Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23085349-23085856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUZP1
Entrez Gene ID
7798
Gene Name
leucine zipper protein 1
Gene Aliases
LUZP
UniGene
Hs.257900
Gene Chromosome Location
Chr.1: 23084017-23177808 on Build GRCh38
Transcripts
NM_001142546.1 NM_033631.3 XM_011542090.2 XM_011542091.2 XM_017002252.1

SNP Detail

SNP ID Chromosome Location
rs997066157 Chr.1: 23085590-23085590 on Build GRCh38
rs558035750 Chr.1: 23085814-23085814 on Build GRCh38
rs977214753 Chr.1: 23085682-23085682 on Build GRCh38
rs979887566 Chr.1: 23085623-23085625 on Build GRCh38
rs950067764 Chr.1: 23085771-23085771 on Build GRCh38
rs758817186 Chr.1: 23085521-23085521 on Build GRCh38
rs925855816 Chr.1: 23085465-23085465 on Build GRCh38
rs914247333 Chr.1: 23085581-23085581 on Build GRCh38
rs752172895 Chr.1: 23085740-23085740 on Build GRCh38
rs139076782 Chr.1: 23085678-23085678 on Build GRCh38
rs780728077 Chr.1: 23085785-23085785 on Build GRCh38
rs764312251 Chr.1: 23085627-23085627 on Build GRCh38
rs940115367 Chr.1: 23085824-23085824 on Build GRCh38
rs367589444 Chr.1: 23085513-23085514 on Build GRCh38
rs935907759 Chr.1: 23085467-23085467 on Build GRCh38
rs911652671 Chr.1: 23085431-23085431 on Build GRCh38
rs967569766 Chr.1: 23085677-23085677 on Build GRCh38
rs757743713 Chr.1: 23085757-23085757 on Build GRCh38
rs868134335 Chr.1: 23085532-23085532 on Build GRCh38
rs749139360 Chr.1: 23085549-23085550 on Build GRCh38
rs901911852 Chr.1: 23085782-23085782 on Build GRCh38
rs568497437 Chr.1: 23085517-23085517 on Build GRCh38
rs887219375 Chr.1: 23085543-23085543 on Build GRCh38
rs1041605965 Chr.1: 23085445-23085445 on Build GRCh38
rs780760288 Chr.1: 23085506-23085508 on Build GRCh38
rs536894053 Chr.1: 23085414-23085414 on Build GRCh38
rs1037283806 Chr.1: 23085706-23085706 on Build GRCh38
rs1044133012 Chr.1: 23085467-23085467 on Build GRCh38
rs997582837 Chr.1: 23085808-23085808 on Build GRCh38
rs887816181 Chr.1: 23085766-23085767 on Build GRCh38
rs925785596 Chr.1: 23085709-23085709 on Build GRCh38
rs138360916 Chr.1: 23085796-23085797 on Build GRCh38
rs887196860 Chr.1: 23085810-23085810 on Build GRCh38
rs953219183 Chr.1: 23085406-23085406 on Build GRCh38
rs897049372 Chr.1: 23085478-23085479 on Build GRCh38
rs561831387 Chr.1: 23085663-23085663 on Build GRCh38
rs867223383 Chr.1: 23085501-23085501 on Build GRCh38
rs112137541 Chr.1: 23085731-23085731 on Build GRCh38
rs987569413 Chr.1: 23085638-23085638 on Build GRCh38
rs1017667644 Chr.1: 23085776-23085776 on Build GRCh38
rs539409672 Chr.1: 23085807-23085807 on Build GRCh38
rs945885327 Chr.1: 23085444-23085444 on Build GRCh38
rs1028591867 Chr.1: 23085607-23085607 on Build GRCh38
rs565547184 Chr.1: 23085530-23085531 on Build GRCh38
rs962898897 Chr.1: 23085565-23085565 on Build GRCh38
rs1055771492 Chr.1: 23085469-23085469 on Build GRCh38
rs752428622 Chr.1: 23085727-23085727 on Build GRCh38
rs921610100 Chr.1: 23085383-23085383 on Build GRCh38
rs764732780 Chr.1: 23085655-23085655 on Build GRCh38
rs1014733753 Chr.1: 23085508-23085508 on Build GRCh38
rs915838213 Chr.1: 23085764-23085764 on Build GRCh38
rs928347622 Chr.1: 23085648-23085654 on Build GRCh38
rs1036972814 Chr.1: 23085470-23085470 on Build GRCh38
rs914216180 Chr.1: 23085431-23085432 on Build GRCh38
rs766492201 Chr.1: 23085480-23085480 on Build GRCh38
rs896962288 Chr.1: 23085710-23085710 on Build GRCh38
rs753673144 Chr.1: 23085514-23085514 on Build GRCh38
rs1017097223 Chr.1: 23085548-23085548 on Build GRCh38
rs911636845 Chr.1: 23085662-23085662 on Build GRCh38
rs146186530 Chr.1: 23085594-23085594 on Build GRCh38
rs549153655 Chr.1: 23085495-23085495 on Build GRCh38
rs1004331713 Chr.1: 23085546-23085546 on Build GRCh38

View Full Product Details