Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23509885-23510325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F2
Entrez Gene ID
1870
Gene Name
E2F transcription factor 2
Gene Aliases
E2F-2
UniGene
Hs.194333
Gene Chromosome Location
Chr.1: 23505696-23531266 on Build GRCh38
Transcripts
NM_004091.3 XM_005245748.3 XM_005245749.3 XM_011540868.2 XM_011540870.2 XM_011540871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320337 c.1121T>A p.L374Q Chr.1: 23510073-23510073 on Build GRCh38
82118 c.1239G>T p.L413L Chr.1: 23509955-23509955 on Build GRCh38
906301 c.1156G>A p.E386K Chr.1: 23510038-23510038 on Build GRCh38
2049175 c.1058C>T p.A353V Chr.1: 23510136-23510136 on Build GRCh38
6329732 c.1188C>T p.C396C Chr.1: 23510006-23510006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142571891 Chr.1: 23509986-23509986 on Build GRCh38
rs1014160601 Chr.1: 23509981-23509981 on Build GRCh38
rs374607130 Chr.1: 23510189-23510189 on Build GRCh38
rs758120108 Chr.1: 23510158-23510158 on Build GRCh38
rs368795371 Chr.1: 23509941-23509941 on Build GRCh38
rs772598051 Chr.1: 23509948-23509948 on Build GRCh38
rs764168899 Chr.1: 23509964-23509964 on Build GRCh38
rs149926753 Chr.1: 23510173-23510173 on Build GRCh38
rs964025725 Chr.1: 23510203-23510203 on Build GRCh38
rs530082478 Chr.1: 23510061-23510061 on Build GRCh38
rs745450429 Chr.1: 23510136-23510136 on Build GRCh38
rs773372814 Chr.1: 23510099-23510099 on Build GRCh38
rs753858882 Chr.1: 23509967-23509967 on Build GRCh38
rs781075661 Chr.1: 23509913-23509913 on Build GRCh38
rs769106531 Chr.1: 23510174-23510174 on Build GRCh38
rs776096433 Chr.1: 23509958-23509958 on Build GRCh38
rs755992777 Chr.1: 23510113-23510113 on Build GRCh38
rs756781681 Chr.1: 23510129-23510129 on Build GRCh38
rs752620979 Chr.1: 23510009-23510009 on Build GRCh38
rs143218447 Chr.1: 23509915-23509915 on Build GRCh38
rs548500891 Chr.1: 23510018-23510018 on Build GRCh38
rs886819893 Chr.1: 23510010-23510010 on Build GRCh38
rs770482896 Chr.1: 23509936-23509936 on Build GRCh38
rs752426258 Chr.1: 23510112-23510112 on Build GRCh38
rs1018300326 Chr.1: 23510199-23510199 on Build GRCh38
rs957123568 Chr.1: 23510206-23510206 on Build GRCh38
rs750682155 Chr.1: 23509977-23509977 on Build GRCh38
rs777245600 Chr.1: 23510066-23510066 on Build GRCh38
rs759951612 Chr.1: 23509943-23509943 on Build GRCh38
rs370898219 Chr.1: 23510013-23510013 on Build GRCh38
rs3218207 Chr.1: 23510204-23510204 on Build GRCh38
rs778584443 Chr.1: 23510135-23510135 on Build GRCh38
rs866938398 Chr.1: 23510094-23510094 on Build GRCh38
rs753848134 Chr.1: 23510127-23510127 on Build GRCh38
rs552752216 Chr.1: 23510218-23510218 on Build GRCh38
rs776962247 Chr.1: 23510176-23510176 on Build GRCh38
rs183643474 Chr.1: 23510050-23510050 on Build GRCh38
rs967839185 Chr.1: 23510236-23510236 on Build GRCh38
rs761768883 Chr.1: 23510074-23510074 on Build GRCh38
rs937994233 Chr.1: 23510249-23510249 on Build GRCh38
rs369246681 Chr.1: 23510081-23510081 on Build GRCh38
rs926264748 Chr.1: 23510055-23510055 on Build GRCh38
rs146493961 Chr.1: 23510271-23510271 on Build GRCh38
rs925413474 Chr.1: 23510224-23510224 on Build GRCh38
rs563073079 Chr.1: 23509963-23509963 on Build GRCh38
rs763913178 Chr.1: 23510115-23510115 on Build GRCh38
rs777160606 Chr.1: 23509925-23509925 on Build GRCh38
rs767232696 Chr.1: 23510111-23510111 on Build GRCh38
rs777829086 Chr.1: 23510017-23510017 on Build GRCh38
rs767859361 Chr.1: 23510274-23510274 on Build GRCh38
rs773284390 Chr.1: 23510195-23510195 on Build GRCh38
rs754726067 Chr.1: 23509979-23509979 on Build GRCh38
rs762013274 Chr.1: 23509969-23509969 on Build GRCh38
rs367755068 Chr.1: 23510160-23510160 on Build GRCh38
rs377589757 Chr.1: 23510205-23510205 on Build GRCh38
rs778672424 Chr.1: 23510022-23510022 on Build GRCh38
rs772511958 Chr.1: 23510059-23510059 on Build GRCh38
rs151287835 Chr.1: 23510060-23510060 on Build GRCh38
rs3218206 Chr.1: 23510273-23510273 on Build GRCh38
rs1049336725 Chr.1: 23510125-23510125 on Build GRCh38
rs915353981 Chr.1: 23510220-23510220 on Build GRCh38
rs775452581 Chr.1: 23510259-23510259 on Build GRCh38
rs374297603 Chr.1: 23509934-23509934 on Build GRCh38
rs765424043 Chr.1: 23509976-23509976 on Build GRCh38
rs748709598 Chr.1: 23510182-23510182 on Build GRCh38
rs763176248 Chr.1: 23510109-23510109 on Build GRCh38
rs570651971 Chr.1: 23510237-23510237 on Build GRCh38
rs1016525076 Chr.1: 23510025-23510025 on Build GRCh38
rs377140105 Chr.1: 23509919-23509919 on Build GRCh38

View Full Product Details