Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23695718-23696215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL11
Entrez Gene ID
6135
Gene Name
ribosomal protein L11
Gene Aliases
DBA7, GIG34, L11
UniGene
Hs.719951
Gene Chromosome Location
Chr.1: 23691779-23696425 on Build GRCh38
Transcripts
NM_000975.3 NM_001199802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919965 c.477A>G p.K159K Chr.1: 23695878-23695878 on Build GRCh38
4934615 c.442A>G p.T148A Chr.1: 23695843-23695843 on Build GRCh38
906316 c.424G>A p.A142T Chr.1: 23695825-23695825 on Build GRCh38
906317 c.437G>A p.R146H Chr.1: 23695838-23695838 on Build GRCh38
7425799 c.406A>T p.R136W Chr.1: 23695807-23695807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005222030 Chr.1: 23696054-23696055 on Build GRCh38
rs373013413 Chr.1: 23696001-23696001 on Build GRCh38
rs561892112 Chr.1: 23696111-23696111 on Build GRCh38
rs947352542 Chr.1: 23696166-23696166 on Build GRCh38
rs758442388 Chr.1: 23696006-23696006 on Build GRCh38
rs201128306 Chr.1: 23695889-23695889 on Build GRCh38
rs919757627 Chr.1: 23696019-23696019 on Build GRCh38
rs753304343 Chr.1: 23695831-23695833 on Build GRCh38
rs971153514 Chr.1: 23696149-23696149 on Build GRCh38
rs2280437 Chr.1: 23695947-23695947 on Build GRCh38
rs751490105 Chr.1: 23695842-23695842 on Build GRCh38
rs760198106 Chr.1: 23696005-23696007 on Build GRCh38
rs963822319 Chr.1: 23696090-23696090 on Build GRCh38
rs902799494 Chr.1: 23696076-23696076 on Build GRCh38
rs11556040 Chr.1: 23695836-23695836 on Build GRCh38
rs891140034 Chr.1: 23696058-23696058 on Build GRCh38
rs770776620 Chr.1: 23695838-23695838 on Build GRCh38
rs772274492 Chr.1: 23695752-23695752 on Build GRCh38
rs1015250131 Chr.1: 23696171-23696171 on Build GRCh38
rs754966145 Chr.1: 23695853-23695853 on Build GRCh38
rs879056713 Chr.1: 23695909-23695909 on Build GRCh38
rs1001063696 Chr.1: 23695837-23695837 on Build GRCh38
rs895284727 Chr.1: 23696101-23696101 on Build GRCh38
rs1032514832 Chr.1: 23696089-23696089 on Build GRCh38
rs587777119 Chr.1: 23695877-23695878 on Build GRCh38
rs542148988 Chr.1: 23695750-23695750 on Build GRCh38
rs905961633 Chr.1: 23695978-23695978 on Build GRCh38
rs1018002665 Chr.1: 23696071-23696071 on Build GRCh38
rs867427651 Chr.1: 23695924-23695924 on Build GRCh38
rs989049618 Chr.1: 23696139-23696139 on Build GRCh38
rs775683622 Chr.1: 23695959-23695959 on Build GRCh38
rs3208746 Chr.1: 23695879-23695879 on Build GRCh38
rs369530329 Chr.1: 23695873-23695873 on Build GRCh38
rs779487276 Chr.1: 23695942-23695942 on Build GRCh38
rs1023114482 Chr.1: 23696118-23696118 on Build GRCh38
rs760371367 Chr.1: 23695767-23695767 on Build GRCh38
rs771421542 Chr.1: 23695747-23695747 on Build GRCh38
rs981541343 Chr.1: 23696162-23696162 on Build GRCh38
rs145233387 Chr.1: 23695917-23695917 on Build GRCh38
rs371228733 Chr.1: 23695794-23695794 on Build GRCh38
rs1057000026 Chr.1: 23695985-23695985 on Build GRCh38
rs368426196 Chr.1: 23695751-23695751 on Build GRCh38
rs749610453 Chr.1: 23695892-23695892 on Build GRCh38
rs531857888 Chr.1: 23696091-23696091 on Build GRCh38
rs762667446 Chr.1: 23695808-23695808 on Build GRCh38
rs878927531 Chr.1: 23695910-23695910 on Build GRCh38
rs777724022 Chr.1: 23696021-23696021 on Build GRCh38
rs991197389 Chr.1: 23695877-23695877 on Build GRCh38
rs11556033 Chr.1: 23695902-23695902 on Build GRCh38
rs895720837 Chr.1: 23695989-23695989 on Build GRCh38
rs114817969 Chr.1: 23695993-23695993 on Build GRCh38
rs1032506542 Chr.1: 23695845-23695845 on Build GRCh38
rs932738841 Chr.1: 23696020-23696020 on Build GRCh38
rs1804303 Chr.1: 23695823-23695823 on Build GRCh38
rs777313952 Chr.1: 23695891-23695891 on Build GRCh38
rs1030521816 Chr.1: 23696109-23696109 on Build GRCh38
rs974903869 Chr.1: 23696103-23696103 on Build GRCh38
rs764265677 Chr.1: 23695793-23695793 on Build GRCh38
rs1025994135 Chr.1: 23696017-23696017 on Build GRCh38
rs1008620624 Chr.1: 23696062-23696062 on Build GRCh38
rs978742399 Chr.1: 23696173-23696173 on Build GRCh38
rs879099403 Chr.1: 23696069-23696069 on Build GRCh38
rs766991539 Chr.1: 23695855-23695855 on Build GRCh38
rs755941072 Chr.1: 23695770-23695770 on Build GRCh38
rs766268959 Chr.1: 23695825-23695825 on Build GRCh38
rs543615416 Chr.1: 23696092-23696092 on Build GRCh38
rs763112075 Chr.1: 23696093-23696093 on Build GRCh38
rs200545084 Chr.1: 23695981-23695981 on Build GRCh38
rs760931298 Chr.1: 23695781-23695781 on Build GRCh38
rs913262496 Chr.1: 23695839-23695839 on Build GRCh38
rs1001115443 Chr.1: 23696084-23696084 on Build GRCh38
rs747513965 Chr.1: 23696033-23696033 on Build GRCh38
rs757566843 Chr.1: 23695916-23695916 on Build GRCh38

View Full Product Details