Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23743176-23743714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEB3
Entrez Gene ID
6924
Gene Name
transcription elongation factor B subunit 3
Gene Aliases
EloA, SIII, SIII p110, TCEB3A
UniGene
Hs.15535
Gene Chromosome Location
Chr.1: 23743366-23762059 on Build GRCh38
Transcripts
NM_003198.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403168 c.29T>C p.V10A Chr.1: 23743532-23743532 on Build GRCh38
6403167 c.107T>C p.V36A Chr.1: 23743532-23743532 on Build GRCh38
5723442 c.72G>C p.V24V Chr.1: 23743497-23743497 on Build GRCh38
6412322 c.28G>T p.V10L Chr.1: 23743531-23743531 on Build GRCh38
6412321 c.106G>T p.V36L Chr.1: 23743531-23743531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567747723 Chr.1: 23743288-23743288 on Build GRCh38
rs939529796 Chr.1: 23743491-23743491 on Build GRCh38
rs768736517 Chr.1: 23743395-23743395 on Build GRCh38
rs1031574839 Chr.1: 23743657-23743657 on Build GRCh38
rs965987143 Chr.1: 23743241-23743241 on Build GRCh38
rs1010042747 Chr.1: 23743293-23743293 on Build GRCh38
rs367781237 Chr.1: 23743454-23743454 on Build GRCh38
rs956183050 Chr.1: 23743660-23743660 on Build GRCh38
rs917451406 Chr.1: 23743370-23743370 on Build GRCh38
rs755555300 Chr.1: 23743595-23743595 on Build GRCh38
rs924266639 Chr.1: 23743259-23743260 on Build GRCh38
rs993926550 Chr.1: 23743332-23743332 on Build GRCh38
rs560571710 Chr.1: 23743687-23743687 on Build GRCh38
rs905448509 Chr.1: 23743322-23743322 on Build GRCh38
rs772440540 Chr.1: 23743503-23743503 on Build GRCh38
rs778839375 Chr.1: 23743464-23743464 on Build GRCh38
rs545679389 Chr.1: 23743207-23743207 on Build GRCh38
rs930146296 Chr.1: 23743371-23743371 on Build GRCh38
rs1056697768 Chr.1: 23743496-23743496 on Build GRCh38
rs968792465 Chr.1: 23743313-23743313 on Build GRCh38
rs975996133 Chr.1: 23743375-23743376 on Build GRCh38
rs1007391353 Chr.1: 23743347-23743348 on Build GRCh38
rs553458405 Chr.1: 23743307-23743307 on Build GRCh38
rs914345855 Chr.1: 23743272-23743272 on Build GRCh38
rs765016806 Chr.1: 23743514-23743514 on Build GRCh38
rs1005847874 Chr.1: 23743233-23743233 on Build GRCh38
rs992811329 Chr.1: 23743596-23743596 on Build GRCh38
rs904640928 Chr.1: 23743566-23743566 on Build GRCh38
rs563082501 Chr.1: 23743222-23743222 on Build GRCh38
rs907627327 Chr.1: 23743396-23743396 on Build GRCh38
rs951630182 Chr.1: 23743664-23743664 on Build GRCh38
rs886159391 Chr.1: 23743230-23743230 on Build GRCh38
rs761529812 Chr.1: 23743509-23743509 on Build GRCh38
rs997782756 Chr.1: 23743656-23743656 on Build GRCh38
rs901652489 Chr.1: 23743609-23743609 on Build GRCh38
rs780358880 Chr.1: 23743254-23743254 on Build GRCh38
rs535057141 Chr.1: 23743301-23743301 on Build GRCh38
rs556867091 Chr.1: 23743468-23743468 on Build GRCh38
rs577352375 Chr.1: 23743438-23743438 on Build GRCh38
rs989892780 Chr.1: 23743261-23743261 on Build GRCh38
rs12086741 Chr.1: 23743472-23743472 on Build GRCh38
rs565392243 Chr.1: 23743276-23743276 on Build GRCh38
rs774092119 Chr.1: 23743463-23743463 on Build GRCh38
rs780716634 Chr.1: 23743419-23743419 on Build GRCh38
rs934332105 Chr.1: 23743260-23743260 on Build GRCh38
rs993059803 Chr.1: 23743368-23743368 on Build GRCh38
rs990249156 Chr.1: 23743452-23743452 on Build GRCh38
rs1037297728 Chr.1: 23743238-23743238 on Build GRCh38
rs775294825 Chr.1: 23743477-23743477 on Build GRCh38
rs939564635 Chr.1: 23743324-23743324 on Build GRCh38
rs1045240016 Chr.1: 23743277-23743277 on Build GRCh38
rs539514265 Chr.1: 23743367-23743367 on Build GRCh38
rs1010486691 Chr.1: 23743498-23743498 on Build GRCh38
rs1049125525 Chr.1: 23743587-23743587 on Build GRCh38
rs927269786 Chr.1: 23743326-23743326 on Build GRCh38
rs192950496 Chr.1: 23743558-23743558 on Build GRCh38
rs917401666 Chr.1: 23743201-23743201 on Build GRCh38
rs978765010 Chr.1: 23743317-23743317 on Build GRCh38
rs760734858 Chr.1: 23743490-23743490 on Build GRCh38
rs1024993823 Chr.1: 23743659-23743659 on Build GRCh38
rs185512297 Chr.1: 23743594-23743594 on Build GRCh38
rs780628476 Chr.1: 23743561-23743561 on Build GRCh38
rs749506630 Chr.1: 23743315-23743315 on Build GRCh38
rs897798710 Chr.1: 23743331-23743331 on Build GRCh38
rs750771570 Chr.1: 23743554-23743554 on Build GRCh38
rs763500834 Chr.1: 23743495-23743495 on Build GRCh38
rs900462603 Chr.1: 23743248-23743248 on Build GRCh38
rs558923580 Chr.1: 23743372-23743372 on Build GRCh38
rs887891208 Chr.1: 23743342-23743342 on Build GRCh38
rs887860069 Chr.1: 23743588-23743588 on Build GRCh38
rs992454153 Chr.1: 23743661-23743661 on Build GRCh38
rs747040396 Chr.1: 23743422-23743422 on Build GRCh38
rs1020081769 Chr.1: 23743518-23743518 on Build GRCh38
rs1047202145 Chr.1: 23743377-23743377 on Build GRCh38
rs748304702 Chr.1: 23743502-23743502 on Build GRCh38
rs996048112 Chr.1: 23743253-23743253 on Build GRCh38
rs1032024143 Chr.1: 23743381-23743381 on Build GRCh38
rs983391519 Chr.1: 23743667-23743667 on Build GRCh38
rs565589815 Chr.1: 23743334-23743334 on Build GRCh38
rs1030189465 Chr.1: 23743271-23743271 on Build GRCh38
rs1000278163 Chr.1: 23743570-23743570 on Build GRCh38
rs1021204505 Chr.1: 23743676-23743676 on Build GRCh38
rs941779064 Chr.1: 23743451-23743451 on Build GRCh38
rs949186264 Chr.1: 23743214-23743214 on Build GRCh38
rs748295525 Chr.1: 23743460-23743460 on Build GRCh38

View Full Product Details