Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23792856-23793353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPLA2
Entrez Gene ID
11313
Gene Name
lysophospholipase II
Gene Aliases
APT-2, APT2, DJ886K2.4
UniGene
Hs.533479
Gene Chromosome Location
Chr.1: 23790789-23795539 on Build GRCh38
Transcripts
NM_007260.2 XM_005245728.4 XM_017000165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5581590 c.130C>A p.L44I Chr.1: 23793170-23793170 on Build GRCh38
5581591 c.130C>A p.L44I Chr.1: 23793170-23793170 on Build GRCh38
5926276 c.148C>T p.P50S Chr.1: 23793188-23793188 on Build GRCh38
6705334 c.105C>T p.D35D Chr.1: 23793034-23793034 on Build GRCh38
2049816 c.83delT p.L30fs*35 Chr.1: 23793012-23793012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756191714 Chr.1: 23792976-23792976 on Build GRCh38
rs754736273 Chr.1: 23793109-23793109 on Build GRCh38
rs775862639 Chr.1: 23793212-23793212 on Build GRCh38
rs931883828 Chr.1: 23793184-23793184 on Build GRCh38
rs904531463 Chr.1: 23792981-23792981 on Build GRCh38
rs777462448 Chr.1: 23793054-23793055 on Build GRCh38
rs369751419 Chr.1: 23793124-23793124 on Build GRCh38
rs371527090 Chr.1: 23792986-23792986 on Build GRCh38
rs370129638 Chr.1: 23793220-23793220 on Build GRCh38
rs778590879 Chr.1: 23793194-23793194 on Build GRCh38
rs370052150 Chr.1: 23793244-23793245 on Build GRCh38
rs372601977 Chr.1: 23792963-23792963 on Build GRCh38
rs992745530 Chr.1: 23792971-23792971 on Build GRCh38
rs2232979 Chr.1: 23792893-23792893 on Build GRCh38
rs1020334005 Chr.1: 23793120-23793120 on Build GRCh38
rs1010199342 Chr.1: 23793119-23793119 on Build GRCh38
rs1024528714 Chr.1: 23793326-23793326 on Build GRCh38
rs746815475 Chr.1: 23792999-23792999 on Build GRCh38
rs753864890 Chr.1: 23793101-23793101 on Build GRCh38
rs1042206246 Chr.1: 23793263-23793263 on Build GRCh38
rs1034139289 Chr.1: 23792935-23792935 on Build GRCh38
rs757274144 Chr.1: 23793112-23793112 on Build GRCh38
rs958536435 Chr.1: 23793144-23793144 on Build GRCh38
rs759975979 Chr.1: 23793070-23793070 on Build GRCh38
rs374787618 Chr.1: 23793005-23793005 on Build GRCh38
rs752344350 Chr.1: 23792968-23792969 on Build GRCh38
rs968526176 Chr.1: 23792925-23792925 on Build GRCh38
rs537012768 Chr.1: 23792987-23792987 on Build GRCh38
rs771859306 Chr.1: 23793211-23793211 on Build GRCh38
rs574024225 Chr.1: 23793241-23793242 on Build GRCh38
rs2232980 Chr.1: 23792970-23792970 on Build GRCh38
rs757031249 Chr.1: 23793106-23793106 on Build GRCh38
rs368040980 Chr.1: 23793183-23793183 on Build GRCh38
rs2232981 Chr.1: 23792983-23792983 on Build GRCh38
rs755891364 Chr.1: 23793050-23793051 on Build GRCh38
rs978532877 Chr.1: 23792930-23792930 on Build GRCh38
rs371361178 Chr.1: 23793122-23793122 on Build GRCh38
rs376835219 Chr.1: 23793123-23793123 on Build GRCh38
rs768125088 Chr.1: 23792973-23792973 on Build GRCh38
rs767259355 Chr.1: 23793176-23793176 on Build GRCh38
rs762354950 Chr.1: 23793242-23793242 on Build GRCh38
rs529404899 Chr.1: 23793105-23793105 on Build GRCh38
rs570634166 Chr.1: 23793142-23793142 on Build GRCh38
rs774117027 Chr.1: 23793130-23793130 on Build GRCh38
rs922108622 Chr.1: 23793299-23793299 on Build GRCh38
rs890473453 Chr.1: 23793110-23793110 on Build GRCh38
rs777568986 Chr.1: 23793186-23793186 on Build GRCh38
rs747318475 Chr.1: 23793228-23793228 on Build GRCh38
rs551905424 Chr.1: 23793073-23793073 on Build GRCh38
rs757440115 Chr.1: 23793103-23793103 on Build GRCh38
rs748075123 Chr.1: 23793037-23793037 on Build GRCh38
rs749069180 Chr.1: 23793294-23793294 on Build GRCh38
rs945865912 Chr.1: 23793227-23793227 on Build GRCh38
rs766716526 Chr.1: 23792969-23792969 on Build GRCh38
rs761873997 Chr.1: 23793155-23793155 on Build GRCh38
rs374788476 Chr.1: 23793215-23793215 on Build GRCh38
rs764518777 Chr.1: 23793140-23793140 on Build GRCh38
rs777767043 Chr.1: 23792977-23792977 on Build GRCh38
rs199697474 Chr.1: 23793219-23793219 on Build GRCh38
rs1000219306 Chr.1: 23793275-23793275 on Build GRCh38
rs894761232 Chr.1: 23793303-23793303 on Build GRCh38
rs765340005 Chr.1: 23793157-23793157 on Build GRCh38
rs770722930 Chr.1: 23793061-23793061 on Build GRCh38
rs753762946 Chr.1: 23793104-23793105 on Build GRCh38
rs2232982 Chr.1: 23793137-23793137 on Build GRCh38
rs1024883737 Chr.1: 23793094-23793094 on Build GRCh38
rs775750902 Chr.1: 23793138-23793138 on Build GRCh38
rs550681434 Chr.1: 23792882-23792882 on Build GRCh38
rs774204993 Chr.1: 23793063-23793063 on Build GRCh38
rs563036143 Chr.1: 23792883-23792883 on Build GRCh38
rs137894016 Chr.1: 23793166-23793166 on Build GRCh38
rs769129056 Chr.1: 23793231-23793231 on Build GRCh38
rs368623413 Chr.1: 23792958-23792958 on Build GRCh38
rs758642203 Chr.1: 23793173-23793173 on Build GRCh38
rs779754947 Chr.1: 23792994-23792994 on Build GRCh38
rs759315333 Chr.1: 23793135-23793135 on Build GRCh38
rs369248220 Chr.1: 23793050-23793050 on Build GRCh38
rs371799001 Chr.1: 23793193-23793193 on Build GRCh38
rs963034088 Chr.1: 23793148-23793148 on Build GRCh38
rs768494389 Chr.1: 23793001-23793001 on Build GRCh38
rs542626512 Chr.1: 23793112-23793112 on Build GRCh38
rs770632972 Chr.1: 23793127-23793127 on Build GRCh38
rs772617928 Chr.1: 23793053-23793053 on Build GRCh38
rs776970224 Chr.1: 23793239-23793239 on Build GRCh38
rs1020302800 Chr.1: 23792899-23792899 on Build GRCh38
rs745379103 Chr.1: 23793182-23793182 on Build GRCh38
rs768301834 Chr.1: 23793320-23793320 on Build GRCh38
rs753237290 Chr.1: 23793090-23793090 on Build GRCh38
rs762601559 Chr.1: 23793058-23793058 on Build GRCh38
rs112266814 Chr.1: 23792974-23792974 on Build GRCh38
rs376858572 Chr.1: 23793092-23793092 on Build GRCh38
rs932148995 Chr.1: 23793305-23793305 on Build GRCh38
rs377182145 Chr.1: 23793091-23793091 on Build GRCh38
rs745481334 Chr.1: 23793196-23793196 on Build GRCh38
rs189482841 Chr.1: 23793273-23793273 on Build GRCh38

View Full Product Details