Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24128063-24128577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA1
Entrez Gene ID
58985
Gene Name
interleukin 22 receptor subunit alpha 1
Gene Aliases
CRF2-9, IL22R, IL22R1
UniGene
Hs.110915
Gene Chromosome Location
Chr.1: 24119771-24143285 on Build GRCh38
Transcripts
NM_021258.3 XM_011541882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5977922 c.568G>A p.G190S Chr.1: 24128243-24128243 on Build GRCh38
3486380 c.651C>T p.C217C Chr.1: 24128160-24128160 on Build GRCh38
3934607 c.603C>T p.I201I Chr.1: 24128208-24128208 on Build GRCh38
5380306 c.553_554delGA p.E185fs*2 Chr.1: 24128257-24128258 on Build GRCh38
6125544 c.655G>T p.V219L Chr.1: 24128156-24128156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780179419 Chr.1: 24128263-24128263 on Build GRCh38
rs200299551 Chr.1: 24128530-24128530 on Build GRCh38
rs149743170 Chr.1: 24128530-24128531 on Build GRCh38
rs894940984 Chr.1: 24128105-24128105 on Build GRCh38
rs545237896 Chr.1: 24128353-24128353 on Build GRCh38
rs916374453 Chr.1: 24128473-24128473 on Build GRCh38
rs376051408 Chr.1: 24128101-24128101 on Build GRCh38
rs1053805100 Chr.1: 24128548-24128548 on Build GRCh38
rs761900564 Chr.1: 24128321-24128321 on Build GRCh38
rs538351492 Chr.1: 24128476-24128476 on Build GRCh38
rs537373224 Chr.1: 24128289-24128289 on Build GRCh38
rs758996710 Chr.1: 24128401-24128401 on Build GRCh38
rs770651348 Chr.1: 24128241-24128241 on Build GRCh38
rs774713240 Chr.1: 24128148-24128148 on Build GRCh38
rs200830098 Chr.1: 24128311-24128311 on Build GRCh38
rs753981262 Chr.1: 24128171-24128171 on Build GRCh38
rs777545837 Chr.1: 24128175-24128175 on Build GRCh38
rs756658665 Chr.1: 24128168-24128168 on Build GRCh38
rs184662831 Chr.1: 24128471-24128471 on Build GRCh38
rs776453511 Chr.1: 24128114-24128114 on Build GRCh38
rs1014585360 Chr.1: 24128147-24128147 on Build GRCh38
rs754069658 Chr.1: 24128318-24128318 on Build GRCh38
rs201360269 Chr.1: 24128132-24128132 on Build GRCh38
rs985614025 Chr.1: 24128212-24128212 on Build GRCh38
rs56091453 Chr.1: 24128539-24128540 on Build GRCh38
rs978163283 Chr.1: 24128526-24128527 on Build GRCh38
rs761162614 Chr.1: 24128292-24128292 on Build GRCh38
rs985690344 Chr.1: 24128138-24128138 on Build GRCh38
rs79895649 Chr.1: 24128412-24128412 on Build GRCh38
rs201644800 Chr.1: 24128133-24128133 on Build GRCh38
rs920187839 Chr.1: 24128187-24128187 on Build GRCh38
rs41268129 Chr.1: 24128322-24128322 on Build GRCh38
rs947759244 Chr.1: 24128489-24128489 on Build GRCh38
rs944120990 Chr.1: 24128229-24128229 on Build GRCh38
rs751316829 Chr.1: 24128183-24128183 on Build GRCh38
rs757399703 Chr.1: 24128174-24128174 on Build GRCh38
rs948282699 Chr.1: 24128256-24128256 on Build GRCh38
rs772297728 Chr.1: 24128247-24128247 on Build GRCh38
rs548872112 Chr.1: 24128399-24128399 on Build GRCh38
rs963041249 Chr.1: 24128410-24128410 on Build GRCh38
rs769910295 Chr.1: 24128199-24128199 on Build GRCh38
rs1014950031 Chr.1: 24128312-24128312 on Build GRCh38
rs912743387 Chr.1: 24128225-24128225 on Build GRCh38
rs748253442 Chr.1: 24128532-24128533 on Build GRCh38
rs35240302 Chr.1: 24128244-24128244 on Build GRCh38
rs34379702 Chr.1: 24128186-24128186 on Build GRCh38
rs752675167 Chr.1: 24128111-24128111 on Build GRCh38
rs767615299 Chr.1: 24128168-24128169 on Build GRCh38
rs748813374 Chr.1: 24128239-24128239 on Build GRCh38
rs1012094554 Chr.1: 24128112-24128112 on Build GRCh38
rs397814093 Chr.1: 24128549-24128550 on Build GRCh38
rs764141100 Chr.1: 24128169-24128169 on Build GRCh38
rs916612843 Chr.1: 24128548-24128549 on Build GRCh38
rs16829204 Chr.1: 24128198-24128198 on Build GRCh38
rs1029112328 Chr.1: 24128346-24128346 on Build GRCh38
rs188885194 Chr.1: 24128414-24128414 on Build GRCh38
rs192222983 Chr.1: 24128457-24128457 on Build GRCh38
rs768023224 Chr.1: 24128159-24128159 on Build GRCh38
rs544118695 Chr.1: 24128209-24128209 on Build GRCh38
rs746782860 Chr.1: 24128103-24128103 on Build GRCh38
rs755033835 Chr.1: 24128358-24128358 on Build GRCh38
rs199754892 Chr.1: 24128119-24128119 on Build GRCh38
rs995232634 Chr.1: 24128334-24128334 on Build GRCh38
rs928092256 Chr.1: 24128550-24128550 on Build GRCh38
rs954274006 Chr.1: 24128458-24128458 on Build GRCh38
rs965971225 Chr.1: 24128511-24128512 on Build GRCh38
rs1027167870 Chr.1: 24128419-24128419 on Build GRCh38
rs764855153 Chr.1: 24128104-24128104 on Build GRCh38
rs71814307 Chr.1: 24128540-24128541 on Build GRCh38
rs769397737 Chr.1: 24128131-24128131 on Build GRCh38
rs781079339 Chr.1: 24128191-24128191 on Build GRCh38
rs34782294 Chr.1: 24128146-24128146 on Build GRCh38
rs924030052 Chr.1: 24128528-24128529 on Build GRCh38
rs540310802 Chr.1: 24128546-24128549 on Build GRCh38
rs61775472 Chr.1: 24128264-24128264 on Build GRCh38
rs1015090033 Chr.1: 24128404-24128404 on Build GRCh38
rs144915672 Chr.1: 24128158-24128158 on Build GRCh38
rs779014817 Chr.1: 24128181-24128181 on Build GRCh38
rs546260555 Chr.1: 24128463-24128463 on Build GRCh38
rs780524878 Chr.1: 24128472-24128472 on Build GRCh38
rs912724614 Chr.1: 24128499-24128499 on Build GRCh38
rs185047620 Chr.1: 24128243-24128243 on Build GRCh38
rs61560438 Chr.1: 24128376-24128376 on Build GRCh38

View Full Product Details