Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24360709-24361202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Gene Symbol
STPG1
Entrez Gene ID
90529
Gene Name
sperm tail PG-rich repeat containing 1
Gene Aliases
C1orf201, MAPO2
UniGene
Hs.403187
Gene Chromosome Location
Chr.1: 24356999-24415533 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3486509 c.887G>T p.R296L Chr.1: 24360892-24360892 on Build GRCh38
5458382 c.604C>T p.P202S Chr.1: 24361034-24361034 on Build GRCh38
906868 c.760C>G p.P254A Chr.1: 24360878-24360878 on Build GRCh38
1745252 c.765_784del20 p.Q256fs*>26 Chr.1: 24360854-24360873 on Build GRCh38
5899339 c.866C>T p.S289L Chr.1: 24360913-24360913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757594954 Chr.1: 24360893-24360893 on Build GRCh38
rs868621625 Chr.1: 24360935-24360936 on Build GRCh38
rs142046508 Chr.1: 24360931-24360931 on Build GRCh38
rs532555701 Chr.1: 24361005-24361005 on Build GRCh38
rs777707045 Chr.1: 24361023-24361023 on Build GRCh38
rs373923229 Chr.1: 24360920-24360920 on Build GRCh38
rs983092920 Chr.1: 24361158-24361158 on Build GRCh38
rs34154883 Chr.1: 24360869-24360869 on Build GRCh38
rs1033732518 Chr.1: 24360852-24360852 on Build GRCh38
rs376956450 Chr.1: 24360938-24360938 on Build GRCh38
rs929012560 Chr.1: 24361095-24361095 on Build GRCh38
rs11249094 Chr.1: 24361182-24361182 on Build GRCh38
rs997779200 Chr.1: 24360796-24360796 on Build GRCh38
rs765483377 Chr.1: 24361081-24361081 on Build GRCh38
rs376495615 Chr.1: 24361066-24361066 on Build GRCh38
rs560627804 Chr.1: 24360865-24360865 on Build GRCh38
rs551473023 Chr.1: 24361013-24361013 on Build GRCh38
rs374239929 Chr.1: 24360968-24360968 on Build GRCh38
rs533375823 Chr.1: 24361036-24361036 on Build GRCh38
rs750618882 Chr.1: 24360983-24360983 on Build GRCh38
rs141586885 Chr.1: 24360951-24360951 on Build GRCh38
rs778622982 Chr.1: 24361034-24361034 on Build GRCh38
rs775758927 Chr.1: 24361061-24361061 on Build GRCh38
rs376548001 Chr.1: 24360832-24360832 on Build GRCh38
rs909689321 Chr.1: 24360908-24360908 on Build GRCh38
rs532147 Chr.1: 24361163-24361163 on Build GRCh38
rs866287688 Chr.1: 24360934-24360935 on Build GRCh38
rs182549082 Chr.1: 24360853-24360853 on Build GRCh38
rs548395528 Chr.1: 24361129-24361129 on Build GRCh38
rs145821697 Chr.1: 24360824-24360824 on Build GRCh38
rs376523924 Chr.1: 24360804-24360804 on Build GRCh38
rs769337400 Chr.1: 24360917-24360917 on Build GRCh38
rs375637757 Chr.1: 24361074-24361074 on Build GRCh38
rs926445672 Chr.1: 24361055-24361055 on Build GRCh38
rs536585788 Chr.1: 24361060-24361060 on Build GRCh38
rs1002347142 Chr.1: 24361003-24361003 on Build GRCh38
rs529236101 Chr.1: 24361111-24361111 on Build GRCh38
rs139547956 Chr.1: 24361009-24361009 on Build GRCh38
rs138744711 Chr.1: 24360881-24360881 on Build GRCh38
rs748302130 Chr.1: 24360815-24360815 on Build GRCh38
rs769392979 Chr.1: 24361097-24361097 on Build GRCh38
rs748824680 Chr.1: 24360922-24360922 on Build GRCh38
rs776459539 Chr.1: 24360857-24360857 on Build GRCh38
rs777291870 Chr.1: 24361067-24361067 on Build GRCh38
rs761134513 Chr.1: 24361062-24361062 on Build GRCh38
rs763364337 Chr.1: 24360965-24360968 on Build GRCh38
rs773614850 Chr.1: 24360937-24360938 on Build GRCh38
rs776050688 Chr.1: 24360949-24360949 on Build GRCh38
rs747319327 Chr.1: 24360937-24360937 on Build GRCh38
rs765501672 Chr.1: 24360970-24360970 on Build GRCh38
rs78286970 Chr.1: 24360830-24360830 on Build GRCh38
rs747640321 Chr.1: 24360858-24360858 on Build GRCh38
rs946593350 Chr.1: 24360837-24360837 on Build GRCh38
rs564024150 Chr.1: 24360892-24360892 on Build GRCh38
rs781122156 Chr.1: 24360909-24360909 on Build GRCh38
rs900044051 Chr.1: 24361139-24361139 on Build GRCh38
rs964116603 Chr.1: 24360766-24360766 on Build GRCh38
rs34378136 Chr.1: 24361059-24361060 on Build GRCh38
rs754801122 Chr.1: 24360993-24360993 on Build GRCh38
rs774655402 Chr.1: 24360879-24360879 on Build GRCh38
rs372204694 Chr.1: 24361006-24361006 on Build GRCh38
rs751943695 Chr.1: 24360880-24360880 on Build GRCh38
rs899969119 Chr.1: 24360936-24360936 on Build GRCh38
rs998293958 Chr.1: 24361157-24361157 on Build GRCh38
rs772384019 Chr.1: 24360940-24360940 on Build GRCh38
rs375583064 Chr.1: 24361059-24361059 on Build GRCh38
rs386629580 Chr.1: 24361163-24361164 on Build GRCh38
rs1018339461 Chr.1: 24360761-24360761 on Build GRCh38
rs75509295 Chr.1: 24361164-24361164 on Build GRCh38
rs779308505 Chr.1: 24360899-24360899 on Build GRCh38
rs941616665 Chr.1: 24361135-24361135 on Build GRCh38
rs575719376 Chr.1: 24360907-24360907 on Build GRCh38
rs749586874 Chr.1: 24360826-24360826 on Build GRCh38
rs899496020 Chr.1: 24360768-24360768 on Build GRCh38
rs1022725095 Chr.1: 24361173-24361173 on Build GRCh38
rs1029286889 Chr.1: 24360797-24360797 on Build GRCh38
rs956916936 Chr.1: 24360827-24360827 on Build GRCh38
rs1022270796 Chr.1: 24360978-24360978 on Build GRCh38
rs375050484 Chr.1: 24360882-24360882 on Build GRCh38
rs199849506 Chr.1: 24360957-24360957 on Build GRCh38
rs771234785 Chr.1: 24360970-24360971 on Build GRCh38
rs369485283 Chr.1: 24360848-24360848 on Build GRCh38
rs1039927601 Chr.1: 24361137-24361137 on Build GRCh38
rs183353247 Chr.1: 24360886-24360886 on Build GRCh38
rs770107500 Chr.1: 24360936-24360937 on Build GRCh38
rs528552913 Chr.1: 24360956-24360956 on Build GRCh38
rs950772779 Chr.1: 24361125-24361125 on Build GRCh38
rs557233910 Chr.1: 24360757-24360757 on Build GRCh38
rs762640146 Chr.1: 24360876-24360876 on Build GRCh38
rs748103478 Chr.1: 24360915-24360915 on Build GRCh38
rs766695337 Chr.1: 24360877-24360877 on Build GRCh38
rs982760173 Chr.1: 24360888-24360888 on Build GRCh38
rs142852156 Chr.1: 24361033-24361033 on Build GRCh38
rs968265981 Chr.1: 24360829-24360829 on Build GRCh38
rs975181652 Chr.1: 24361168-24361168 on Build GRCh38
rs565096415 Chr.1: 24360994-24360994 on Build GRCh38
rs191866200 Chr.1: 24360883-24360883 on Build GRCh38
rs759978684 Chr.1: 24361085-24361085 on Build GRCh38

View Full Product Details