Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72289835-72290333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF6
Entrez Gene ID
60677
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
BRUNOL6
UniGene
Hs.348342
Gene Chromosome Location
Chr.15: 72284727-72320184 on Build GRCh38
Transcripts
NM_001172684.1 NM_001172685.1 NM_052840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040631 c.453A>T p.R151S Chr.15: 72290197-72290197 on Build GRCh38
6209045 c.585C>T p.H195H Chr.15: 72289957-72289957 on Build GRCh38
6414497 c.563A>G p.Q188R Chr.15: 72289979-72289979 on Build GRCh38
5383784 c.463C>T p.Q155* Chr.15: 72290187-72290187 on Build GRCh38
4671432 c.172G>A p.G58S Chr.15: 72290139-72290139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995626335 Chr.15: 72289930-72289930 on Build GRCh38
rs149269869 Chr.15: 72290278-72290278 on Build GRCh38
rs751848232 Chr.15: 72290216-72290216 on Build GRCh38
rs747838355 Chr.15: 72289931-72289931 on Build GRCh38
rs954091177 Chr.15: 72289933-72289933 on Build GRCh38
rs201448755 Chr.15: 72289976-72289976 on Build GRCh38
rs200747208 Chr.15: 72290257-72290257 on Build GRCh38
rs1020144695 Chr.15: 72289986-72289986 on Build GRCh38
rs758048583 Chr.15: 72290280-72290280 on Build GRCh38
rs547665260 Chr.15: 72289936-72289936 on Build GRCh38
rs144738598 Chr.15: 72289975-72289975 on Build GRCh38
rs199610139 Chr.15: 72290258-72290258 on Build GRCh38
rs374757353 Chr.15: 72290035-72290035 on Build GRCh38
rs766387195 Chr.15: 72289965-72289965 on Build GRCh38
rs1015829123 Chr.15: 72289895-72289895 on Build GRCh38
rs768281082 Chr.15: 72289901-72289901 on Build GRCh38
rs751418072 Chr.15: 72290024-72290025 on Build GRCh38
rs757661728 Chr.15: 72290045-72290045 on Build GRCh38
rs771470304 Chr.15: 72290203-72290203 on Build GRCh38
rs759524400 Chr.15: 72290158-72290158 on Build GRCh38
rs747702531 Chr.15: 72290109-72290109 on Build GRCh38
rs773815459 Chr.15: 72289907-72289907 on Build GRCh38
rs760595545 Chr.15: 72289964-72289964 on Build GRCh38
rs973603279 Chr.15: 72289946-72289946 on Build GRCh38
rs200622052 Chr.15: 72290260-72290260 on Build GRCh38
rs537650831 Chr.15: 72290139-72290139 on Build GRCh38
rs917313145 Chr.15: 72290106-72290106 on Build GRCh38
rs374506900 Chr.15: 72290115-72290115 on Build GRCh38
rs202152101 Chr.15: 72290201-72290202 on Build GRCh38
rs777553263 Chr.15: 72290299-72290299 on Build GRCh38
rs566167445 Chr.15: 72289943-72289943 on Build GRCh38
rs754377086 Chr.15: 72290059-72290059 on Build GRCh38
rs868080974 Chr.15: 72289998-72289998 on Build GRCh38
rs567305506 Chr.15: 72290087-72290087 on Build GRCh38
rs927128902 Chr.15: 72290239-72290239 on Build GRCh38
rs375713379 Chr.15: 72290145-72290145 on Build GRCh38
rs765191400 Chr.15: 72290169-72290169 on Build GRCh38
rs776776022 Chr.15: 72289967-72289967 on Build GRCh38
rs778512637 Chr.15: 72290066-72290066 on Build GRCh38
rs369685906 Chr.15: 72290303-72290303 on Build GRCh38
rs1056793592 Chr.15: 72289861-72289861 on Build GRCh38
rs895496676 Chr.15: 72289876-72289876 on Build GRCh38
rs763073658 Chr.15: 72290202-72290202 on Build GRCh38
rs759518980 Chr.15: 72289972-72289972 on Build GRCh38
rs758679995 Chr.15: 72290028-72290028 on Build GRCh38
rs770784305 Chr.15: 72290307-72290307 on Build GRCh38
rs146928966 Chr.15: 72290233-72290233 on Build GRCh38
rs913139946 Chr.15: 72290091-72290091 on Build GRCh38
rs138690830 Chr.15: 72290249-72290249 on Build GRCh38
rs762154571 Chr.15: 72290218-72290218 on Build GRCh38
rs770906804 Chr.15: 72290144-72290144 on Build GRCh38
rs751903396 Chr.15: 72290040-72290040 on Build GRCh38
rs188442869 Chr.15: 72290056-72290056 on Build GRCh38
rs773039652 Chr.15: 72290132-72290132 on Build GRCh38
rs749264776 Chr.15: 72289906-72289906 on Build GRCh38
rs771826892 Chr.15: 72290111-72290111 on Build GRCh38
rs181135362 Chr.15: 72290289-72290289 on Build GRCh38
rs748718952 Chr.15: 72289898-72289898 on Build GRCh38
rs764396887 Chr.15: 72290030-72290030 on Build GRCh38
rs767869120 Chr.15: 72290250-72290250 on Build GRCh38
rs370052513 Chr.15: 72290259-72290259 on Build GRCh38
rs1000711792 Chr.15: 72290209-72290209 on Build GRCh38
rs34566074 Chr.15: 72290195-72290195 on Build GRCh38
rs781703343 Chr.15: 72289890-72289890 on Build GRCh38
rs781664298 Chr.15: 72290046-72290046 on Build GRCh38

View Full Product Details