Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74798152-74798663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSK
Entrez Gene ID
1445
Gene Name
c-src tyrosine kinase
Gene Aliases
-
UniGene
Hs.77793
Gene Chromosome Location
Chr.15: 74782064-74803198 on Build GRCh38
Transcripts
NM_001127190.1 NM_004383.2 XM_005254165.4 XM_017021925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362834 c.5C>G p.S2* Chr.15: 74798302-74798302 on Build GRCh38
4483410 c.26C>T p.P9L Chr.15: 74798625-74798625 on Build GRCh38
964901 c.18C>T p.A6A Chr.15: 74798617-74798617 on Build GRCh38
4056916 c.33T>C p.G11G Chr.15: 74798632-74798632 on Build GRCh38
4802085 c.17C>T p.A6V Chr.15: 74798616-74798616 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748128881 Chr.15: 74798588-74798588 on Build GRCh38
rs34621608 Chr.15: 74798292-74798292 on Build GRCh38
rs775379253 Chr.15: 74798378-74798378 on Build GRCh38
rs769276600 Chr.15: 74798482-74798482 on Build GRCh38
rs754301895 Chr.15: 74798479-74798479 on Build GRCh38
rs750462626 Chr.15: 74798547-74798547 on Build GRCh38
rs926272487 Chr.15: 74798181-74798181 on Build GRCh38
rs764121171 Chr.15: 74798392-74798392 on Build GRCh38
rs780409316 Chr.15: 74798451-74798451 on Build GRCh38
rs879781107 Chr.15: 74798187-74798187 on Build GRCh38
rs752949343 Chr.15: 74798617-74798617 on Build GRCh38
rs533915414 Chr.15: 74798455-74798455 on Build GRCh38
rs750688826 Chr.15: 74798424-74798424 on Build GRCh38
rs751618916 Chr.15: 74798255-74798255 on Build GRCh38
rs539638409 Chr.15: 74798342-74798342 on Build GRCh38
rs986288951 Chr.15: 74798365-74798365 on Build GRCh38
rs777608542 Chr.15: 74798428-74798431 on Build GRCh38
rs1034416690 Chr.15: 74798214-74798214 on Build GRCh38
rs756239294 Chr.15: 74798442-74798442 on Build GRCh38
rs749131234 Chr.15: 74798577-74798577 on Build GRCh38
rs779378936 Chr.15: 74798462-74798462 on Build GRCh38
rs543814060 Chr.15: 74798512-74798512 on Build GRCh38
rs180782355 Chr.15: 74798590-74798590 on Build GRCh38
rs761927513 Chr.15: 74798419-74798419 on Build GRCh38
rs754259592 Chr.15: 74798527-74798527 on Build GRCh38
rs376509733 Chr.15: 74798597-74798597 on Build GRCh38
rs755313963 Chr.15: 74798541-74798541 on Build GRCh38
rs749997068 Chr.15: 74798619-74798619 on Build GRCh38
rs764485759 Chr.15: 74798478-74798478 on Build GRCh38
rs777272259 Chr.15: 74798316-74798316 on Build GRCh38
rs558023410 Chr.15: 74798385-74798385 on Build GRCh38
rs201857254 Chr.15: 74798620-74798620 on Build GRCh38
rs750801661 Chr.15: 74798273-74798273 on Build GRCh38
rs780770731 Chr.15: 74798554-74798554 on Build GRCh38
rs746550185 Chr.15: 74798474-74798474 on Build GRCh38
rs780239018 Chr.15: 74798280-74798280 on Build GRCh38
rs550014244 Chr.15: 74798489-74798489 on Build GRCh38
rs761615445 Chr.15: 74798490-74798490 on Build GRCh38
rs746518753 Chr.15: 74798323-74798323 on Build GRCh38
rs758785044 Chr.15: 74798553-74798553 on Build GRCh38
rs199583396 Chr.15: 74798274-74798274 on Build GRCh38
rs377132612 Chr.15: 74798241-74798241 on Build GRCh38
rs745361756 Chr.15: 74798555-74798555 on Build GRCh38
rs920734793 Chr.15: 74798355-74798355 on Build GRCh38
rs188895080 Chr.15: 74798421-74798421 on Build GRCh38
rs371601033 Chr.15: 74798324-74798324 on Build GRCh38
rs750410782 Chr.15: 74798494-74798494 on Build GRCh38
rs200104962 Chr.15: 74798596-74798596 on Build GRCh38
rs1014352439 Chr.15: 74798509-74798509 on Build GRCh38
rs769095776 Chr.15: 74798306-74798306 on Build GRCh38
rs779826778 Chr.15: 74798576-74798576 on Build GRCh38
rs900483557 Chr.15: 74798503-74798503 on Build GRCh38
rs769325298 Chr.15: 74798274-74798274 on Build GRCh38
rs905038917 Chr.15: 74798183-74798183 on Build GRCh38
rs147315449 Chr.15: 74798629-74798629 on Build GRCh38
rs148597207 Chr.15: 74798486-74798486 on Build GRCh38
rs781458941 Chr.15: 74798266-74798266 on Build GRCh38
rs779667521 Chr.15: 74798624-74798624 on Build GRCh38
rs751743360 Chr.15: 74798409-74798409 on Build GRCh38
rs758375410 Chr.15: 74798248-74798248 on Build GRCh38
rs767656743 Chr.15: 74798493-74798493 on Build GRCh38
rs34072318 Chr.15: 74798546-74798546 on Build GRCh38
rs758566357 Chr.15: 74798568-74798568 on Build GRCh38
rs757484565 Chr.15: 74798261-74798261 on Build GRCh38
rs773409430 Chr.15: 74798591-74798591 on Build GRCh38
rs746964297 Chr.15: 74798599-74798599 on Build GRCh38
rs917673642 Chr.15: 74798329-74798329 on Build GRCh38
rs770448298 Chr.15: 74798475-74798475 on Build GRCh38
rs779308254 Chr.15: 74798504-74798504 on Build GRCh38
rs770548268 Chr.15: 74798340-74798340 on Build GRCh38
rs760957740 Chr.15: 74798510-74798510 on Build GRCh38
rs774239919 Chr.15: 74798585-74798585 on Build GRCh38
rs754136746 Chr.15: 74798454-74798454 on Build GRCh38
rs200997069 Chr.15: 74798364-74798364 on Build GRCh38
rs60357799 Chr.15: 74798614-74798614 on Build GRCh38
rs759294003 Chr.15: 74798344-74798344 on Build GRCh38
rs758951634 Chr.15: 74798618-74798618 on Build GRCh38
rs768602655 Chr.15: 74798582-74798582 on Build GRCh38
rs980331231 Chr.15: 74798609-74798609 on Build GRCh38
rs748909623 Chr.15: 74798604-74798605 on Build GRCh38
rs766637907 Chr.15: 74798592-74798592 on Build GRCh38
rs755885702 Chr.15: 74798557-74798557 on Build GRCh38
rs753230129 Chr.15: 74798228-74798228 on Build GRCh38
rs764294944 Chr.15: 74798251-74798251 on Build GRCh38

View Full Product Details