Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75355690-75356180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
UniGene
Hs.26232
Gene Chromosome Location
Chr.15: 75355792-75368680 on Build GRCh38
Transcripts
NM_001256494.1 NM_001256495.1 NM_001256496.1 NM_006715.3 XM_005254384.2 XM_017022186.1 XM_017022187.1 XM_017022188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1939230 c.2991C>A p.A997A Chr.15: 75356115-75356115 on Build GRCh38
1939231 c.2940C>T p.H980H Chr.15: 75356166-75356166 on Build GRCh38
471070 c.3031C>A p.H1011N Chr.15: 75355998-75355998 on Build GRCh38
4056967 c.2961G>A p.W987* Chr.15: 75356145-75356145 on Build GRCh38
1374599 c.3098T>C p.L1033P Chr.15: 75355931-75355931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764882521 Chr.15: 75356160-75356160 on Build GRCh38
rs1038276859 Chr.15: 75356080-75356080 on Build GRCh38
rs774630337 Chr.15: 75356146-75356146 on Build GRCh38
rs777625418 Chr.15: 75356098-75356098 on Build GRCh38
rs747425562 Chr.15: 75355967-75355967 on Build GRCh38
rs900723642 Chr.15: 75355743-75355743 on Build GRCh38
rs200716401 Chr.15: 75356084-75356084 on Build GRCh38
rs1045988414 Chr.15: 75355820-75355821 on Build GRCh38
rs779226176 Chr.15: 75355950-75355950 on Build GRCh38
rs752792380 Chr.15: 75356065-75356065 on Build GRCh38
rs1128989 Chr.15: 75355871-75355871 on Build GRCh38
rs370541771 Chr.15: 75356136-75356136 on Build GRCh38
rs15970 Chr.15: 75356014-75356014 on Build GRCh38
rs377590568 Chr.15: 75356052-75356052 on Build GRCh38
rs140762875 Chr.15: 75356148-75356148 on Build GRCh38
rs762592247 Chr.15: 75355910-75355910 on Build GRCh38
rs769539126 Chr.15: 75355975-75355975 on Build GRCh38
rs750756332 Chr.15: 75356153-75356153 on Build GRCh38
rs538103458 Chr.15: 75356008-75356008 on Build GRCh38
rs775597447 Chr.15: 75356133-75356133 on Build GRCh38
rs760490250 Chr.15: 75355995-75355995 on Build GRCh38
rs556865249 Chr.15: 75355824-75355824 on Build GRCh38
rs745692150 Chr.15: 75356128-75356128 on Build GRCh38
rs112273984 Chr.15: 75355805-75355805 on Build GRCh38
rs990625950 Chr.15: 75356073-75356073 on Build GRCh38
rs779386791 Chr.15: 75356079-75356080 on Build GRCh38
rs199706127 Chr.15: 75355898-75355898 on Build GRCh38
rs978362067 Chr.15: 75355955-75355955 on Build GRCh38
rs770721304 Chr.15: 75356044-75356044 on Build GRCh38
rs778267823 Chr.15: 75355966-75355966 on Build GRCh38
rs778045672 Chr.15: 75355885-75355885 on Build GRCh38
rs959252750 Chr.15: 75355851-75355851 on Build GRCh38
rs966396271 Chr.15: 75355755-75355755 on Build GRCh38
rs1018096804 Chr.15: 75355948-75355948 on Build GRCh38
rs778461649 Chr.15: 75356034-75356034 on Build GRCh38
rs771953912 Chr.15: 75356036-75356036 on Build GRCh38
rs778465964 Chr.15: 75356019-75356019 on Build GRCh38
rs370543184 Chr.15: 75356029-75356029 on Build GRCh38
rs772496039 Chr.15: 75355867-75355867 on Build GRCh38
rs745408548 Chr.15: 75356158-75356159 on Build GRCh38
rs925043740 Chr.15: 75355767-75355767 on Build GRCh38
rs753878575 Chr.15: 75355932-75355932 on Build GRCh38
rs755102911 Chr.15: 75355943-75355943 on Build GRCh38
rs369456817 Chr.15: 75355976-75355976 on Build GRCh38
rs370700941 Chr.15: 75356071-75356071 on Build GRCh38
rs1006270685 Chr.15: 75355940-75355940 on Build GRCh38
rs768248874 Chr.15: 75356033-75356033 on Build GRCh38
rs139259204 Chr.15: 75356090-75356092 on Build GRCh38
rs976030009 Chr.15: 75355730-75355730 on Build GRCh38
rs150070963 Chr.15: 75356028-75356028 on Build GRCh38
rs756453791 Chr.15: 75356157-75356157 on Build GRCh38
rs548988769 Chr.15: 75355745-75355745 on Build GRCh38
rs995093527 Chr.15: 75355816-75355816 on Build GRCh38
rs754077447 Chr.15: 75356003-75356003 on Build GRCh38
rs139418901 Chr.15: 75355986-75355986 on Build GRCh38
rs1128995 Chr.15: 75355854-75355854 on Build GRCh38
rs1033281042 Chr.15: 75356112-75356112 on Build GRCh38
rs5745933 Chr.15: 75355797-75355797 on Build GRCh38
rs554272385 Chr.15: 75355974-75355974 on Build GRCh38
rs1033314699 Chr.15: 75355821-75355821 on Build GRCh38
rs962474223 Chr.15: 75355814-75355814 on Build GRCh38
rs758917954 Chr.15: 75356016-75356016 on Build GRCh38
rs745509147 Chr.15: 75355884-75355884 on Build GRCh38
rs767342082 Chr.15: 75355929-75355929 on Build GRCh38
rs1128978 Chr.15: 75355958-75355958 on Build GRCh38
rs199637132 Chr.15: 75356015-75356015 on Build GRCh38
rs778458841 Chr.15: 75356085-75356085 on Build GRCh38
rs775117015 Chr.15: 75355903-75355903 on Build GRCh38
rs767979691 Chr.15: 75356082-75356082 on Build GRCh38
rs776077716 Chr.15: 75355874-75355874 on Build GRCh38
rs200547980 Chr.15: 75355866-75355866 on Build GRCh38
rs762975142 Chr.15: 75356070-75356070 on Build GRCh38
rs186846962 Chr.15: 75356078-75356078 on Build GRCh38
rs754355595 Chr.15: 75355919-75355919 on Build GRCh38
rs773801306 Chr.15: 75355985-75355985 on Build GRCh38
rs189284876 Chr.15: 75355757-75355757 on Build GRCh38
rs886591371 Chr.15: 75355773-75355773 on Build GRCh38
rs763888919 Chr.15: 75355914-75355914 on Build GRCh38
rs182211802 Chr.15: 75356020-75356020 on Build GRCh38
rs746948219 Chr.15: 75356100-75356100 on Build GRCh38
rs770915774 Chr.15: 75356126-75356126 on Build GRCh38
rs760408496 Chr.15: 75356041-75356041 on Build GRCh38
rs967033543 Chr.15: 75356156-75356156 on Build GRCh38
rs775916608 Chr.15: 75355747-75355747 on Build GRCh38
rs747642374 Chr.15: 75356086-75356086 on Build GRCh38
rs764646840 Chr.15: 75356077-75356077 on Build GRCh38
rs759793641 Chr.15: 75356058-75356058 on Build GRCh38
rs765429313 Chr.15: 75356064-75356064 on Build GRCh38
rs766535676 Chr.15: 75355996-75355996 on Build GRCh38
rs753897562 Chr.15: 75355915-75355915 on Build GRCh38
rs1128999 Chr.15: 75355819-75355819 on Build GRCh38
rs373931168 Chr.15: 75355977-75355977 on Build GRCh38
rs771954600 Chr.15: 75355990-75355990 on Build GRCh38
rs756309307 Chr.15: 75355930-75355930 on Build GRCh38
rs763060309 Chr.15: 75356134-75356134 on Build GRCh38
rs757819142 Chr.15: 75355921-75355922 on Build GRCh38
rs753250089 Chr.15: 75355954-75355954 on Build GRCh38
rs770019171 Chr.15: 75356131-75356131 on Build GRCh38
rs371876324 Chr.15: 75356127-75356127 on Build GRCh38
rs761449790 Chr.15: 75355916-75355916 on Build GRCh38
rs772963654 Chr.15: 75355993-75355993 on Build GRCh38
rs768176221 Chr.15: 75356150-75356150 on Build GRCh38
rs376869970 Chr.15: 75356012-75356012 on Build GRCh38
rs561303224 Chr.15: 75356161-75356161 on Build GRCh38
rs201238629 Chr.15: 75356040-75356040 on Build GRCh38
rs1128987 Chr.15: 75355878-75355878 on Build GRCh38
rs757913166 Chr.15: 75356092-75356092 on Build GRCh38

View Full Product Details