Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75375576-75376078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIN3A
Entrez Gene ID
25942
Gene Name
SIN3 transcription regulator family member A
Gene Aliases
-
UniGene
Hs.513039
Gene Chromosome Location
Chr.15: 75369379-75455819 on Build GRCh38
Transcripts
NM_001145357.1 NM_001145358.1 NM_015477.2 XM_006720465.3 XM_006720466.3 XM_006720467.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
195877 c.3418C>T p.R1140* Chr.15: 75375838-75375838 on Build GRCh38
1374606 c.3419G>A p.R1140Q Chr.15: 75375837-75375837 on Build GRCh38
7096425 c.3444G>C p.K1148N Chr.15: 75375812-75375812 on Build GRCh38
6257379 c.3584C>A p.A1195D Chr.15: 75375672-75375672 on Build GRCh38
5635100 c.3547G>T p.E1183* Chr.15: 75375709-75375709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777090934 Chr.15: 75375677-75375677 on Build GRCh38
rs766896566 Chr.15: 75375891-75375891 on Build GRCh38
rs754243706 Chr.15: 75375837-75375837 on Build GRCh38
rs946672382 Chr.15: 75375864-75375864 on Build GRCh38
rs138854954 Chr.15: 75375868-75375868 on Build GRCh38
rs1001901331 Chr.15: 75376033-75376033 on Build GRCh38
rs1034760611 Chr.15: 75376051-75376051 on Build GRCh38
rs1027013727 Chr.15: 75375622-75375622 on Build GRCh38
rs993758890 Chr.15: 75375608-75375608 on Build GRCh38
rs761876235 Chr.15: 75375803-75375803 on Build GRCh38
rs556160377 Chr.15: 75375861-75375861 on Build GRCh38
rs780874168 Chr.15: 75375851-75375851 on Build GRCh38
rs372034400 Chr.15: 75375818-75375818 on Build GRCh38
rs749092824 Chr.15: 75375859-75375859 on Build GRCh38
rs776172017 Chr.15: 75375648-75375649 on Build GRCh38
rs150503184 Chr.15: 75375738-75375738 on Build GRCh38
rs757004591 Chr.15: 75375911-75375911 on Build GRCh38
rs375053575 Chr.15: 75375619-75375619 on Build GRCh38
rs573001904 Chr.15: 75376029-75376029 on Build GRCh38
rs769304744 Chr.15: 75375805-75375805 on Build GRCh38
rs202118407 Chr.15: 75375661-75375661 on Build GRCh38
rs770636953 Chr.15: 75375732-75375732 on Build GRCh38
rs537899729 Chr.15: 75375802-75375802 on Build GRCh38
rs761191233 Chr.15: 75375821-75375821 on Build GRCh38
rs746516200 Chr.15: 75375689-75375689 on Build GRCh38
rs749964156 Chr.15: 75375923-75375923 on Build GRCh38
rs60213317 Chr.15: 75375790-75375790 on Build GRCh38
rs777072630 Chr.15: 75375902-75375902 on Build GRCh38
rs772514204 Chr.15: 75375623-75375623 on Build GRCh38
rs145577994 Chr.15: 75375866-75375866 on Build GRCh38
rs142815984 Chr.15: 75375906-75375906 on Build GRCh38
rs926564869 Chr.15: 75375948-75375948 on Build GRCh38
rs758076139 Chr.15: 75375675-75375675 on Build GRCh38
rs752254316 Chr.15: 75375669-75375669 on Build GRCh38
rs756438000 Chr.15: 75375655-75375655 on Build GRCh38
rs201447452 Chr.15: 75375878-75375878 on Build GRCh38
rs774601051 Chr.15: 75375865-75375865 on Build GRCh38
rs181485654 Chr.15: 75375624-75375624 on Build GRCh38
rs144295678 Chr.15: 75375799-75375799 on Build GRCh38
rs763801195 Chr.15: 75375844-75375844 on Build GRCh38
rs959302324 Chr.15: 75375952-75375952 on Build GRCh38
rs768051074 Chr.15: 75375924-75375924 on Build GRCh38
rs1040518758 Chr.15: 75375894-75375894 on Build GRCh38
rs1026333745 Chr.15: 75375938-75375938 on Build GRCh38
rs887998890 Chr.15: 75375957-75375957 on Build GRCh38
rs745758382 Chr.15: 75375854-75375854 on Build GRCh38
rs747408351 Chr.15: 75375894-75375895 on Build GRCh38
rs1009645328 Chr.15: 75375901-75375901 on Build GRCh38
rs375820448 Chr.15: 75375903-75375903 on Build GRCh38
rs756808431 Chr.15: 75375848-75375848 on Build GRCh38
rs897964621 Chr.15: 75375806-75375806 on Build GRCh38
rs147800016 Chr.15: 75375843-75375843 on Build GRCh38
rs939053954 Chr.15: 75375822-75375822 on Build GRCh38
rs769666153 Chr.15: 75375770-75375770 on Build GRCh38
rs1006482775 Chr.15: 75375985-75375985 on Build GRCh38
rs745852705 Chr.15: 75375743-75375743 on Build GRCh38
rs969076616 Chr.15: 75375696-75375696 on Build GRCh38
rs761115423 Chr.15: 75375890-75375890 on Build GRCh38
rs985341985 Chr.15: 75375685-75375685 on Build GRCh38
rs756123555 Chr.15: 75375856-75375856 on Build GRCh38
rs991826892 Chr.15: 75375965-75375965 on Build GRCh38
rs141700684 Chr.15: 75375845-75375845 on Build GRCh38
rs767065732 Chr.15: 75375919-75375919 on Build GRCh38
rs1023147564 Chr.15: 75375686-75375686 on Build GRCh38
rs1023049938 Chr.15: 75376001-75376001 on Build GRCh38
rs371714083 Chr.15: 75375830-75375830 on Build GRCh38

View Full Product Details