Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77055610-77056131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN3
Entrez Gene ID
10099
Gene Name
tetraspanin 3
Gene Aliases
TM4-A, TM4SF8, TSPAN-3
UniGene
Hs.744863
Gene Chromosome Location
Chr.15: 77044017-77071228 on Build GRCh38
Transcripts
NM_001168412.1 NM_005724.5 NM_198902.2 XM_017021857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4057057 c.278T>G p.V93G Chr.15: 77055841-77055841 on Build GRCh38
1374692 c.238C>T p.R80C Chr.15: 77056081-77056081 on Build GRCh38
1940074 c.229C>T p.R77W Chr.15: 77056090-77056090 on Build GRCh38
6810635 c.255G>A p.T85T Chr.15: 77056064-77056064 on Build GRCh38
1678601 c.289G>T p.E97* Chr.15: 77055830-77055830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772333818 Chr.15: 77055906-77055911 on Build GRCh38
rs757883139 Chr.15: 77055879-77055879 on Build GRCh38
rs764241691 Chr.15: 77056041-77056041 on Build GRCh38
rs369282184 Chr.15: 77056035-77056035 on Build GRCh38
rs375851256 Chr.15: 77056016-77056016 on Build GRCh38
rs778024681 Chr.15: 77055857-77055857 on Build GRCh38
rs1022094331 Chr.15: 77055714-77055714 on Build GRCh38
rs185197169 Chr.15: 77055937-77055937 on Build GRCh38
rs376931567 Chr.15: 77056081-77056081 on Build GRCh38
rs929752200 Chr.15: 77055995-77055995 on Build GRCh38
rs759701807 Chr.15: 77055820-77055822 on Build GRCh38
rs773077958 Chr.15: 77055756-77055756 on Build GRCh38
rs891802776 Chr.15: 77056059-77056059 on Build GRCh38
rs148163067 Chr.15: 77056090-77056090 on Build GRCh38
rs142023609 Chr.15: 77056101-77056101 on Build GRCh38
rs566586852 Chr.15: 77055951-77055951 on Build GRCh38
rs746580028 Chr.15: 77056091-77056091 on Build GRCh38
rs765431679 Chr.15: 77055802-77055802 on Build GRCh38
rs761296667 Chr.15: 77055739-77055739 on Build GRCh38
rs976436129 Chr.15: 77055954-77055954 on Build GRCh38
rs936696794 Chr.15: 77055919-77055919 on Build GRCh38
rs909586032 Chr.15: 77056006-77056006 on Build GRCh38
rs768350451 Chr.15: 77055889-77055889 on Build GRCh38
rs755204868 Chr.15: 77055787-77055787 on Build GRCh38
rs555404533 Chr.15: 77055986-77055986 on Build GRCh38
rs111689029 Chr.15: 77055640-77055640 on Build GRCh38
rs767020436 Chr.15: 77055747-77055747 on Build GRCh38
rs1054160340 Chr.15: 77055929-77055929 on Build GRCh38
rs772737867 Chr.15: 77055903-77055903 on Build GRCh38
rs781602123 Chr.15: 77055880-77055880 on Build GRCh38
rs968916359 Chr.15: 77055730-77055730 on Build GRCh38
rs760627106 Chr.15: 77055776-77055776 on Build GRCh38
rs569382363 Chr.15: 77055685-77055685 on Build GRCh38
rs781617303 Chr.15: 77056057-77056057 on Build GRCh38
rs867653823 Chr.15: 77056058-77056058 on Build GRCh38
rs533831329 Chr.15: 77055979-77055979 on Build GRCh38
rs539999250 Chr.15: 77055819-77055819 on Build GRCh38
rs760574109 Chr.15: 77055910-77055910 on Build GRCh38
rs369375820 Chr.15: 77055777-77055777 on Build GRCh38
rs763301747 Chr.15: 77056036-77056036 on Build GRCh38
rs114063611 Chr.15: 77056052-77056052 on Build GRCh38
rs1003396978 Chr.15: 77056042-77056042 on Build GRCh38
rs1010312426 Chr.15: 77055712-77055712 on Build GRCh38
rs770196309 Chr.15: 77056053-77056053 on Build GRCh38
rs752075831 Chr.15: 77055871-77055871 on Build GRCh38
rs111545013 Chr.15: 77055999-77055999 on Build GRCh38
rs538395738 Chr.15: 77055661-77055661 on Build GRCh38
rs753616086 Chr.15: 77055778-77055778 on Build GRCh38
rs758482457 Chr.15: 77055825-77055825 on Build GRCh38
rs867807913 Chr.15: 77055892-77055892 on Build GRCh38
rs11550569 Chr.15: 77056095-77056095 on Build GRCh38
rs776361443 Chr.15: 77056096-77056096 on Build GRCh38
rs778364921 Chr.15: 77055890-77055890 on Build GRCh38
rs771520534 Chr.15: 77055899-77055899 on Build GRCh38
rs747814096 Chr.15: 77055894-77055894 on Build GRCh38
rs986727219 Chr.15: 77055718-77055718 on Build GRCh38
rs771749795 Chr.15: 77056084-77056084 on Build GRCh38
rs956343160 Chr.15: 77055705-77055705 on Build GRCh38
rs143800573 Chr.15: 77056065-77056065 on Build GRCh38
rs115731547 Chr.15: 77056024-77056024 on Build GRCh38
rs756378132 Chr.15: 77056068-77056068 on Build GRCh38
rs367828134 Chr.15: 77056018-77056018 on Build GRCh38
rs551243802 Chr.15: 77055674-77055674 on Build GRCh38
rs918254125 Chr.15: 77055978-77055978 on Build GRCh38
rs11550567 Chr.15: 77056103-77056103 on Build GRCh38
rs770903340 Chr.15: 77056015-77056015 on Build GRCh38
rs746276424 Chr.15: 77055886-77055886 on Build GRCh38

View Full Product Details