Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77111604-77112111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEAK1
Entrez Gene ID
79834
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
SGK269
UniGene
Hs.731402
Gene Chromosome Location
Chr.15: 77100829-77420104 on Build GRCh38
Transcripts
NM_024776.3 XM_005254670.2 XM_005254671.2 XM_005254673.2 XM_005254674.2 XM_005254675.4 XM_011522033.2 XM_011522034.2 XM_011522035.2 XM_011522036.2 XM_011522037.2 XM_011522038.2 XM_011522039.2 XM_011522040.1 XM_011522042.2

SNP Detail

SNP ID Chromosome Location
rs1032708715 Chr.15: 77111975-77111975 on Build GRCh38
rs1056381612 Chr.15: 77111826-77111826 on Build GRCh38
rs1016486585 Chr.15: 77111638-77111638 on Build GRCh38
rs1012918222 Chr.15: 77111740-77111740 on Build GRCh38
rs1038381067 Chr.15: 77111885-77111885 on Build GRCh38
rs140945955 Chr.15: 77111772-77111772 on Build GRCh38
rs140113344 Chr.15: 77111741-77111741 on Build GRCh38
rs955538585 Chr.15: 77111686-77111686 on Build GRCh38
rs910283419 Chr.15: 77112044-77112044 on Build GRCh38
rs771585658 Chr.15: 77111857-77111857 on Build GRCh38
rs144951678 Chr.15: 77111768-77111768 on Build GRCh38
rs540579499 Chr.15: 77111816-77111816 on Build GRCh38
rs933059489 Chr.15: 77111808-77111808 on Build GRCh38
rs923055881 Chr.15: 77111789-77111789 on Build GRCh38
rs945177066 Chr.15: 77111853-77111853 on Build GRCh38
rs551016808 Chr.15: 77112008-77112008 on Build GRCh38
rs191461124 Chr.15: 77111932-77111932 on Build GRCh38
rs775044679 Chr.15: 77111864-77111864 on Build GRCh38
rs987798782 Chr.15: 77111783-77111783 on Build GRCh38
rs762412382 Chr.15: 77111928-77111928 on Build GRCh38
rs529509103 Chr.15: 77111842-77111842 on Build GRCh38
rs149865757 Chr.15: 77111756-77111756 on Build GRCh38
rs979778678 Chr.15: 77111758-77111758 on Build GRCh38
rs942220319 Chr.15: 77112057-77112057 on Build GRCh38
rs184766435 Chr.15: 77112033-77112033 on Build GRCh38
rs1000692256 Chr.15: 77111953-77111953 on Build GRCh38
rs960103966 Chr.15: 77111729-77111730 on Build GRCh38
rs117994354 Chr.15: 77111811-77111811 on Build GRCh38
rs1042267750 Chr.15: 77111890-77111890 on Build GRCh38
rs533536766 Chr.15: 77112034-77112034 on Build GRCh38
rs962739502 Chr.15: 77112060-77112060 on Build GRCh38
rs1035763127 Chr.15: 77111711-77111711 on Build GRCh38
rs35165014 Chr.15: 77111910-77111911 on Build GRCh38
rs77974454 Chr.15: 77111947-77111947 on Build GRCh38
rs186604142 Chr.15: 77111630-77111630 on Build GRCh38
rs988017715 Chr.15: 77112023-77112023 on Build GRCh38
rs4886850 Chr.15: 77111818-77111818 on Build GRCh38
rs997296378 Chr.15: 77111956-77111956 on Build GRCh38
rs895789712 Chr.15: 77111942-77111942 on Build GRCh38
rs1030207224 Chr.15: 77111681-77111681 on Build GRCh38
rs77632502 Chr.15: 77111948-77111948 on Build GRCh38
rs141974531 Chr.15: 77111681-77111686 on Build GRCh38
rs1029865128 Chr.15: 77112006-77112006 on Build GRCh38
rs375040888 Chr.15: 77111745-77111745 on Build GRCh38
rs942187369 Chr.15: 77111843-77111843 on Build GRCh38
rs922939619 Chr.15: 77112024-77112024 on Build GRCh38
rs747478364 Chr.15: 77111837-77111837 on Build GRCh38
rs898062256 Chr.15: 77111887-77111887 on Build GRCh38
rs993490267 Chr.15: 77111901-77111901 on Build GRCh38
rs941130979 Chr.15: 77111760-77111760 on Build GRCh38
rs1012969303 Chr.15: 77111943-77111943 on Build GRCh38
rs973850565 Chr.15: 77112089-77112089 on Build GRCh38
rs1015727233 Chr.15: 77112035-77112035 on Build GRCh38

View Full Product Details