Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80122619-80123131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFAND6
Entrez Gene ID
54469
Gene Name
zinc finger AN1-type containing 6
Gene Aliases
AWP1, ZA20D3, ZFAND5B
UniGene
Hs.596679
Gene Chromosome Location
Chr.15: 80059568-80138393 on Build GRCh38
Transcripts
NM_001242911.1 NM_001242912.1 NM_001242913.1 NM_001242914.1 NM_001242915.1 NM_001242916.1 NM_001242917.1 NM_001242918.1 NM_001242919.1 NM_019006.3 XM_011521684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903893 c.314T>G p.L105W Chr.15: 80122750-80122750 on Build GRCh38
701666 c.283C>T p.L95F Chr.15: 80122719-80122719 on Build GRCh38
3387082 c.311A>G p.Q104R Chr.15: 80122747-80122747 on Build GRCh38
6143003 c.364+1G>A p.? Chr.15: 80122801-80122801 on Build GRCh38
6425177 c.301G>T p.A101S Chr.15: 80122737-80122737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756729074 Chr.15: 80122994-80122994 on Build GRCh38
rs778390153 Chr.15: 80122683-80122683 on Build GRCh38
rs368985546 Chr.15: 80122723-80122723 on Build GRCh38
rs745534585 Chr.15: 80123084-80123084 on Build GRCh38
rs763695689 Chr.15: 80122960-80122960 on Build GRCh38
rs1029788365 Chr.15: 80123048-80123048 on Build GRCh38
rs891540469 Chr.15: 80123037-80123041 on Build GRCh38
rs941343389 Chr.15: 80122873-80122873 on Build GRCh38
rs151059961 Chr.15: 80122724-80122724 on Build GRCh38
rs147509612 Chr.15: 80122740-80122740 on Build GRCh38
rs934500639 Chr.15: 80122964-80122964 on Build GRCh38
rs201758936 Chr.15: 80122645-80122645 on Build GRCh38
rs117247288 Chr.15: 80123085-80123085 on Build GRCh38
rs371672655 Chr.15: 80122701-80122701 on Build GRCh38
rs747026426 Chr.15: 80122697-80122697 on Build GRCh38
rs915229009 Chr.15: 80122894-80122894 on Build GRCh38
rs755177634 Chr.15: 80122843-80122843 on Build GRCh38
rs895721560 Chr.15: 80122655-80122655 on Build GRCh38
rs778099390 Chr.15: 80122819-80122819 on Build GRCh38
rs748592585 Chr.15: 80122659-80122659 on Build GRCh38
rs867438423 Chr.15: 80123104-80123104 on Build GRCh38
rs969710220 Chr.15: 80122704-80122704 on Build GRCh38
rs72630427 Chr.15: 80122962-80122962 on Build GRCh38
rs918655783 Chr.15: 80122897-80122897 on Build GRCh38
rs1015803758 Chr.15: 80122755-80122755 on Build GRCh38
rs768373723 Chr.15: 80122725-80122725 on Build GRCh38
rs188075840 Chr.15: 80123094-80123094 on Build GRCh38
rs112813114 Chr.15: 80122858-80122858 on Build GRCh38
rs577250371 Chr.15: 80123005-80123005 on Build GRCh38
rs917001815 Chr.15: 80123025-80123025 on Build GRCh38
rs767326491 Chr.15: 80122737-80122737 on Build GRCh38
rs961534443 Chr.15: 80122773-80122773 on Build GRCh38
rs776781451 Chr.15: 80122709-80122709 on Build GRCh38
rs1029759450 Chr.15: 80122817-80122817 on Build GRCh38
rs770856908 Chr.15: 80122831-80122831 on Build GRCh38
rs79104576 Chr.15: 80122881-80122881 on Build GRCh38
rs781252049 Chr.15: 80122832-80122832 on Build GRCh38
rs868304000 Chr.15: 80122658-80122658 on Build GRCh38
rs760056399 Chr.15: 80122742-80122742 on Build GRCh38
rs750356337 Chr.15: 80122720-80122720 on Build GRCh38
rs112075519 Chr.15: 80122706-80122706 on Build GRCh38
rs146488669 Chr.15: 80122784-80122784 on Build GRCh38
rs573241410 Chr.15: 80123093-80123093 on Build GRCh38
rs201635973 Chr.15: 80122754-80122754 on Build GRCh38
rs911221530 Chr.15: 80122863-80122863 on Build GRCh38
rs1047586949 Chr.15: 80122916-80122916 on Build GRCh38
rs954127323 Chr.15: 80122864-80122864 on Build GRCh38
rs1044481135 Chr.15: 80123054-80123054 on Build GRCh38
rs929152013 Chr.15: 80122907-80122907 on Build GRCh38
rs11554805 Chr.15: 80122711-80122711 on Build GRCh38
rs552181058 Chr.15: 80122731-80122731 on Build GRCh38
rs772448424 Chr.15: 80122665-80122665 on Build GRCh38
rs200534739 Chr.15: 80122732-80122732 on Build GRCh38
rs905537284 Chr.15: 80123016-80123016 on Build GRCh38
rs771069921 Chr.15: 80122702-80122702 on Build GRCh38
rs746013771 Chr.15: 80122846-80122846 on Build GRCh38
rs559318164 Chr.15: 80123090-80123090 on Build GRCh38
rs1010402510 Chr.15: 80123039-80123039 on Build GRCh38
rs753477601 Chr.15: 80122749-80122749 on Build GRCh38
rs988927952 Chr.15: 80122750-80122750 on Build GRCh38
rs747571037 Chr.15: 80122821-80122821 on Build GRCh38
rs747552499 Chr.15: 80122756-80122756 on Build GRCh38
rs765930669 Chr.15: 80122748-80122748 on Build GRCh38
rs780827797 Chr.15: 80123076-80123076 on Build GRCh38
rs948477810 Chr.15: 80123032-80123032 on Build GRCh38
rs74647991 Chr.15: 80122927-80122927 on Build GRCh38
rs546096513 Chr.15: 80123103-80123103 on Build GRCh38
rs758833474 Chr.15: 80122813-80122813 on Build GRCh38
rs149918624 Chr.15: 80122781-80122781 on Build GRCh38
rs978642673 Chr.15: 80122934-80122934 on Build GRCh38
rs543836621 Chr.15: 80122654-80122654 on Build GRCh38
rs895770411 Chr.15: 80122919-80122919 on Build GRCh38

View Full Product Details