Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81001123-81001612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MESDC1
Entrez Gene ID
59274
Gene Name
mesoderm development candidate 1
Gene Aliases
-
UniGene
Hs.513071
Gene Chromosome Location
Chr.15: 81000954-81004004 on Build GRCh38
Transcripts
NM_022566.2

SNP Detail

SNP ID Chromosome Location
rs987197719 Chr.15: 81001341-81001341 on Build GRCh38
rs896004692 Chr.15: 81001464-81001464 on Build GRCh38
rs1042529340 Chr.15: 81001307-81001307 on Build GRCh38
rs937620355 Chr.15: 81001585-81001585 on Build GRCh38
rs1020747179 Chr.15: 81001331-81001332 on Build GRCh38
rs931449033 Chr.15: 81001381-81001381 on Build GRCh38
rs908739598 Chr.15: 81001569-81001569 on Build GRCh38
rs932548469 Chr.15: 81001266-81001266 on Build GRCh38
rs569133565 Chr.15: 81001286-81001286 on Build GRCh38
rs141417718 Chr.15: 81001354-81001354 on Build GRCh38
rs905102181 Chr.15: 81001182-81001182 on Build GRCh38
rs985348821 Chr.15: 81001270-81001270 on Build GRCh38
rs982907557 Chr.15: 81001564-81001564 on Build GRCh38
rs957835127 Chr.15: 81001297-81001297 on Build GRCh38
rs1022948613 Chr.15: 81001185-81001185 on Build GRCh38
rs975611814 Chr.15: 81001350-81001350 on Build GRCh38
rs1053668277 Chr.15: 81001454-81001454 on Build GRCh38
rs937842747 Chr.15: 81001468-81001468 on Build GRCh38
rs1036334603 Chr.15: 81001148-81001148 on Build GRCh38
rs866119200 Chr.15: 81001332-81001332 on Build GRCh38
rs549315728 Chr.15: 81001276-81001276 on Build GRCh38
rs912591738 Chr.15: 81001342-81001342 on Build GRCh38
rs949197868 Chr.15: 81001422-81001422 on Build GRCh38
rs1013633730 Chr.15: 81001143-81001143 on Build GRCh38
rs1020819506 Chr.15: 81001492-81001492 on Build GRCh38
rs1013135836 Chr.15: 81001465-81001465 on Build GRCh38
rs922507371 Chr.15: 81001260-81001260 on Build GRCh38
rs961511021 Chr.15: 81001558-81001558 on Build GRCh38
rs914877085 Chr.15: 81001301-81001301 on Build GRCh38
rs961498467 Chr.15: 81001246-81001246 on Build GRCh38
rs1020133349 Chr.15: 81001534-81001534 on Build GRCh38
rs976634812 Chr.15: 81001259-81001259 on Build GRCh38
rs553800977 Chr.15: 81001545-81001545 on Build GRCh38
rs1013186903 Chr.15: 81001172-81001172 on Build GRCh38
rs1016074364 Chr.15: 81001551-81001551 on Build GRCh38
rs1005588418 Chr.15: 81001541-81001541 on Build GRCh38
rs975394635 Chr.15: 81001540-81001540 on Build GRCh38
rs533629501 Chr.15: 81001543-81001543 on Build GRCh38
rs1047223697 Chr.15: 81001384-81001384 on Build GRCh38
rs960307494 Chr.15: 81001164-81001164 on Build GRCh38
rs1057292094 Chr.15: 81001453-81001453 on Build GRCh38
rs1023230135 Chr.15: 81001505-81001505 on Build GRCh38
rs894457635 Chr.15: 81001453-81001467 on Build GRCh38
rs1024684120 Chr.15: 81001181-81001181 on Build GRCh38
rs942925528 Chr.15: 81001345-81001345 on Build GRCh38
rs908657985 Chr.15: 81001228-81001228 on Build GRCh38
rs952692086 Chr.15: 81001205-81001205 on Build GRCh38
rs577708518 Chr.15: 81001542-81001542 on Build GRCh38
rs967851219 Chr.15: 81001500-81001500 on Build GRCh38
rs1046125884 Chr.15: 81001579-81001579 on Build GRCh38
rs963983437 Chr.15: 81001538-81001538 on Build GRCh38
rs888436119 Chr.15: 81001506-81001506 on Build GRCh38
rs1009399972 Chr.15: 81001469-81001469 on Build GRCh38
rs946439426 Chr.15: 81001302-81001302 on Build GRCh38
rs987980096 Chr.15: 81001324-81001324 on Build GRCh38
rs893728626 Chr.15: 81001456-81001456 on Build GRCh38
rs868460001 Chr.15: 81001215-81001215 on Build GRCh38
rs949138162 Chr.15: 81001574-81001574 on Build GRCh38
rs574167181 Chr.15: 81001546-81001546 on Build GRCh38
rs934812099 Chr.15: 81001438-81001438 on Build GRCh38
rs28387047 Chr.15: 81001587-81001587 on Build GRCh38
rs557785343 Chr.15: 81001503-81001503 on Build GRCh38
rs902284393 Chr.15: 81001436-81001436 on Build GRCh38

View Full Product Details