Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81136431-81136942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP161
Entrez Gene ID
161502
Gene Name
cilia and flagella associated protein 161
Gene Aliases
C15orf26
UniGene
Hs.130979
Gene Chromosome Location
Chr.15: 81099408-81149175 on Build GRCh38
Transcripts
NM_173528.2 XM_006720408.2 XM_011521280.1 XM_017021963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596379 c.287C>T p.P96L Chr.15: 81136643-81136643 on Build GRCh38
1749343 c.390G>C p.L130F Chr.15: 81136746-81136746 on Build GRCh38
6142994 c.341G>A p.S114N Chr.15: 81136697-81136697 on Build GRCh38
4666905 c.380T>G p.F127C Chr.15: 81136736-81136736 on Build GRCh38
2011823 c.172G>A p.V58I Chr.15: 81136528-81136528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753877234 Chr.15: 81136751-81136751 on Build GRCh38
rs768090433 Chr.15: 81136732-81136732 on Build GRCh38
rs768439798 Chr.15: 81136703-81136703 on Build GRCh38
rs759113074 Chr.15: 81136606-81136606 on Build GRCh38
rs756240929 Chr.15: 81136662-81136662 on Build GRCh38
rs757909801 Chr.15: 81136520-81136520 on Build GRCh38
rs981225719 Chr.15: 81136876-81136876 on Build GRCh38
rs781002231 Chr.15: 81136773-81136773 on Build GRCh38
rs367707361 Chr.15: 81136698-81136698 on Build GRCh38
rs376487839 Chr.15: 81136554-81136554 on Build GRCh38
rs8038176 Chr.15: 81136489-81136489 on Build GRCh38
rs957307762 Chr.15: 81136859-81136859 on Build GRCh38
rs959148390 Chr.15: 81136535-81136535 on Build GRCh38
rs200232799 Chr.15: 81136705-81136705 on Build GRCh38
rs753306255 Chr.15: 81136852-81136852 on Build GRCh38
rs372009759 Chr.15: 81136765-81136765 on Build GRCh38
rs746807332 Chr.15: 81136679-81136679 on Build GRCh38
rs543896743 Chr.15: 81136618-81136618 on Build GRCh38
rs202012296 Chr.15: 81136561-81136561 on Build GRCh38
rs78911995 Chr.15: 81136501-81136501 on Build GRCh38
rs765471393 Chr.15: 81136757-81136757 on Build GRCh38
rs368007727 Chr.15: 81136756-81136756 on Build GRCh38
rs762353686 Chr.15: 81136638-81136638 on Build GRCh38
rs764418714 Chr.15: 81136504-81136504 on Build GRCh38
rs200314900 Chr.15: 81136699-81136699 on Build GRCh38
rs1026713834 Chr.15: 81136480-81136480 on Build GRCh38
rs371138907 Chr.15: 81136671-81136671 on Build GRCh38
rs575572430 Chr.15: 81136642-81136642 on Build GRCh38
rs771385890 Chr.15: 81136756-81136757 on Build GRCh38
rs745817180 Chr.15: 81136779-81136779 on Build GRCh38
rs775183319 Chr.15: 81136615-81136615 on Build GRCh38
rs1010639922 Chr.15: 81136807-81136807 on Build GRCh38
rs761151429 Chr.15: 81136740-81136740 on Build GRCh38
rs759127027 Chr.15: 81136495-81136495 on Build GRCh38
rs201483243 Chr.15: 81136643-81136643 on Build GRCh38
rs777486386 Chr.15: 81136678-81136678 on Build GRCh38
rs755213848 Chr.15: 81136675-81136675 on Build GRCh38
rs373978881 Chr.15: 81136527-81136527 on Build GRCh38
rs1019950726 Chr.15: 81136865-81136865 on Build GRCh38
rs751301994 Chr.15: 81136761-81136761 on Build GRCh38
rs996034642 Chr.15: 81136832-81136832 on Build GRCh38
rs781411575 Chr.15: 81136522-81136522 on Build GRCh38
rs755677086 Chr.15: 81136567-81136567 on Build GRCh38
rs899447041 Chr.15: 81136680-81136680 on Build GRCh38
rs1009004293 Chr.15: 81136733-81136733 on Build GRCh38
rs971446810 Chr.15: 81136866-81136866 on Build GRCh38
rs771866435 Chr.15: 81136474-81136474 on Build GRCh38
rs763394362 Chr.15: 81136503-81136503 on Build GRCh38
rs747981930 Chr.15: 81136468-81136468 on Build GRCh38
rs766467350 Chr.15: 81136741-81136741 on Build GRCh38
rs190354276 Chr.15: 81136516-81136516 on Build GRCh38
rs778415768 Chr.15: 81136759-81136760 on Build GRCh38
rs182043667 Chr.15: 81136789-81136789 on Build GRCh38
rs563596352 Chr.15: 81136632-81136632 on Build GRCh38
rs888453713 Chr.15: 81136500-81136500 on Build GRCh38
rs745559208 Chr.15: 81136557-81136557 on Build GRCh38
rs773748600 Chr.15: 81136734-81136734 on Build GRCh38
rs796943845 Chr.15: 81136759-81136760 on Build GRCh38
rs769279001 Chr.15: 81136609-81136609 on Build GRCh38
rs78284409 Chr.15: 81136907-81136907 on Build GRCh38
rs377413316 Chr.15: 81136502-81136502 on Build GRCh38
rs748755771 Chr.15: 81136800-81136800 on Build GRCh38
rs762555812 Chr.15: 81136616-81136616 on Build GRCh38
rs762300584 Chr.15: 81136713-81136713 on Build GRCh38
rs747849720 Chr.15: 81136550-81136550 on Build GRCh38
rs748960955 Chr.15: 81136702-81136702 on Build GRCh38
rs543392858 Chr.15: 81136590-81136590 on Build GRCh38
rs201116091 Chr.15: 81136685-81136685 on Build GRCh38
rs780356046 Chr.15: 81136544-81136544 on Build GRCh38
rs756224699 Chr.15: 81136528-81136528 on Build GRCh38
rs1022016956 Chr.15: 81136851-81136851 on Build GRCh38
rs776175061 Chr.15: 81136491-81136491 on Build GRCh38
rs202018244 Chr.15: 81136634-81136634 on Build GRCh38
rs773177868 Chr.15: 81136476-81136476 on Build GRCh38
rs371319193 Chr.15: 81136555-81136555 on Build GRCh38
rs745839156 Chr.15: 81136693-81136693 on Build GRCh38
rs749796122 Chr.15: 81136629-81136641 on Build GRCh38
rs757142572 Chr.15: 81136681-81136681 on Build GRCh38
rs1045852284 Chr.15: 81136633-81136633 on Build GRCh38
rs770868343 Chr.15: 81136579-81136579 on Build GRCh38
rs921756799 Chr.15: 81136639-81136639 on Build GRCh38
rs562300386 Chr.15: 81136864-81136864 on Build GRCh38
rs375594608 Chr.15: 81136790-81136790 on Build GRCh38
rs576400926 Chr.15: 81136456-81136456 on Build GRCh38
rs752084250 Chr.15: 81136517-81136517 on Build GRCh38
rs374697061 Chr.15: 81136672-81136672 on Build GRCh38
rs539826859 Chr.15: 81136481-81136481 on Build GRCh38
rs1032843778 Chr.15: 81136846-81136846 on Build GRCh38

View Full Product Details