Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11678706-11679215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNN
Entrez Gene ID
8303
Gene Name
stannin
Gene Aliases
-
UniGene
Hs.618526
Gene Chromosome Location
Chr.16: 11668433-11679159 on Build GRCh38
Transcripts
NM_003498.5 XM_017023741.1 XM_017023742.1 NM_001303447.1 NM_001324022.1 NM_001324024.1 NM_001324025.1 NM_015914.6 NR_136671.1 NR_136672.1 NR_136673.1 NR_136674.1 XM_011522515.2 XM_011522516.2 XM_017023268.1

Gene Symbol
TXNDC11
Entrez Gene ID
51061
Gene Name
thioredoxin domain containing 11
Gene Aliases
EFP1
UniGene
Hs.313847
Gene Chromosome Location
Chr.16: 11679080-11742878 on Build GRCh38
Transcripts
NM_003498.5 XM_017023741.1 XM_017023742.1 NM_001303447.1 NM_001324022.1 NM_001324024.1 NM_001324025.1 NM_015914.6 NR_136671.1 NR_136672.1 NR_136673.1 NR_136674.1 XM_011522515.2 XM_011522516.2 XM_017023268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
286214 c.2862G>T p.E954D Chr.16: 11679210-11679210 on Build GRCh38
4873094 c.2943G>T p.E981D Chr.16: 11679210-11679210 on Build GRCh38
5887451 c.2864A>C p.N955T Chr.16: 11679208-11679208 on Build GRCh38
5887452 c.2945A>C p.N982T Chr.16: 11679208-11679208 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959958782 Chr.16: 11679079-11679079 on Build GRCh38
rs773236199 Chr.16: 11678982-11678982 on Build GRCh38
rs948235118 Chr.16: 11679114-11679114 on Build GRCh38
rs920660959 Chr.16: 11678747-11678747 on Build GRCh38
rs745947611 Chr.16: 11679075-11679075 on Build GRCh38
rs1002367926 Chr.16: 11678987-11678988 on Build GRCh38
rs928379592 Chr.16: 11678772-11678772 on Build GRCh38
rs930740019 Chr.16: 11678750-11678750 on Build GRCh38
rs961603310 Chr.16: 11679106-11679109 on Build GRCh38
rs200077141 Chr.16: 11678744-11678744 on Build GRCh38
rs1033460972 Chr.16: 11679002-11679002 on Build GRCh38
rs865909922 Chr.16: 11678995-11678995 on Build GRCh38
rs961723015 Chr.16: 11678752-11678752 on Build GRCh38
rs971724391 Chr.16: 11678771-11678771 on Build GRCh38
rs542395467 Chr.16: 11678735-11678735 on Build GRCh38
rs149601553 Chr.16: 11678741-11678742 on Build GRCh38
rs1052407578 Chr.16: 11678902-11678902 on Build GRCh38
rs1057045189 Chr.16: 11678786-11678786 on Build GRCh38
rs772120791 Chr.16: 11679108-11679108 on Build GRCh38
rs1013446119 Chr.16: 11679096-11679096 on Build GRCh38
rs8191347 Chr.16: 11678745-11678746 on Build GRCh38
rs1029015595 Chr.16: 11678972-11678972 on Build GRCh38
rs920592963 Chr.16: 11679130-11679130 on Build GRCh38
rs938422915 Chr.16: 11678775-11678775 on Build GRCh38
rs952114636 Chr.16: 11679170-11679170 on Build GRCh38
rs1015506281 Chr.16: 11679105-11679105 on Build GRCh38
rs757416382 Chr.16: 11679162-11679162 on Build GRCh38
rs111726209 Chr.16: 11678744-11678745 on Build GRCh38
rs997640834 Chr.16: 11678922-11678922 on Build GRCh38
rs572417689 Chr.16: 11678888-11678888 on Build GRCh38
rs1016144892 Chr.16: 11679006-11679006 on Build GRCh38
rs562437497 Chr.16: 11679126-11679126 on Build GRCh38
rs746114850 Chr.16: 11679181-11679181 on Build GRCh38
rs953202235 Chr.16: 11678729-11678729 on Build GRCh38
rs36012200 Chr.16: 11679147-11679148 on Build GRCh38
rs34330598 Chr.16: 11678921-11678922 on Build GRCh38
rs972161442 Chr.16: 11679050-11679050 on Build GRCh38
rs901942719 Chr.16: 11678913-11678913 on Build GRCh38
rs35393498 Chr.16: 11678766-11678766 on Build GRCh38
rs979561855 Chr.16: 11679138-11679138 on Build GRCh38
rs903564361 Chr.16: 11678954-11678954 on Build GRCh38
rs1044040273 Chr.16: 11678860-11678860 on Build GRCh38
rs960523764 Chr.16: 11679063-11679063 on Build GRCh38
rs545731522 Chr.16: 11679125-11679125 on Build GRCh38
rs550070163 Chr.16: 11679047-11679047 on Build GRCh38
rs948181669 Chr.16: 11678849-11678849 on Build GRCh38
rs141628246 Chr.16: 11679191-11679191 on Build GRCh38
rs774007294 Chr.16: 11678740-11678741 on Build GRCh38
rs555841878 Chr.16: 11678810-11678810 on Build GRCh38
rs1043803990 Chr.16: 11678953-11678953 on Build GRCh38
rs986177858 Chr.16: 11679171-11679171 on Build GRCh38
rs1009209090 Chr.16: 11678910-11678910 on Build GRCh38
rs868546249 Chr.16: 11678826-11678826 on Build GRCh38
rs8191348 Chr.16: 11679097-11679097 on Build GRCh38
rs747921672 Chr.16: 11678828-11678828 on Build GRCh38
rs910713762 Chr.16: 11678759-11678760 on Build GRCh38
rs904095503 Chr.16: 11678887-11678887 on Build GRCh38
rs188239514 Chr.16: 11678998-11678998 on Build GRCh38
rs779075493 Chr.16: 11679180-11679180 on Build GRCh38
rs752000865 Chr.16: 11679182-11679184 on Build GRCh38
rs984697389 Chr.16: 11678730-11678730 on Build GRCh38
rs761249934 Chr.16: 11678823-11678823 on Build GRCh38
rs370523801 Chr.16: 11679027-11679027 on Build GRCh38

View Full Product Details