Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 14435567-14436097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARN
Entrez Gene ID
5073
Gene Name
poly(A)-specific ribonuclease
Gene Aliases
DAN, DKCB6, PFBMFT4
UniGene
Hs.253197
Gene Chromosome Location
Chr.16: 14435700-14630277 on Build GRCh38
Transcripts
NM_001134477.2 NM_001242992.1 NM_002582.3

SNP Detail

SNP ID Chromosome Location
rs1004943385 Chr.16: 14435903-14435903 on Build GRCh38
rs987635115 Chr.16: 14435764-14435764 on Build GRCh38
rs914964211 Chr.16: 14436011-14436011 on Build GRCh38
rs540791345 Chr.16: 14436044-14436044 on Build GRCh38
rs748380192 Chr.16: 14435899-14435904 on Build GRCh38
rs750015711 Chr.16: 14435788-14435788 on Build GRCh38
rs751276606 Chr.16: 14435830-14435830 on Build GRCh38
rs924068361 Chr.16: 14435990-14435990 on Build GRCh38
rs950801542 Chr.16: 14435699-14435699 on Build GRCh38
rs535190492 Chr.16: 14435703-14435703 on Build GRCh38
rs77289113 Chr.16: 14435941-14435941 on Build GRCh38
rs922164295 Chr.16: 14435728-14435728 on Build GRCh38
rs778320819 Chr.16: 14436032-14436032 on Build GRCh38
rs557670454 Chr.16: 14435915-14435915 on Build GRCh38
rs906349756 Chr.16: 14435673-14435674 on Build GRCh38
rs57289814 Chr.16: 14435945-14435945 on Build GRCh38
rs374288002 Chr.16: 14435897-14435898 on Build GRCh38
rs963982745 Chr.16: 14435724-14435724 on Build GRCh38
rs763074608 Chr.16: 14435897-14435904 on Build GRCh38
rs71373026 Chr.16: 14435897-14435902 on Build GRCh38
rs1051538438 Chr.16: 14435593-14435593 on Build GRCh38
rs754634940 Chr.16: 14436029-14436029 on Build GRCh38
rs559142230 Chr.16: 14436069-14436069 on Build GRCh38
rs956680069 Chr.16: 14436002-14436002 on Build GRCh38
rs1053448125 Chr.16: 14435636-14435636 on Build GRCh38
rs1053865624 Chr.16: 14435799-14435799 on Build GRCh38
rs1003320116 Chr.16: 14435674-14435674 on Build GRCh38
rs77726332 Chr.16: 14435897-14435897 on Build GRCh38
rs893449808 Chr.16: 14435801-14435801 on Build GRCh38
rs939230622 Chr.16: 14436040-14436040 on Build GRCh38
rs564780605 Chr.16: 14435995-14435995 on Build GRCh38
rs879118030 Chr.16: 14435897-14435900 on Build GRCh38
rs140287792 Chr.16: 14435858-14435858 on Build GRCh38
rs996561862 Chr.16: 14435953-14435953 on Build GRCh38
rs893722874 Chr.16: 14435647-14435648 on Build GRCh38
rs902245710 Chr.16: 14435618-14435618 on Build GRCh38
rs57360536 Chr.16: 14435949-14435950 on Build GRCh38
rs774652867 Chr.16: 14435934-14435940 on Build GRCh38
rs538421852 Chr.16: 14435876-14435878 on Build GRCh38
rs1046896580 Chr.16: 14436060-14436060 on Build GRCh38
rs765070890 Chr.16: 14435688-14435688 on Build GRCh38
rs546515145 Chr.16: 14435803-14435803 on Build GRCh38
rs543451522 Chr.16: 14435949-14435949 on Build GRCh38
rs886527076 Chr.16: 14435859-14435859 on Build GRCh38
rs553620636 Chr.16: 14435821-14435821 on Build GRCh38
rs576538524 Chr.16: 14436033-14436033 on Build GRCh38
rs935036053 Chr.16: 14435789-14435789 on Build GRCh38
rs947771857 Chr.16: 14436019-14436019 on Build GRCh38
rs989217221 Chr.16: 14436010-14436010 on Build GRCh38
rs1029286779 Chr.16: 14435961-14435961 on Build GRCh38
rs575158231 Chr.16: 14435842-14435842 on Build GRCh38
rs767942651 Chr.16: 14435896-14435897 on Build GRCh38
rs1023907873 Chr.16: 14435661-14435661 on Build GRCh38
rs1045020162 Chr.16: 14435804-14435804 on Build GRCh38
rs1012127086 Chr.16: 14435654-14435654 on Build GRCh38
rs1016688758 Chr.16: 14435931-14435931 on Build GRCh38
rs570963176 Chr.16: 14435687-14435687 on Build GRCh38
rs923718496 Chr.16: 14435771-14435771 on Build GRCh38
rs761786111 Chr.16: 14435672-14435672 on Build GRCh38
rs773144491 Chr.16: 14435934-14435938 on Build GRCh38
rs149952396 Chr.16: 14435620-14435620 on Build GRCh38
rs1005781930 Chr.16: 14436072-14436072 on Build GRCh38
rs933499457 Chr.16: 14435759-14435759 on Build GRCh38
rs775224304 Chr.16: 14435716-14435716 on Build GRCh38
rs987644950 Chr.16: 14435987-14435990 on Build GRCh38
rs983864906 Chr.16: 14435704-14435704 on Build GRCh38
rs974991770 Chr.16: 14435727-14435727 on Build GRCh38
rs906443554 Chr.16: 14435811-14435811 on Build GRCh38
rs954933256 Chr.16: 14435967-14435967 on Build GRCh38

View Full Product Details