Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 14672467-14672977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G10
Entrez Gene ID
8399
Gene Name
phospholipase A2 group X
Gene Aliases
GXPLA2, GXSPLA2, SPLA2, sPLA2-X
UniGene
Hs.567366
Gene Chromosome Location
Chr.16: 14672548-14694663 on Build GRCh38
Transcripts
NM_003561.2 NR_133651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4714881 c.361G>A p.A121T Chr.16: 14672744-14672744 on Build GRCh38
7325941 c.396_397delTG p.C132fs*1 Chr.16: 14672708-14672709 on Build GRCh38
4058461 c.402G>T p.Q134H Chr.16: 14672703-14672703 on Build GRCh38
4714880 c.379G>T p.E127* Chr.16: 14672726-14672726 on Build GRCh38
6078353 c.450C>G p.L150L Chr.16: 14672655-14672655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772560235 Chr.16: 14672574-14672574 on Build GRCh38
rs995585310 Chr.16: 14672730-14672730 on Build GRCh38
rs752005685 Chr.16: 14672790-14672790 on Build GRCh38
rs1005629665 Chr.16: 14672575-14672575 on Build GRCh38
rs755321415 Chr.16: 14672666-14672666 on Build GRCh38
rs776877289 Chr.16: 14672600-14672600 on Build GRCh38
rs116330346 Chr.16: 14672546-14672546 on Build GRCh38
rs763526715 Chr.16: 14672859-14672859 on Build GRCh38
rs767537868 Chr.16: 14672627-14672627 on Build GRCh38
rs778179831 Chr.16: 14672605-14672608 on Build GRCh38
rs929443433 Chr.16: 14672829-14672829 on Build GRCh38
rs760698070 Chr.16: 14672738-14672738 on Build GRCh38
rs1040475197 Chr.16: 14672789-14672789 on Build GRCh38
rs763817301 Chr.16: 14672635-14672635 on Build GRCh38
rs987957414 Chr.16: 14672677-14672677 on Build GRCh38
rs558733732 Chr.16: 14672522-14672522 on Build GRCh38
rs1008026371 Chr.16: 14672906-14672906 on Build GRCh38
rs139270637 Chr.16: 14672734-14672734 on Build GRCh38
rs542572627 Chr.16: 14672916-14672916 on Build GRCh38
rs760505520 Chr.16: 14672633-14672633 on Build GRCh38
rs531379939 Chr.16: 14672935-14672935 on Build GRCh38
rs944764933 Chr.16: 14672788-14672788 on Build GRCh38
rs764674494 Chr.16: 14672780-14672780 on Build GRCh38
rs150828124 Chr.16: 14672731-14672731 on Build GRCh38
rs780599326 Chr.16: 14672580-14672580 on Build GRCh38
rs1037040000 Chr.16: 14672700-14672700 on Build GRCh38
rs530932954 Chr.16: 14672828-14672828 on Build GRCh38
rs757956029 Chr.16: 14672643-14672643 on Build GRCh38
rs113572525 Chr.16: 14672571-14672571 on Build GRCh38
rs780511582 Chr.16: 14672658-14672658 on Build GRCh38
rs749924257 Chr.16: 14672642-14672642 on Build GRCh38
rs549762676 Chr.16: 14672947-14672947 on Build GRCh38
rs764777766 Chr.16: 14672760-14672760 on Build GRCh38
rs192235371 Chr.16: 14672564-14672564 on Build GRCh38
rs755445261 Chr.16: 14672798-14672798 on Build GRCh38
rs776282626 Chr.16: 14672753-14672753 on Build GRCh38
rs896748790 Chr.16: 14672606-14672606 on Build GRCh38
rs1049299151 Chr.16: 14672870-14672870 on Build GRCh38
rs367681146 Chr.16: 14672673-14672673 on Build GRCh38
rs143240053 Chr.16: 14672917-14672917 on Build GRCh38
rs199646389 Chr.16: 14672563-14672563 on Build GRCh38
rs761723158 Chr.16: 14672759-14672759 on Build GRCh38
rs753399179 Chr.16: 14672629-14672629 on Build GRCh38
rs757467624 Chr.16: 14672761-14672761 on Build GRCh38
rs748340733 Chr.16: 14672602-14672602 on Build GRCh38
rs375494472 Chr.16: 14672723-14672723 on Build GRCh38
rs773616537 Chr.16: 14672716-14672716 on Build GRCh38
rs372117900 Chr.16: 14672515-14672515 on Build GRCh38
rs753942842 Chr.16: 14672718-14672719 on Build GRCh38
rs757868056 Chr.16: 14672779-14672779 on Build GRCh38
rs981900332 Chr.16: 14672628-14672628 on Build GRCh38
rs576069069 Chr.16: 14672511-14672511 on Build GRCh38
rs915154882 Chr.16: 14672696-14672696 on Build GRCh38
rs149977312 Chr.16: 14672745-14672745 on Build GRCh38
rs140187592 Chr.16: 14672622-14672622 on Build GRCh38
rs372227161 Chr.16: 14672717-14672717 on Build GRCh38
rs200010548 Chr.16: 14672623-14672623 on Build GRCh38
rs543507853 Chr.16: 14672516-14672516 on Build GRCh38
rs761574144 Chr.16: 14672721-14672721 on Build GRCh38
rs779279208 Chr.16: 14672773-14672774 on Build GRCh38
rs775520047 Chr.16: 14672729-14672729 on Build GRCh38
rs142440622 Chr.16: 14672637-14672637 on Build GRCh38
rs750118985 Chr.16: 14672777-14672777 on Build GRCh38
rs747549327 Chr.16: 14672664-14672664 on Build GRCh38
rs777450617 Chr.16: 14672562-14672562 on Build GRCh38
rs139866515 Chr.16: 14672672-14672672 on Build GRCh38
rs200070548 Chr.16: 14672719-14672719 on Build GRCh38
rs769905506 Chr.16: 14672695-14672695 on Build GRCh38
rs753468912 Chr.16: 14672636-14672636 on Build GRCh38
rs1019913508 Chr.16: 14672914-14672914 on Build GRCh38
rs747354625 Chr.16: 14672596-14672596 on Build GRCh38
rs769855614 Chr.16: 14672611-14672611 on Build GRCh38
rs370170527 Chr.16: 14672565-14672565 on Build GRCh38
rs760373314 Chr.16: 14672542-14672542 on Build GRCh38
rs147705409 Chr.16: 14672747-14672747 on Build GRCh38

View Full Product Details