Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15055801-15056302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTAN1
Entrez Gene ID
123803
Gene Name
N-terminal asparagine amidase
Gene Aliases
PNAA, PNAD
UniGene
Hs.592045
Gene Chromosome Location
Chr.16: 15037853-15056079 on Build GRCh38
Transcripts
NM_001270766.1 NM_001270767.1 NM_173474.3 NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1

Gene Symbol
PDXDC1
Entrez Gene ID
23042
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Gene Aliases
LP8165
UniGene
Hs.370781
Gene Chromosome Location
Chr.16: 14974591-15139410 on Build GRCh38
Transcripts
NM_001270766.1 NM_001270767.1 NM_173474.3 NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
399320 c.40C>T p.Q14* Chr.16: 15055932-15055932 on Build GRCh38
5657724 c.39G>T p.P13P Chr.16: 15055933-15055933 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009773057 Chr.16: 15055921-15055921 on Build GRCh38
rs537239666 Chr.16: 15056086-15056086 on Build GRCh38
rs368851607 Chr.16: 15055822-15055822 on Build GRCh38
rs917717457 Chr.16: 15055981-15055981 on Build GRCh38
rs1019039096 Chr.16: 15055917-15055917 on Build GRCh38
rs1040759827 Chr.16: 15056126-15056126 on Build GRCh38
rs912921513 Chr.16: 15056214-15056214 on Build GRCh38
rs1043206481 Chr.16: 15056241-15056241 on Build GRCh38
rs972637035 Chr.16: 15056009-15056009 on Build GRCh38
rs902106682 Chr.16: 15055920-15055920 on Build GRCh38
rs1020989567 Chr.16: 15055936-15055936 on Build GRCh38
rs952771056 Chr.16: 15056146-15056146 on Build GRCh38
rs889522149 Chr.16: 15055867-15055867 on Build GRCh38
rs943751846 Chr.16: 15056013-15056013 on Build GRCh38
rs984186894 Chr.16: 15056149-15056149 on Build GRCh38
rs528448907 Chr.16: 15056033-15056034 on Build GRCh38
rs554704790 Chr.16: 15056228-15056228 on Build GRCh38
rs376534889 Chr.16: 15056163-15056163 on Build GRCh38
rs757458131 Chr.16: 15056262-15056262 on Build GRCh38
rs559213237 Chr.16: 15056099-15056099 on Build GRCh38
rs930992804 Chr.16: 15056011-15056012 on Build GRCh38
rs919851607 Chr.16: 15056079-15056079 on Build GRCh38
rs1052798138 Chr.16: 15056180-15056180 on Build GRCh38
rs895139172 Chr.16: 15056213-15056213 on Build GRCh38
rs1013704640 Chr.16: 15056226-15056226 on Build GRCh38
rs943826209 Chr.16: 15056101-15056101 on Build GRCh38
rs139773894 Chr.16: 15056017-15056017 on Build GRCh38
rs571011250 Chr.16: 15056131-15056131 on Build GRCh38
rs979562568 Chr.16: 15055956-15055956 on Build GRCh38
rs968441723 Chr.16: 15055938-15055938 on Build GRCh38
rs959444079 Chr.16: 15056040-15056041 on Build GRCh38
rs994414997 Chr.16: 15056277-15056277 on Build GRCh38
rs889620325 Chr.16: 15056253-15056253 on Build GRCh38
rs972570774 Chr.16: 15056074-15056074 on Build GRCh38
rs889542588 Chr.16: 15056012-15056012 on Build GRCh38
rs904080115 Chr.16: 15056030-15056030 on Build GRCh38
rs916713115 Chr.16: 15056159-15056159 on Build GRCh38
rs1049376975 Chr.16: 15056012-15056012 on Build GRCh38
rs961426839 Chr.16: 15056274-15056274 on Build GRCh38
rs939958871 Chr.16: 15055991-15055991 on Build GRCh38
rs572057307 Chr.16: 15056179-15056179 on Build GRCh38
rs1033649420 Chr.16: 15056040-15056040 on Build GRCh38
rs1001284590 Chr.16: 15056031-15056031 on Build GRCh38
rs3934730 Chr.16: 15056250-15056250 on Build GRCh38
rs992240739 Chr.16: 15056053-15056053 on Build GRCh38
rs939883122 Chr.16: 15055833-15055833 on Build GRCh38
rs910888623 Chr.16: 15056094-15056094 on Build GRCh38
rs553770544 Chr.16: 15056165-15056165 on Build GRCh38
rs1025672332 Chr.16: 15056137-15056137 on Build GRCh38
rs919630328 Chr.16: 15056011-15056011 on Build GRCh38
rs1026780726 Chr.16: 15056279-15056279 on Build GRCh38
rs1024848000 Chr.16: 15056061-15056061 on Build GRCh38
rs959413405 Chr.16: 15055963-15055963 on Build GRCh38
rs182927365 Chr.16: 15056223-15056223 on Build GRCh38
rs1040561497 Chr.16: 15056015-15056015 on Build GRCh38
rs566737619 Chr.16: 15055827-15055827 on Build GRCh38
rs991998119 Chr.16: 15055972-15055972 on Build GRCh38
rs985533354 Chr.16: 15056093-15056093 on Build GRCh38
rs538527904 Chr.16: 15055858-15055858 on Build GRCh38
rs961186310 Chr.16: 15056069-15056069 on Build GRCh38
rs969966368 Chr.16: 15056162-15056162 on Build GRCh38
rs1049926071 Chr.16: 15055863-15055863 on Build GRCh38
rs902013703 Chr.16: 15056016-15056016 on Build GRCh38
rs1036699043 Chr.16: 15055840-15055840 on Build GRCh38
rs569962511 Chr.16: 15056043-15056043 on Build GRCh38
rs1001082482 Chr.16: 15056192-15056192 on Build GRCh38
rs898150282 Chr.16: 15055848-15055848 on Build GRCh38
rs943079880 Chr.16: 15056049-15056049 on Build GRCh38
rs1055574613 Chr.16: 15056193-15056193 on Build GRCh38
rs1007697441 Chr.16: 15055880-15055880 on Build GRCh38
rs972734238 Chr.16: 15055842-15055842 on Build GRCh38

View Full Product Details