Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15407421-15407929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPV17L
Entrez Gene ID
255027
Gene Name
MPV17 mitochondrial inner membrane protein like
Gene Aliases
M-LPH, MLPH1, MLPH2, MPV17L1
UniGene
Hs.720673
Gene Chromosome Location
Chr.16: 15395569-15409686 on Build GRCh38
Transcripts
NM_001128423.1 NM_173803.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263362 c.396C>T p.Y132Y Chr.16: 15407838-15407838 on Build GRCh38
7263363 c.325C>T p.L109L Chr.16: 15407838-15407838 on Build GRCh38
1708766 c.401_402CC>TT p.P134L Chr.16: 15407843-15407844 on Build GRCh38
1708767 c.330_331CC>TT p.L111F Chr.16: 15407843-15407844 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547566231 Chr.16: 15407482-15407482 on Build GRCh38
rs763391285 Chr.16: 15407861-15407861 on Build GRCh38
rs745314811 Chr.16: 15407802-15407802 on Build GRCh38
rs770118742 Chr.16: 15407888-15407888 on Build GRCh38
rs777792683 Chr.16: 15407839-15407840 on Build GRCh38
rs751143877 Chr.16: 15407871-15407871 on Build GRCh38
rs111659587 Chr.16: 15407890-15407890 on Build GRCh38
rs768868139 Chr.16: 15407817-15407817 on Build GRCh38
rs554916773 Chr.16: 15407529-15407529 on Build GRCh38
rs745894143 Chr.16: 15407709-15407709 on Build GRCh38
rs1035389505 Chr.16: 15407455-15407455 on Build GRCh38
rs5815831 Chr.16: 15407513-15407514 on Build GRCh38
rs777952939 Chr.16: 15407889-15407889 on Build GRCh38
rs868722122 Chr.16: 15407466-15407466 on Build GRCh38
rs567261988 Chr.16: 15407577-15407577 on Build GRCh38
rs753612895 Chr.16: 15407851-15407851 on Build GRCh38
rs531756682 Chr.16: 15407868-15407868 on Build GRCh38
rs200953251 Chr.16: 15407802-15407803 on Build GRCh38
rs554007786 Chr.16: 15407715-15407715 on Build GRCh38
rs376653858 Chr.16: 15407843-15407843 on Build GRCh38
rs1027935557 Chr.16: 15407532-15407532 on Build GRCh38
rs781560059 Chr.16: 15407876-15407876 on Build GRCh38
rs778444567 Chr.16: 15407800-15407800 on Build GRCh38
rs371219439 Chr.16: 15407862-15407862 on Build GRCh38
rs190144901 Chr.16: 15407587-15407587 on Build GRCh38
rs773493839 Chr.16: 15407824-15407824 on Build GRCh38
rs572099410 Chr.16: 15407812-15407812 on Build GRCh38
rs149762442 Chr.16: 15407462-15407462 on Build GRCh38
rs752247621 Chr.16: 15407812-15407813 on Build GRCh38
rs764938039 Chr.16: 15407858-15407858 on Build GRCh38
rs761517541 Chr.16: 15407856-15407856 on Build GRCh38
rs185335553 Chr.16: 15407486-15407486 on Build GRCh38
rs369371748 Chr.16: 15407816-15407816 on Build GRCh38
rs763962358 Chr.16: 15407848-15407848 on Build GRCh38
rs577798247 Chr.16: 15407650-15407650 on Build GRCh38
rs62039098 Chr.16: 15407484-15407484 on Build GRCh38
rs749554728 Chr.16: 15407895-15407895 on Build GRCh38
rs781577467 Chr.16: 15407822-15407822 on Build GRCh38
rs141439179 Chr.16: 15407834-15407834 on Build GRCh38
rs371983552 Chr.16: 15407454-15407454 on Build GRCh38
rs1036809394 Chr.16: 15407838-15407838 on Build GRCh38
rs144408494 Chr.16: 15407823-15407823 on Build GRCh38
rs753608097 Chr.16: 15407781-15407781 on Build GRCh38
rs986484383 Chr.16: 15407580-15407580 on Build GRCh38
rs755460574 Chr.16: 15407874-15407874 on Build GRCh38
rs756885709 Chr.16: 15407791-15407791 on Build GRCh38
rs148793965 Chr.16: 15407831-15407831 on Build GRCh38
rs748545952 Chr.16: 15407877-15407877 on Build GRCh38
rs148942772 Chr.16: 15407844-15407844 on Build GRCh38
rs780692155 Chr.16: 15407815-15407815 on Build GRCh38
rs771175224 Chr.16: 15407906-15407906 on Build GRCh38
rs962415137 Chr.16: 15407523-15407530 on Build GRCh38
rs542512244 Chr.16: 15407801-15407802 on Build GRCh38
rs796391901 Chr.16: 15407802-15407803 on Build GRCh38
rs545263772 Chr.16: 15407670-15407670 on Build GRCh38
rs1004154286 Chr.16: 15407835-15407835 on Build GRCh38
rs946771170 Chr.16: 15407748-15407749 on Build GRCh38
rs763257092 Chr.16: 15407802-15407802 on Build GRCh38

View Full Product Details