Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 17108845-17109329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XYLT1
Entrez Gene ID
64131
Gene Name
xylosyltransferase 1
Gene Aliases
DBQD2, PXYLT1, XT-I, XT1, XTI, XYLTI, xylT-I
UniGene
Hs.22907
Gene Chromosome Location
Chr.16: 17102324-17470934 on Build GRCh38
Transcripts
NM_022166.3 XM_017023540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6856050 c.2681A>G p.N894S Chr.16: 17108894-17108894 on Build GRCh38
5541870 c.2691C>T p.S897S Chr.16: 17108884-17108884 on Build GRCh38
1233000 c.2723C>T p.S908L Chr.16: 17108852-17108852 on Build GRCh38
5909547 c.2604G>A p.M868I Chr.16: 17108971-17108971 on Build GRCh38
4821082 c.2637C>T p.L879L Chr.16: 17108938-17108938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771630753 Chr.16: 17109220-17109220 on Build GRCh38
rs1038115539 Chr.16: 17109290-17109290 on Build GRCh38
rs375238626 Chr.16: 17108985-17108985 on Build GRCh38
rs553595317 Chr.16: 17109283-17109283 on Build GRCh38
rs767728751 Chr.16: 17108972-17108972 on Build GRCh38
rs149423967 Chr.16: 17108909-17108909 on Build GRCh38
rs772914063 Chr.16: 17109023-17109023 on Build GRCh38
rs767933314 Chr.16: 17109038-17109038 on Build GRCh38
rs386789435 Chr.16: 17109107-17109108 on Build GRCh38
rs200752508 Chr.16: 17109066-17109066 on Build GRCh38
rs769733883 Chr.16: 17108872-17108872 on Build GRCh38
rs375566768 Chr.16: 17109026-17109026 on Build GRCh38
rs780187085 Chr.16: 17108992-17108992 on Build GRCh38
rs572010848 Chr.16: 17108928-17108928 on Build GRCh38
rs141353269 Chr.16: 17108881-17108881 on Build GRCh38
rs940580106 Chr.16: 17109078-17109078 on Build GRCh38
rs558755151 Chr.16: 17109226-17109226 on Build GRCh38
rs772538035 Chr.16: 17108941-17108941 on Build GRCh38
rs560977789 Chr.16: 17108947-17108947 on Build GRCh38
rs372238772 Chr.16: 17109025-17109025 on Build GRCh38
rs979080952 Chr.16: 17108933-17108933 on Build GRCh38
rs564847916 Chr.16: 17109035-17109035 on Build GRCh38
rs368097793 Chr.16: 17108882-17108882 on Build GRCh38
rs768545631 Chr.16: 17108927-17108927 on Build GRCh38
rs1015588911 Chr.16: 17109297-17109297 on Build GRCh38
rs776112557 Chr.16: 17108885-17108885 on Build GRCh38
rs370964557 Chr.16: 17109004-17109004 on Build GRCh38
rs761331511 Chr.16: 17108930-17108930 on Build GRCh38
rs933523803 Chr.16: 17109234-17109234 on Build GRCh38
rs756228697 Chr.16: 17108981-17108981 on Build GRCh38
rs752844900 Chr.16: 17108910-17108910 on Build GRCh38
rs761434857 Chr.16: 17108892-17108892 on Build GRCh38
rs190476242 Chr.16: 17109191-17109191 on Build GRCh38
rs199588257 Chr.16: 17109014-17109014 on Build GRCh38
rs923514488 Chr.16: 17109230-17109230 on Build GRCh38
rs538813398 Chr.16: 17109114-17109114 on Build GRCh38
rs34511974 Chr.16: 17108944-17108944 on Build GRCh38
rs548885176 Chr.16: 17109118-17109118 on Build GRCh38
rs879420215 Chr.16: 17109107-17109108 on Build GRCh38
rs769620977 Chr.16: 17108936-17108936 on Build GRCh38
rs780133822 Chr.16: 17108926-17108926 on Build GRCh38
rs942091422 Chr.16: 17109274-17109274 on Build GRCh38
rs184391112 Chr.16: 17108873-17108873 on Build GRCh38
rs749059906 Chr.16: 17109083-17109083 on Build GRCh38
rs537585931 Chr.16: 17108870-17108870 on Build GRCh38
rs924260 Chr.16: 17108920-17108920 on Build GRCh38
rs570864241 Chr.16: 17109246-17109246 on Build GRCh38
rs537072873 Chr.16: 17109208-17109208 on Build GRCh38
rs924259 Chr.16: 17109108-17109108 on Build GRCh38
rs757379467 Chr.16: 17108990-17108990 on Build GRCh38
rs143481827 Chr.16: 17108919-17108919 on Build GRCh38
rs114032393 Chr.16: 17109225-17109225 on Build GRCh38
rs1047727049 Chr.16: 17108869-17108869 on Build GRCh38
rs1047871736 Chr.16: 17109250-17109250 on Build GRCh38
rs1016839043 Chr.16: 17108879-17108879 on Build GRCh38
rs375282606 Chr.16: 17109253-17109254 on Build GRCh38
rs35309694 Chr.16: 17108900-17108900 on Build GRCh38
rs752999045 Chr.16: 17108976-17108976 on Build GRCh38
rs755332140 Chr.16: 17108907-17108907 on Build GRCh38
rs769391314 Chr.16: 17109015-17109015 on Build GRCh38
rs1033934388 Chr.16: 17109102-17109102 on Build GRCh38
rs772958671 Chr.16: 17109196-17109196 on Build GRCh38
rs764602771 Chr.16: 17108893-17108893 on Build GRCh38
rs966434051 Chr.16: 17109122-17109122 on Build GRCh38
rs979112291 Chr.16: 17109124-17109124 on Build GRCh38
rs139025663 Chr.16: 17108901-17108901 on Build GRCh38
rs768526010 Chr.16: 17109001-17109001 on Build GRCh38
rs894193236 Chr.16: 17109117-17109117 on Build GRCh38
rs775364743 Chr.16: 17108876-17108876 on Build GRCh38
rs6498661 Chr.16: 17109107-17109107 on Build GRCh38
rs780784292 Chr.16: 17109009-17109009 on Build GRCh38
rs1042687620 Chr.16: 17109166-17109166 on Build GRCh38
rs1037647981 Chr.16: 17109047-17109047 on Build GRCh38
rs531190343 Chr.16: 17108984-17108984 on Build GRCh38
rs202017658 Chr.16: 17108943-17108943 on Build GRCh38
rs556062365 Chr.16: 17109263-17109263 on Build GRCh38
rs746893762 Chr.16: 17108993-17108993 on Build GRCh38

View Full Product Details