Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19022723-19023238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC7
Entrez Gene ID
79905
Gene Name
transmembrane channel like 7
Gene Aliases
-
UniGene
Hs.187377
Gene Chromosome Location
Chr.16: 18983934-19063942 on Build GRCh38
Transcripts
NM_001160364.2 NM_001300732.1 NM_001324263.1 NM_001324265.1 NM_001324268.1 NM_024847.4 NR_136733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
967822 c.652G>T p.V218L Chr.16: 19023136-19023136 on Build GRCh38
5936242 c.665G>A p.G222E Chr.16: 19023149-19023149 on Build GRCh38
3927534 c.651A>G p.P217P Chr.16: 19023135-19023135 on Build GRCh38
3506628 c.690C>T p.I230I Chr.16: 19023174-19023174 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760020258 Chr.16: 19023111-19023111 on Build GRCh38
rs745952582 Chr.16: 19023075-19023075 on Build GRCh38
rs1027239706 Chr.16: 19022819-19022819 on Build GRCh38
rs186616626 Chr.16: 19022966-19022966 on Build GRCh38
rs752331207 Chr.16: 19022881-19022881 on Build GRCh38
rs374105814 Chr.16: 19023210-19023210 on Build GRCh38
rs527628140 Chr.16: 19023191-19023191 on Build GRCh38
rs1035109737 Chr.16: 19022968-19022968 on Build GRCh38
rs921676046 Chr.16: 19022976-19022976 on Build GRCh38
rs868176971 Chr.16: 19023107-19023107 on Build GRCh38
rs1040105306 Chr.16: 19022852-19022852 on Build GRCh38
rs971035174 Chr.16: 19022894-19022894 on Build GRCh38
rs187461695 Chr.16: 19023123-19023123 on Build GRCh38
rs780892783 Chr.16: 19023209-19023209 on Build GRCh38
rs995550780 Chr.16: 19022789-19022789 on Build GRCh38
rs377326089 Chr.16: 19023109-19023109 on Build GRCh38
rs977748506 Chr.16: 19022906-19022906 on Build GRCh38
rs758231812 Chr.16: 19022912-19022912 on Build GRCh38
rs897241399 Chr.16: 19022946-19022946 on Build GRCh38
rs752199350 Chr.16: 19023021-19023022 on Build GRCh38
rs767915875 Chr.16: 19023121-19023121 on Build GRCh38
rs891390242 Chr.16: 19023017-19023017 on Build GRCh38
rs1050408309 Chr.16: 19022897-19022897 on Build GRCh38
rs530133143 Chr.16: 19023066-19023066 on Build GRCh38
rs576463421 Chr.16: 19022892-19022892 on Build GRCh38
rs764399604 Chr.16: 19023159-19023159 on Build GRCh38
rs927989346 Chr.16: 19022750-19022750 on Build GRCh38
rs750581075 Chr.16: 19023156-19023159 on Build GRCh38
rs190248230 Chr.16: 19023025-19023025 on Build GRCh38
rs758815804 Chr.16: 19023182-19023183 on Build GRCh38
rs760905718 Chr.16: 19023133-19023133 on Build GRCh38
rs749488629 Chr.16: 19023155-19023155 on Build GRCh38
rs1008460599 Chr.16: 19023048-19023048 on Build GRCh38
rs559461824 Chr.16: 19023065-19023065 on Build GRCh38
rs959267898 Chr.16: 19022821-19022821 on Build GRCh38
rs1015574695 Chr.16: 19022776-19022776 on Build GRCh38
rs923686524 Chr.16: 19022828-19022828 on Build GRCh38
rs766955953 Chr.16: 19023196-19023199 on Build GRCh38
rs1037035665 Chr.16: 19023053-19023053 on Build GRCh38
rs147994584 Chr.16: 19023160-19023160 on Build GRCh38
rs769906977 Chr.16: 19023076-19023076 on Build GRCh38
rs757174590 Chr.16: 19022994-19022994 on Build GRCh38
rs769187591 Chr.16: 19023192-19023192 on Build GRCh38
rs765162741 Chr.16: 19023077-19023077 on Build GRCh38
rs142240612 Chr.16: 19022918-19022918 on Build GRCh38
rs867926458 Chr.16: 19023009-19023009 on Build GRCh38
rs757418401 Chr.16: 19023173-19023173 on Build GRCh38
rs575460672 Chr.16: 19022792-19022792 on Build GRCh38
rs781120496 Chr.16: 19023006-19023006 on Build GRCh38
rs574566414 Chr.16: 19022751-19022751 on Build GRCh38
rs900135372 Chr.16: 19022856-19022856 on Build GRCh38
rs759019056 Chr.16: 19022831-19022831 on Build GRCh38
rs55743338 Chr.16: 19022840-19022840 on Build GRCh38
rs557148727 Chr.16: 19022780-19022780 on Build GRCh38
rs371162152 Chr.16: 19023068-19023068 on Build GRCh38
rs959402865 Chr.16: 19022756-19022756 on Build GRCh38
rs754944887 Chr.16: 19023207-19023207 on Build GRCh38
rs1015803755 Chr.16: 19022923-19022923 on Build GRCh38
rs772703840 Chr.16: 19023087-19023087 on Build GRCh38
rs1012107521 Chr.16: 19023167-19023167 on Build GRCh38
rs753319167 Chr.16: 19022757-19022757 on Build GRCh38
rs1022613698 Chr.16: 19023031-19023031 on Build GRCh38
rs534350387 Chr.16: 19023008-19023008 on Build GRCh38
rs182459231 Chr.16: 19023095-19023095 on Build GRCh38
rs921721514 Chr.16: 19023131-19023131 on Build GRCh38
rs757363195 Chr.16: 19023063-19023063 on Build GRCh38
rs754888785 Chr.16: 19023072-19023072 on Build GRCh38
rs1008261082 Chr.16: 19022771-19022771 on Build GRCh38
rs117741115 Chr.16: 19023084-19023084 on Build GRCh38
rs61171024 Chr.16: 19022768-19022768 on Build GRCh38
rs751306169 Chr.16: 19022935-19022935 on Build GRCh38
rs770357727 Chr.16: 19023101-19023101 on Build GRCh38

View Full Product Details