Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19030144-19030568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC7
Entrez Gene ID
79905
Gene Name
transmembrane channel like 7
Gene Aliases
-
UniGene
Hs.187377
Gene Chromosome Location
Chr.16: 18983934-19063942 on Build GRCh38
Transcripts
NM_001160364.2 NM_001300732.1 NM_001324263.1 NM_001324265.1 NM_001324268.1 NM_024847.4 NR_136733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907685 c.712-8C>T p.? Chr.16: 19030216-19030216 on Build GRCh38
4128760 c.761G>A p.G254E Chr.16: 19030273-19030273 on Build GRCh38
702603 c.755T>C p.I252T Chr.16: 19030267-19030267 on Build GRCh38
2128752 c.743G>A p.G248E Chr.16: 19030255-19030255 on Build GRCh38
2128754 c.782C>T p.T261I Chr.16: 19030294-19030294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753535460 Chr.16: 19030247-19030247 on Build GRCh38
rs766306886 Chr.16: 19030304-19030304 on Build GRCh38
rs751938939 Chr.16: 19030363-19030364 on Build GRCh38
rs752522247 Chr.16: 19030226-19030226 on Build GRCh38
rs769013357 Chr.16: 19030340-19030340 on Build GRCh38
rs772596832 Chr.16: 19030180-19030180 on Build GRCh38
rs765774247 Chr.16: 19030474-19030474 on Build GRCh38
rs771088373 Chr.16: 19030408-19030408 on Build GRCh38
rs774261095 Chr.16: 19030415-19030415 on Build GRCh38
rs1056762273 Chr.16: 19030255-19030255 on Build GRCh38
rs374745847 Chr.16: 19030254-19030254 on Build GRCh38
rs777412602 Chr.16: 19030232-19030232 on Build GRCh38
rs756000809 Chr.16: 19030230-19030230 on Build GRCh38
rs763069013 Chr.16: 19030405-19030405 on Build GRCh38
rs370887725 Chr.16: 19030185-19030185 on Build GRCh38
rs919088075 Chr.16: 19030522-19030522 on Build GRCh38
rs762167845 Chr.16: 19030297-19030297 on Build GRCh38
rs4072393 Chr.16: 19030273-19030273 on Build GRCh38
rs763142503 Chr.16: 19030303-19030303 on Build GRCh38
rs941851312 Chr.16: 19030472-19030472 on Build GRCh38
rs371761861 Chr.16: 19030310-19030310 on Build GRCh38
rs755351338 Chr.16: 19030402-19030402 on Build GRCh38
rs747601851 Chr.16: 19030334-19030334 on Build GRCh38
rs561902485 Chr.16: 19030377-19030377 on Build GRCh38
rs373579773 Chr.16: 19030272-19030273 on Build GRCh38
rs928985995 Chr.16: 19030436-19030436 on Build GRCh38
rs746410789 Chr.16: 19030179-19030179 on Build GRCh38
rs557552101 Chr.16: 19030168-19030168 on Build GRCh38
rs374196164 Chr.16: 19030361-19030361 on Build GRCh38
rs201407524 Chr.16: 19030216-19030216 on Build GRCh38
rs773576000 Chr.16: 19030395-19030395 on Build GRCh38
rs140007425 Chr.16: 19030312-19030312 on Build GRCh38
rs369657549 Chr.16: 19030421-19030421 on Build GRCh38
rs776979452 Chr.16: 19030202-19030202 on Build GRCh38
rs529007755 Chr.16: 19030442-19030442 on Build GRCh38
rs201491513 Chr.16: 19030212-19030212 on Build GRCh38
rs374240709 Chr.16: 19030209-19030209 on Build GRCh38
rs200360239 Chr.16: 19030380-19030380 on Build GRCh38
rs776839663 Chr.16: 19030295-19030295 on Build GRCh38
rs956849083 Chr.16: 19030352-19030352 on Build GRCh38
rs745403744 Chr.16: 19030256-19030256 on Build GRCh38
rs546403187 Chr.16: 19030170-19030170 on Build GRCh38
rs1045184781 Chr.16: 19030514-19030514 on Build GRCh38
rs764056388 Chr.16: 19030424-19030424 on Build GRCh38
rs769123002 Chr.16: 19030278-19030278 on Build GRCh38
rs773522471 Chr.16: 19030302-19030302 on Build GRCh38
rs141948391 Chr.16: 19030253-19030253 on Build GRCh38
rs756995644 Chr.16: 19030313-19030313 on Build GRCh38
rs1035391137 Chr.16: 19030262-19030262 on Build GRCh38
rs771425199 Chr.16: 19030269-19030269 on Build GRCh38
rs763987902 Chr.16: 19030311-19030311 on Build GRCh38
rs984530453 Chr.16: 19030439-19030439 on Build GRCh38
rs757994336 Chr.16: 19030332-19030332 on Build GRCh38
rs764300088 Chr.16: 19030197-19030197 on Build GRCh38
rs910495413 Chr.16: 19030469-19030469 on Build GRCh38
rs372138127 Chr.16: 19030217-19030217 on Build GRCh38
rs750503461 Chr.16: 19030213-19030213 on Build GRCh38
rs4381586 Chr.16: 19030169-19030169 on Build GRCh38
rs561493305 Chr.16: 19030277-19030277 on Build GRCh38
rs770230228 Chr.16: 19030387-19030387 on Build GRCh38
rs547040980 Chr.16: 19030374-19030374 on Build GRCh38
rs761114055 Chr.16: 19030192-19030192 on Build GRCh38
rs367565977 Chr.16: 19030299-19030299 on Build GRCh38
rs755461576 Chr.16: 19030331-19030331 on Build GRCh38
rs369414340 Chr.16: 19030333-19030333 on Build GRCh38
rs375390148 Chr.16: 19030329-19030329 on Build GRCh38

View Full Product Details