Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19860449-19860947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRC5B
Entrez Gene ID
51704
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
RAIG-2, RAIG2
UniGene
Hs.148685
Gene Chromosome Location
Chr.16: 19858971-19886045 on Build GRCh38
Transcripts
NM_001304771.1 NM_016235.2 XM_006721051.2 XM_006721052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
329381 c.1182G>A p.P394P Chr.16: 19860530-19860530 on Build GRCh38
6312748 c.1170C>A p.I390I Chr.16: 19860542-19860542 on Build GRCh38
2129286 c.1189C>A p.H397N Chr.16: 19860523-19860523 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756077882 Chr.16: 19860559-19860559 on Build GRCh38
rs1044739085 Chr.16: 19860844-19860844 on Build GRCh38
rs767265211 Chr.16: 19860541-19860541 on Build GRCh38
rs375866997 Chr.16: 19860573-19860573 on Build GRCh38
rs919462112 Chr.16: 19860760-19860760 on Build GRCh38
rs771851958 Chr.16: 19860626-19860626 on Build GRCh38
rs768964139 Chr.16: 19860479-19860479 on Build GRCh38
rs1011116730 Chr.16: 19860922-19860922 on Build GRCh38
rs1013651485 Chr.16: 19860702-19860702 on Build GRCh38
rs1024985812 Chr.16: 19860740-19860740 on Build GRCh38
rs746575819 Chr.16: 19860683-19860683 on Build GRCh38
rs542188869 Chr.16: 19860712-19860712 on Build GRCh38
rs1022598105 Chr.16: 19860677-19860677 on Build GRCh38
rs774970596 Chr.16: 19860625-19860625 on Build GRCh38
rs892307549 Chr.16: 19860894-19860894 on Build GRCh38
rs958564025 Chr.16: 19860824-19860824 on Build GRCh38
rs748392111 Chr.16: 19860559-19860559 on Build GRCh38
rs765254302 Chr.16: 19860568-19860568 on Build GRCh38
rs947760128 Chr.16: 19860832-19860832 on Build GRCh38
rs147744945 Chr.16: 19860531-19860531 on Build GRCh38
rs930922267 Chr.16: 19860519-19860519 on Build GRCh38
rs906191147 Chr.16: 19860845-19860845 on Build GRCh38
rs769394087 Chr.16: 19860492-19860492 on Build GRCh38
rs139694238 Chr.16: 19860538-19860538 on Build GRCh38
rs755344090 Chr.16: 19860592-19860592 on Build GRCh38
rs745813888 Chr.16: 19860779-19860779 on Build GRCh38
rs769645890 Chr.16: 19860918-19860918 on Build GRCh38
rs571222346 Chr.16: 19860859-19860859 on Build GRCh38
rs898367726 Chr.16: 19860615-19860615 on Build GRCh38
rs761798492 Chr.16: 19860555-19860555 on Build GRCh38
rs557850505 Chr.16: 19860818-19860818 on Build GRCh38
rs747249237 Chr.16: 19860590-19860590 on Build GRCh38
rs770742359 Chr.16: 19860685-19860685 on Build GRCh38
rs772877895 Chr.16: 19860510-19860510 on Build GRCh38
rs564301504 Chr.16: 19860810-19860810 on Build GRCh38
rs541130292 Chr.16: 19860520-19860520 on Build GRCh38
rs144982445 Chr.16: 19860807-19860807 on Build GRCh38
rs1052126987 Chr.16: 19860871-19860871 on Build GRCh38
rs956535192 Chr.16: 19860657-19860657 on Build GRCh38
rs1030814958 Chr.16: 19860614-19860614 on Build GRCh38
rs777261121 Chr.16: 19860483-19860483 on Build GRCh38
rs563864924 Chr.16: 19860766-19860766 on Build GRCh38
rs916125924 Chr.16: 19860825-19860825 on Build GRCh38
rs781257504 Chr.16: 19860755-19860755 on Build GRCh38
rs577424623 Chr.16: 19860484-19860484 on Build GRCh38
rs766068160 Chr.16: 19860530-19860530 on Build GRCh38
rs768077422 Chr.16: 19860863-19860863 on Build GRCh38
rs966144423 Chr.16: 19860754-19860754 on Build GRCh38
rs11865578 Chr.16: 19860660-19860660 on Build GRCh38
rs780314845 Chr.16: 19860583-19860583 on Build GRCh38
rs752600969 Chr.16: 19860550-19860550 on Build GRCh38
rs1003234876 Chr.16: 19860610-19860610 on Build GRCh38
rs183656593 Chr.16: 19860749-19860749 on Build GRCh38
rs531110754 Chr.16: 19860784-19860784 on Build GRCh38
rs769924647 Chr.16: 19860581-19860582 on Build GRCh38
rs191744313 Chr.16: 19860582-19860582 on Build GRCh38
rs1002574107 Chr.16: 19860781-19860781 on Build GRCh38

View Full Product Details