Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67398732-67399222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC1
Entrez Gene ID
29800
Gene Name
zinc finger DHHC-type containing 1
Gene Aliases
C16orf1, HSU90653, ZNF377
UniGene
Hs.624541
Gene Chromosome Location
Chr.16: 67394419-67416494 on Build GRCh38
Transcripts
NM_001323627.1 NM_013304.2 XM_005255915.1 XM_006721185.1 XM_011523058.1 XM_011523059.1 XM_011523060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2994831 c.653A>T p.E218V Chr.16: 67398822-67398822 on Build GRCh38
1189299 c.609G>T p.E203D Chr.16: 67398866-67398866 on Build GRCh38
703860 c.596A>T p.Y199F Chr.16: 67398879-67398879 on Build GRCh38
2994833 c.603C>T p.F201F Chr.16: 67398872-67398872 on Build GRCh38
6189851 c.652G>C p.E218Q Chr.16: 67398823-67398823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781125546 Chr.16: 67398943-67398943 on Build GRCh38
rs529064124 Chr.16: 67399052-67399052 on Build GRCh38
rs931458772 Chr.16: 67398999-67398999 on Build GRCh38
rs775245008 Chr.16: 67398804-67398804 on Build GRCh38
rs943929966 Chr.16: 67399048-67399048 on Build GRCh38
rs910930618 Chr.16: 67399025-67399025 on Build GRCh38
rs767848800 Chr.16: 67398898-67398898 on Build GRCh38
rs367620530 Chr.16: 67398772-67398772 on Build GRCh38
rs779123839 Chr.16: 67398850-67398850 on Build GRCh38
rs773957524 Chr.16: 67398881-67398881 on Build GRCh38
rs368581927 Chr.16: 67398978-67398978 on Build GRCh38
rs1014492378 Chr.16: 67398838-67398838 on Build GRCh38
rs549801855 Chr.16: 67399040-67399040 on Build GRCh38
rs754156041 Chr.16: 67398844-67398844 on Build GRCh38
rs755142654 Chr.16: 67398847-67398847 on Build GRCh38
rs773750953 Chr.16: 67398899-67398899 on Build GRCh38
rs150003209 Chr.16: 67398768-67398768 on Build GRCh38
rs545412296 Chr.16: 67398879-67398879 on Build GRCh38
rs756237481 Chr.16: 67398840-67398840 on Build GRCh38
rs543887530 Chr.16: 67398762-67398762 on Build GRCh38
rs967973182 Chr.16: 67399176-67399176 on Build GRCh38
rs774844883 Chr.16: 67398872-67398872 on Build GRCh38
rs372405273 Chr.16: 67398908-67398908 on Build GRCh38
rs750754097 Chr.16: 67398758-67398758 on Build GRCh38
rs780347496 Chr.16: 67398773-67398773 on Build GRCh38
rs757155278 Chr.16: 67398930-67398930 on Build GRCh38
rs565910338 Chr.16: 67399068-67399068 on Build GRCh38
rs867987964 Chr.16: 67398792-67398792 on Build GRCh38
rs967973447 Chr.16: 67398884-67398884 on Build GRCh38
rs995869046 Chr.16: 67399090-67399090 on Build GRCh38
rs1047231071 Chr.16: 67399012-67399012 on Build GRCh38
rs749451369 Chr.16: 67398780-67398780 on Build GRCh38
rs1001611528 Chr.16: 67399132-67399132 on Build GRCh38
rs766339839 Chr.16: 67398842-67398842 on Build GRCh38
rs888528990 Chr.16: 67399097-67399097 on Build GRCh38
rs765400019 Chr.16: 67398919-67398919 on Build GRCh38
rs374778259 Chr.16: 67398767-67398767 on Build GRCh38
rs752171232 Chr.16: 67398763-67398763 on Build GRCh38
rs372354325 Chr.16: 67398793-67398793 on Build GRCh38
rs370903934 Chr.16: 67398987-67398987 on Build GRCh38
rs756015946 Chr.16: 67398963-67398963 on Build GRCh38
rs1018622200 Chr.16: 67398785-67398785 on Build GRCh38
rs547634556 Chr.16: 67399053-67399053 on Build GRCh38
rs750884254 Chr.16: 67398831-67398831 on Build GRCh38
rs533426749 Chr.16: 67398863-67398863 on Build GRCh38
rs761082000 Chr.16: 67398906-67398906 on Build GRCh38
rs745856568 Chr.16: 67398944-67398944 on Build GRCh38
rs748782236 Chr.16: 67398967-67398967 on Build GRCh38
rs762504455 Chr.16: 67398805-67398805 on Build GRCh38
rs763420916 Chr.16: 67398761-67398761 on Build GRCh38
rs745766676 Chr.16: 67398866-67398866 on Build GRCh38
rs771942617 Chr.16: 67398788-67398788 on Build GRCh38
rs541243929 Chr.16: 67398801-67398801 on Build GRCh38
rs746830838 Chr.16: 67398857-67398857 on Build GRCh38
rs766538951 Chr.16: 67398907-67398907 on Build GRCh38
rs886155481 Chr.16: 67399031-67399031 on Build GRCh38
rs769411072 Chr.16: 67398871-67398871 on Build GRCh38
rs551424949 Chr.16: 67399091-67399091 on Build GRCh38
rs757745354 Chr.16: 67398764-67398764 on Build GRCh38
rs376612420 Chr.16: 67398816-67398816 on Build GRCh38
rs573857179 Chr.16: 67398790-67398790 on Build GRCh38
rs539726044 Chr.16: 67399079-67399079 on Build GRCh38
rs762191085 Chr.16: 67398883-67398883 on Build GRCh38
rs901493548 Chr.16: 67399078-67399078 on Build GRCh38
rs780196047 Chr.16: 67398841-67398841 on Build GRCh38
rs561774582 Chr.16: 67399049-67399049 on Build GRCh38
rs372751146 Chr.16: 67398920-67398920 on Build GRCh38
rs570027663 Chr.16: 67399189-67399189 on Build GRCh38
rs752241973 Chr.16: 67398757-67398757 on Build GRCh38
rs369730444 Chr.16: 67398874-67398874 on Build GRCh38
rs375952924 Chr.16: 67398977-67398977 on Build GRCh38
rs370323251 Chr.16: 67398964-67398964 on Build GRCh38
rs755043579 Chr.16: 67398911-67398911 on Build GRCh38
rs992533536 Chr.16: 67398933-67398933 on Build GRCh38
rs769162137 Chr.16: 67398802-67398802 on Build GRCh38
rs776668404 Chr.16: 67398798-67398798 on Build GRCh38
rs369229169 Chr.16: 67399109-67399109 on Build GRCh38
rs1004630655 Chr.16: 67399046-67399046 on Build GRCh38
rs760053380 Chr.16: 67399073-67399073 on Build GRCh38
rs768201787 Chr.16: 67398877-67398877 on Build GRCh38

View Full Product Details