Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67935490-67935982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB10
Entrez Gene ID
5699
Gene Name
proteasome subunit beta 10
Gene Aliases
LMP10, MECL1, beta2i
UniGene
Hs.9661
Gene Chromosome Location
Chr.16: 67934504-67936877 on Build GRCh38
Transcripts
NM_002801.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777999 c.483G>A p.L161L Chr.16: 67935598-67935598 on Build GRCh38
4883703 c.404C>A p.A135E Chr.16: 67935677-67935677 on Build GRCh38
6035664 c.484C>T p.P162S Chr.16: 67935597-67935597 on Build GRCh38
7177742 c.432G>C p.L144L Chr.16: 67935649-67935649 on Build GRCh38
3818305 c.489C>T p.F163F Chr.16: 67935592-67935592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750867899 Chr.16: 67935514-67935514 on Build GRCh38
rs754244406 Chr.16: 67935935-67935935 on Build GRCh38
rs878911762 Chr.16: 67935738-67935738 on Build GRCh38
rs749251923 Chr.16: 67935735-67935735 on Build GRCh38
rs759161390 Chr.16: 67935678-67935678 on Build GRCh38
rs374539582 Chr.16: 67935516-67935516 on Build GRCh38
rs956708569 Chr.16: 67935810-67935810 on Build GRCh38
rs138394638 Chr.16: 67935624-67935624 on Build GRCh38
rs996370754 Chr.16: 67935712-67935712 on Build GRCh38
rs1040147897 Chr.16: 67935684-67935684 on Build GRCh38
rs1050654704 Chr.16: 67935963-67935963 on Build GRCh38
rs779349651 Chr.16: 67935700-67935700 on Build GRCh38
rs1025664791 Chr.16: 67935709-67935709 on Build GRCh38
rs750969201 Chr.16: 67935617-67935617 on Build GRCh38
rs760140635 Chr.16: 67935931-67935931 on Build GRCh38
rs112794989 Chr.16: 67935868-67935868 on Build GRCh38
rs1012029485 Chr.16: 67935752-67935752 on Build GRCh38
rs370963927 Chr.16: 67935956-67935956 on Build GRCh38
rs1079723 Chr.16: 67935781-67935781 on Build GRCh38
rs756668971 Chr.16: 67935529-67935529 on Build GRCh38
rs1016492821 Chr.16: 67935835-67935835 on Build GRCh38
rs201566321 Chr.16: 67935916-67935916 on Build GRCh38
rs760147515 Chr.16: 67935938-67935938 on Build GRCh38
rs747133219 Chr.16: 67935722-67935722 on Build GRCh38
rs763375288 Chr.16: 67935640-67935645 on Build GRCh38
rs200453196 Chr.16: 67935958-67935958 on Build GRCh38
rs752849317 Chr.16: 67935704-67935704 on Build GRCh38
rs923044203 Chr.16: 67935927-67935927 on Build GRCh38
rs771749532 Chr.16: 67935925-67935925 on Build GRCh38
rs768905112 Chr.16: 67935544-67935545 on Build GRCh38
rs775331213 Chr.16: 67935616-67935616 on Build GRCh38
rs767120162 Chr.16: 67935708-67935709 on Build GRCh38
rs907619781 Chr.16: 67935667-67935667 on Build GRCh38
rs755197510 Chr.16: 67935877-67935877 on Build GRCh38
rs1031034731 Chr.16: 67935778-67935778 on Build GRCh38
rs766717951 Chr.16: 67935641-67935650 on Build GRCh38
rs199551744 Chr.16: 67935739-67935739 on Build GRCh38
rs772876833 Chr.16: 67935930-67935930 on Build GRCh38
rs144563727 Chr.16: 67935602-67935602 on Build GRCh38
rs780389173 Chr.16: 67935534-67935534 on Build GRCh38
rs771156115 Chr.16: 67935731-67935731 on Build GRCh38
rs766825816 Chr.16: 67935661-67935661 on Build GRCh38
rs779815715 Chr.16: 67935734-67935734 on Build GRCh38
rs979984570 Chr.16: 67935866-67935866 on Build GRCh38
rs755234526 Chr.16: 67935688-67935688 on Build GRCh38
rs374418759 Chr.16: 67935829-67935829 on Build GRCh38
rs774256077 Chr.16: 67935741-67935741 on Build GRCh38
rs754403391 Chr.16: 67935686-67935686 on Build GRCh38
rs940385278 Chr.16: 67935681-67935681 on Build GRCh38
rs770354708 Chr.16: 67935630-67935631 on Build GRCh38
rs907496816 Chr.16: 67935889-67935889 on Build GRCh38
rs753155097 Chr.16: 67935954-67935954 on Build GRCh38
rs777849619 Chr.16: 67935914-67935914 on Build GRCh38
rs949284955 Chr.16: 67935879-67935879 on Build GRCh38
rs766427512 Chr.16: 67935847-67935847 on Build GRCh38
rs772221413 Chr.16: 67935585-67935585 on Build GRCh38
rs143808892 Chr.16: 67935657-67935657 on Build GRCh38
rs570917522 Chr.16: 67935962-67935962 on Build GRCh38
rs761464535 Chr.16: 67935631-67935631 on Build GRCh38
rs765922650 Chr.16: 67935933-67935933 on Build GRCh38
rs748411006 Chr.16: 67935584-67935584 on Build GRCh38
rs529994379 Chr.16: 67935660-67935660 on Build GRCh38
rs999301507 Chr.16: 67935539-67935539 on Build GRCh38
rs755394979 Chr.16: 67935571-67935571 on Build GRCh38
rs748323637 Chr.16: 67935907-67935907 on Build GRCh38
rs879375019 Chr.16: 67935747-67935747 on Build GRCh38
rs748136367 Chr.16: 67935618-67935618 on Build GRCh38
rs767015932 Chr.16: 67935512-67935512 on Build GRCh38
rs990729332 Chr.16: 67935872-67935872 on Build GRCh38
rs778967736 Chr.16: 67935577-67935577 on Build GRCh38
rs1001271194 Chr.16: 67935771-67935771 on Build GRCh38
rs553495244 Chr.16: 67935557-67935559 on Build GRCh38
rs758761627 Chr.16: 67935719-67935719 on Build GRCh38
rs971192320 Chr.16: 67935857-67935857 on Build GRCh38
rs940354211 Chr.16: 67935901-67935901 on Build GRCh38
rs765515451 Chr.16: 67935940-67935940 on Build GRCh38
rs752279184 Chr.16: 67935709-67935710 on Build GRCh38
rs769908499 Chr.16: 67935900-67935900 on Build GRCh38
rs902221005 Chr.16: 67935536-67935536 on Build GRCh38
rs774040854 Chr.16: 67935647-67935653 on Build GRCh38
rs201188208 Chr.16: 67935611-67935611 on Build GRCh38
rs755525249 Chr.16: 67935921-67935922 on Build GRCh38
rs749666370 Chr.16: 67935544-67935544 on Build GRCh38
rs11075663 Chr.16: 67935887-67935887 on Build GRCh38
rs14178 Chr.16: 67935628-67935628 on Build GRCh38
rs745797567 Chr.16: 67935603-67935603 on Build GRCh38
rs773525455 Chr.16: 67935640-67935648 on Build GRCh38

View Full Product Details