Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67980332-67980829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPEP3
Entrez Gene ID
64180
Gene Name
dipeptidase 3
Gene Aliases
MBD3
UniGene
Hs.302028
Gene Chromosome Location
Chr.16: 67975663-67980549 on Build GRCh38
Transcripts
NM_001129758.1 NM_022357.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
394435 c.107C>T p.A36V Chr.16: 67980349-67980349 on Build GRCh38
4062062 c.105C>T p.R35R Chr.16: 67980351-67980351 on Build GRCh38
4623384 c.93C>T p.R31R Chr.16: 67980363-67980363 on Build GRCh38
6035665 c.61C>T p.R21* Chr.16: 67980395-67980395 on Build GRCh38
3387552 c.69G>A p.P23P Chr.16: 67980387-67980387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039992060 Chr.16: 67980465-67980465 on Build GRCh38
rs903200622 Chr.16: 67980693-67980693 on Build GRCh38
rs947656979 Chr.16: 67980679-67980679 on Build GRCh38
rs748814678 Chr.16: 67980437-67980437 on Build GRCh38
rs962987030 Chr.16: 67980606-67980606 on Build GRCh38
rs951059242 Chr.16: 67980617-67980617 on Build GRCh38
rs372588067 Chr.16: 67980390-67980390 on Build GRCh38
rs771185451 Chr.16: 67980505-67980505 on Build GRCh38
rs918254284 Chr.16: 67980434-67980434 on Build GRCh38
rs1035715690 Chr.16: 67980552-67980552 on Build GRCh38
rs896338980 Chr.16: 67980584-67980584 on Build GRCh38
rs200300395 Chr.16: 67980389-67980389 on Build GRCh38
rs757063157 Chr.16: 67980413-67980413 on Build GRCh38
rs960510134 Chr.16: 67980386-67980386 on Build GRCh38
rs1034845994 Chr.16: 67980351-67980351 on Build GRCh38
rs576264712 Chr.16: 67980645-67980645 on Build GRCh38
rs949895562 Chr.16: 67980646-67980646 on Build GRCh38
rs953263312 Chr.16: 67980524-67980524 on Build GRCh38
rs1023556507 Chr.16: 67980601-67980601 on Build GRCh38
rs772381907 Chr.16: 67980479-67980479 on Build GRCh38
rs1057155117 Chr.16: 67980717-67980717 on Build GRCh38
rs888743567 Chr.16: 67980751-67980751 on Build GRCh38
rs755879389 Chr.16: 67980435-67980435 on Build GRCh38
rs377542686 Chr.16: 67980472-67980472 on Build GRCh38
rs1012952911 Chr.16: 67980771-67980771 on Build GRCh38
rs371062584 Chr.16: 67980775-67980775 on Build GRCh38
rs986566543 Chr.16: 67980628-67980628 on Build GRCh38
rs1012002253 Chr.16: 67980587-67980587 on Build GRCh38
rs930890719 Chr.16: 67980611-67980611 on Build GRCh38
rs930671562 Chr.16: 67980425-67980425 on Build GRCh38
rs1025534010 Chr.16: 67980800-67980800 on Build GRCh38
rs903171804 Chr.16: 67980540-67980540 on Build GRCh38
rs942821291 Chr.16: 67980461-67980461 on Build GRCh38
rs561987067 Chr.16: 67980614-67980614 on Build GRCh38
rs545613168 Chr.16: 67980520-67980520 on Build GRCh38
rs1002937116 Chr.16: 67980541-67980541 on Build GRCh38
rs142218554 Chr.16: 67980491-67980491 on Build GRCh38
rs894526478 Chr.16: 67980767-67980767 on Build GRCh38
rs985214229 Chr.16: 67980612-67980612 on Build GRCh38
rs967310259 Chr.16: 67980515-67980515 on Build GRCh38
rs764007361 Chr.16: 67980395-67980395 on Build GRCh38
rs375626255 Chr.16: 67980610-67980610 on Build GRCh38
rs975539053 Chr.16: 67980670-67980670 on Build GRCh38
rs1007178804 Chr.16: 67980754-67980754 on Build GRCh38
rs767442609 Chr.16: 67980661-67980662 on Build GRCh38
rs767921700 Chr.16: 67980631-67980631 on Build GRCh38
rs1018683691 Chr.16: 67980770-67980770 on Build GRCh38
rs529274298 Chr.16: 67980761-67980761 on Build GRCh38
rs1056520819 Chr.16: 67980642-67980642 on Build GRCh38
rs767425264 Chr.16: 67980423-67980423 on Build GRCh38
rs974109824 Chr.16: 67980403-67980403 on Build GRCh38
rs751357521 Chr.16: 67980407-67980407 on Build GRCh38
rs898314074 Chr.16: 67980777-67980777 on Build GRCh38
rs938658342 Chr.16: 67980699-67980699 on Build GRCh38
rs1025758612 Chr.16: 67980615-67980615 on Build GRCh38
rs972198221 Chr.16: 67980570-67980570 on Build GRCh38
rs912064652 Chr.16: 67980532-67980532 on Build GRCh38
rs755495304 Chr.16: 67980444-67980444 on Build GRCh38
rs554743775 Chr.16: 67980478-67980478 on Build GRCh38
rs192494106 Chr.16: 67980678-67980678 on Build GRCh38
rs750057887 Chr.16: 67980424-67980424 on Build GRCh38
rs561970622 Chr.16: 67980680-67980680 on Build GRCh38
rs1050817378 Chr.16: 67980442-67980442 on Build GRCh38
rs1000294362 Chr.16: 67980516-67980517 on Build GRCh38
rs912756661 Chr.16: 67980456-67980456 on Build GRCh38
rs778129361 Chr.16: 67980480-67980480 on Build GRCh38
rs995850782 Chr.16: 67980797-67980797 on Build GRCh38
rs895615185 Chr.16: 67980492-67980492 on Build GRCh38
rs966221388 Chr.16: 67980547-67980547 on Build GRCh38
rs766402220 Chr.16: 67980365-67980365 on Build GRCh38
rs1041955989 Chr.16: 67980685-67980685 on Build GRCh38
rs779227802 Chr.16: 67980468-67980468 on Build GRCh38
rs962414774 Chr.16: 67980400-67980400 on Build GRCh38
rs1014180602 Chr.16: 67980502-67980502 on Build GRCh38
rs759155633 Chr.16: 67980543-67980543 on Build GRCh38
rs985810333 Chr.16: 67980526-67980526 on Build GRCh38

View Full Product Details