Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69116643-69117150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS3
Entrez Gene ID
3038
Gene Name
hyaluronan synthase 3
Gene Aliases
-
UniGene
Hs.592069
Gene Chromosome Location
Chr.16: 69105564-69118719 on Build GRCh38
Transcripts
NM_001199280.1 NM_005329.2 NM_138612.2 XM_005255921.2 XM_011523061.2

SNP Detail

SNP ID Chromosome Location
rs966142409 Chr.16: 69116870-69116870 on Build GRCh38
rs530124757 Chr.16: 69116794-69116794 on Build GRCh38
rs934295361 Chr.16: 69116995-69116995 on Build GRCh38
rs947818970 Chr.16: 69116757-69116757 on Build GRCh38
rs16958646 Chr.16: 69116987-69116987 on Build GRCh38
rs1045175727 Chr.16: 69117013-69117013 on Build GRCh38
rs757449114 Chr.16: 69116862-69116862 on Build GRCh38
rs1012599614 Chr.16: 69116879-69116879 on Build GRCh38
rs1024617365 Chr.16: 69116892-69116892 on Build GRCh38
rs935607712 Chr.16: 69116964-69116964 on Build GRCh38
rs948259499 Chr.16: 69117012-69117012 on Build GRCh38
rs887060451 Chr.16: 69116850-69116850 on Build GRCh38
rs998720534 Chr.16: 69116815-69116815 on Build GRCh38
rs979432861 Chr.16: 69116927-69116927 on Build GRCh38
rs924179709 Chr.16: 69116937-69116937 on Build GRCh38
rs116124683 Chr.16: 69117071-69117071 on Build GRCh38
rs1031539005 Chr.16: 69116918-69116918 on Build GRCh38
rs887011233 Chr.16: 69117124-69117124 on Build GRCh38
rs1047234089 Chr.16: 69116848-69116848 on Build GRCh38
rs748928636 Chr.16: 69117028-69117028 on Build GRCh38
rs953106386 Chr.16: 69117049-69117058 on Build GRCh38
rs8057513 Chr.16: 69116714-69116714 on Build GRCh38
rs377429593 Chr.16: 69116856-69116856 on Build GRCh38
rs371186811 Chr.16: 69116942-69116942 on Build GRCh38
rs915230085 Chr.16: 69116966-69116966 on Build GRCh38
rs112703877 Chr.16: 69116996-69116996 on Build GRCh38
rs958387252 Chr.16: 69116868-69116868 on Build GRCh38
rs551012206 Chr.16: 69116779-69116779 on Build GRCh38
rs1056680305 Chr.16: 69116668-69116668 on Build GRCh38
rs559076475 Chr.16: 69116851-69116851 on Build GRCh38
rs1029094554 Chr.16: 69116912-69116912 on Build GRCh38
rs942675838 Chr.16: 69116967-69116967 on Build GRCh38
rs1025482419 Chr.16: 69116802-69116802 on Build GRCh38
rs975205101 Chr.16: 69116897-69116897 on Build GRCh38
rs566964921 Chr.16: 69116863-69116863 on Build GRCh38
rs1012955637 Chr.16: 69117093-69117093 on Build GRCh38
rs571377597 Chr.16: 69117057-69117057 on Build GRCh38
rs941073848 Chr.16: 69117048-69117048 on Build GRCh38
rs569226729 Chr.16: 69116788-69116788 on Build GRCh38
rs745843768 Chr.16: 69116973-69116973 on Build GRCh38
rs989548558 Chr.16: 69116965-69116965 on Build GRCh38
rs1047671797 Chr.16: 69117011-69117011 on Build GRCh38
rs919695007 Chr.16: 69117040-69117040 on Build GRCh38
rs975457475 Chr.16: 69116978-69116978 on Build GRCh38
rs1016589935 Chr.16: 69116866-69116866 on Build GRCh38
rs879033757 Chr.16: 69116792-69116792 on Build GRCh38
rs915282377 Chr.16: 69116688-69116688 on Build GRCh38
rs775363928 Chr.16: 69117123-69117123 on Build GRCh38
rs747208722 Chr.16: 69116765-69116765 on Build GRCh38
rs1053709084 Chr.16: 69116677-69116677 on Build GRCh38
rs556848046 Chr.16: 69117081-69117081 on Build GRCh38
rs139315121 Chr.16: 69116832-69116832 on Build GRCh38
rs898132231 Chr.16: 69116766-69116766 on Build GRCh38
rs968412717 Chr.16: 69116913-69116913 on Build GRCh38
rs1023983508 Chr.16: 69117098-69117098 on Build GRCh38

View Full Product Details