Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70646688-70647224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_001172772.1 NM_152456.2 XM_011522902.2 XM_011522903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
973055 c.11G>A p.G4D Chr.16: 70646958-70646958 on Build GRCh38
3691133 c.25C>T p.R9C Chr.16: 70646972-70646972 on Build GRCh38
6238470 c.5C>G p.P2R Chr.16: 70646952-70646952 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370387616 Chr.16: 70646877-70646877 on Build GRCh38
rs930625549 Chr.16: 70647085-70647085 on Build GRCh38
rs912956076 Chr.16: 70646781-70646781 on Build GRCh38
rs772446015 Chr.16: 70646954-70646954 on Build GRCh38
rs574168160 Chr.16: 70646999-70646999 on Build GRCh38
rs777145598 Chr.16: 70646899-70646899 on Build GRCh38
rs755038942 Chr.16: 70646939-70646939 on Build GRCh38
rs761784434 Chr.16: 70646934-70646934 on Build GRCh38
rs566076969 Chr.16: 70647147-70647147 on Build GRCh38
rs374377265 Chr.16: 70647039-70647039 on Build GRCh38
rs369011177 Chr.16: 70646972-70646972 on Build GRCh38
rs572450078 Chr.16: 70647055-70647055 on Build GRCh38
rs1004813329 Chr.16: 70646829-70646829 on Build GRCh38
rs185531379 Chr.16: 70647072-70647072 on Build GRCh38
rs374367865 Chr.16: 70646914-70646914 on Build GRCh38
rs940695997 Chr.16: 70646932-70646932 on Build GRCh38
rs576906053 Chr.16: 70646740-70646740 on Build GRCh38
rs898714010 Chr.16: 70646998-70646998 on Build GRCh38
rs376027935 Chr.16: 70646776-70646793 on Build GRCh38
rs373269031 Chr.16: 70647148-70647148 on Build GRCh38
rs566779236 Chr.16: 70646942-70646942 on Build GRCh38
rs1003645517 Chr.16: 70646795-70646795 on Build GRCh38
rs988451295 Chr.16: 70646895-70646895 on Build GRCh38
rs541392162 Chr.16: 70646833-70646833 on Build GRCh38
rs1018418361 Chr.16: 70647188-70647188 on Build GRCh38
rs548585613 Chr.16: 70647108-70647108 on Build GRCh38
rs771122772 Chr.16: 70646943-70646943 on Build GRCh38
rs558742486 Chr.16: 70646738-70646738 on Build GRCh38
rs74024417 Chr.16: 70646935-70646935 on Build GRCh38
rs973516661 Chr.16: 70646722-70646722 on Build GRCh38
rs540428536 Chr.16: 70646736-70646736 on Build GRCh38
rs74024416 Chr.16: 70646721-70646721 on Build GRCh38
rs994969753 Chr.16: 70646937-70646937 on Build GRCh38
rs766176480 Chr.16: 70647163-70647163 on Build GRCh38
rs775931818 Chr.16: 70646996-70646996 on Build GRCh38
rs559678912 Chr.16: 70647030-70647030 on Build GRCh38
rs771576382 Chr.16: 70647177-70647177 on Build GRCh38
rs767421408 Chr.16: 70647002-70647002 on Build GRCh38
rs1012276474 Chr.16: 70646982-70646982 on Build GRCh38
rs531761806 Chr.16: 70646876-70646876 on Build GRCh38
rs916643845 Chr.16: 70647117-70647117 on Build GRCh38
rs544292004 Chr.16: 70646746-70646746 on Build GRCh38
rs768246550 Chr.16: 70646992-70646992 on Build GRCh38
rs781329723 Chr.16: 70646728-70646728 on Build GRCh38
rs1057413669 Chr.16: 70646815-70646815 on Build GRCh38
rs3813904 Chr.16: 70646841-70646841 on Build GRCh38
rs1025823694 Chr.16: 70646855-70646855 on Build GRCh38
rs531920606 Chr.16: 70647101-70647101 on Build GRCh38
rs868173399 Chr.16: 70646902-70646902 on Build GRCh38
rs560436922 Chr.16: 70646835-70646835 on Build GRCh38
rs762638551 Chr.16: 70646974-70646974 on Build GRCh38
rs868038962 Chr.16: 70646901-70646901 on Build GRCh38
rs535743576 Chr.16: 70647153-70647153 on Build GRCh38
rs751355881 Chr.16: 70646928-70646928 on Build GRCh38
rs1036233701 Chr.16: 70647082-70647082 on Build GRCh38
rs555767889 Chr.16: 70647166-70647166 on Build GRCh38
rs746390037 Chr.16: 70646967-70646967 on Build GRCh38
rs895846026 Chr.16: 70646819-70646819 on Build GRCh38
rs368387403 Chr.16: 70646922-70646922 on Build GRCh38
rs552363683 Chr.16: 70646851-70646851 on Build GRCh38
rs953619417 Chr.16: 70646731-70646731 on Build GRCh38
rs142146633 Chr.16: 70646946-70646946 on Build GRCh38
rs529362966 Chr.16: 70647128-70647128 on Build GRCh38
rs902573992 Chr.16: 70646794-70646794 on Build GRCh38
rs962891942 Chr.16: 70646839-70646839 on Build GRCh38
rs376123115 Chr.16: 70646969-70646969 on Build GRCh38
rs551273656 Chr.16: 70646771-70646788 on Build GRCh38
rs771359932 Chr.16: 70646955-70646955 on Build GRCh38
rs955605034 Chr.16: 70647015-70647015 on Build GRCh38
rs1008371974 Chr.16: 70647178-70647178 on Build GRCh38
rs780278766 Chr.16: 70646927-70646928 on Build GRCh38
rs775050345 Chr.16: 70646973-70646973 on Build GRCh38
rs892637533 Chr.16: 70647124-70647124 on Build GRCh38
rs529675813 Chr.16: 70646800-70646800 on Build GRCh38
rs956908974 Chr.16: 70646950-70646950 on Build GRCh38
rs751743716 Chr.16: 70646984-70646985 on Build GRCh38
rs371965836 Chr.16: 70646912-70646912 on Build GRCh38
rs750754050 Chr.16: 70647040-70647040 on Build GRCh38
rs562371317 Chr.16: 70646797-70646797 on Build GRCh38
rs778169047 Chr.16: 70646952-70646952 on Build GRCh38
rs889176504 Chr.16: 70647096-70647096 on Build GRCh38
rs1001499800 Chr.16: 70646813-70646813 on Build GRCh38
rs371492920 Chr.16: 70647007-70647007 on Build GRCh38
rs1050545794 Chr.16: 70647095-70647095 on Build GRCh38
rs1052884158 Chr.16: 70646766-70646766 on Build GRCh38
rs968387477 Chr.16: 70647143-70647143 on Build GRCh38
rs368494467 Chr.16: 70646911-70646911 on Build GRCh38
rs541428888 Chr.16: 70647004-70647004 on Build GRCh38
rs947206519 Chr.16: 70646788-70646788 on Build GRCh38
rs1056445927 Chr.16: 70646805-70646805 on Build GRCh38
rs994971517 Chr.16: 70646847-70646847 on Build GRCh38
rs554160387 Chr.16: 70646938-70646938 on Build GRCh38
rs923767952 Chr.16: 70647092-70647092 on Build GRCh38
rs899193508 Chr.16: 70647084-70647084 on Build GRCh38
rs1029195656 Chr.16: 70646862-70646862 on Build GRCh38
rs751379175 Chr.16: 70647192-70647192 on Build GRCh38
rs761094359 Chr.16: 70647045-70647045 on Build GRCh38
rs540011631 Chr.16: 70647027-70647027 on Build GRCh38
rs1008592266 Chr.16: 70647016-70647016 on Build GRCh38
rs573481132 Chr.16: 70647132-70647132 on Build GRCh38
rs369833600 Chr.16: 70647024-70647024 on Build GRCh38
rs573144744 Chr.16: 70646729-70646729 on Build GRCh38
rs1042862762 Chr.16: 70646796-70646796 on Build GRCh38
rs562212914 Chr.16: 70647120-70647120 on Build GRCh38
rs963243065 Chr.16: 70646708-70646708 on Build GRCh38
rs905685888 Chr.16: 70646807-70646807 on Build GRCh38
rs370984741 Chr.16: 70646878-70646878 on Build GRCh38
rs548190195 Chr.16: 70646719-70646719 on Build GRCh38
rs775940328 Chr.16: 70647190-70647190 on Build GRCh38
rs3813905 Chr.16: 70646947-70646947 on Build GRCh38
rs748058543 Chr.16: 70646735-70646735 on Build GRCh38
rs763949176 Chr.16: 70646926-70646926 on Build GRCh38
rs1021976538 Chr.16: 70647058-70647058 on Build GRCh38
rs1022543610 Chr.16: 70647129-70647129 on Build GRCh38
rs556241304 Chr.16: 70646994-70646994 on Build GRCh38
rs547850052 Chr.16: 70647131-70647131 on Build GRCh38
rs374541911 Chr.16: 70646886-70646886 on Build GRCh38
rs200158701 Chr.16: 70646951-70646951 on Build GRCh38
rs958664145 Chr.16: 70647167-70647167 on Build GRCh38
rs571018045 Chr.16: 70646874-70646874 on Build GRCh38
rs188939094 Chr.16: 70647081-70647081 on Build GRCh38
rs192337001 Chr.16: 70646963-70646963 on Build GRCh38

View Full Product Details