Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7382744-7383239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNK1
Entrez Gene ID
8711
Gene Name
tyrosine kinase non receptor 1
Gene Aliases
-
UniGene
Hs.203420
Gene Chromosome Location
Chr.17: 7380559-7389774 on Build GRCh38
Transcripts
NM_001251902.1 NM_003985.4 XM_011524045.2 XM_011524046.2 XM_011524047.2 XM_011524048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4000294 c.81T>A p.L27L Chr.17: 7383007-7383007 on Build GRCh38
6989781 c.114C>T p.F38F Chr.17: 7383040-7383040 on Build GRCh38
4392128 c.85G>A p.E29K Chr.17: 7383011-7383011 on Build GRCh38
4623659 c.9T>C p.P3P Chr.17: 7382935-7382935 on Build GRCh38
984150 c.43C>T p.R15W Chr.17: 7382969-7382969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763242061 Chr.17: 7382957-7382958 on Build GRCh38
rs764324329 Chr.17: 7383021-7383021 on Build GRCh38
rs780834064 Chr.17: 7383068-7383068 on Build GRCh38
rs114938550 Chr.17: 7383107-7383107 on Build GRCh38
rs978081251 Chr.17: 7382773-7382773 on Build GRCh38
rs374753488 Chr.17: 7383027-7383027 on Build GRCh38
rs577697153 Chr.17: 7382969-7382969 on Build GRCh38
rs750573571 Chr.17: 7382957-7382959 on Build GRCh38
rs754186504 Chr.17: 7382957-7382957 on Build GRCh38
rs965986044 Chr.17: 7382814-7382814 on Build GRCh38
rs754509649 Chr.17: 7383178-7383178 on Build GRCh38
rs757897037 Chr.17: 7382967-7382967 on Build GRCh38
rs991349735 Chr.17: 7382818-7382818 on Build GRCh38
rs537553485 Chr.17: 7383140-7383140 on Build GRCh38
rs763332336 Chr.17: 7383126-7383126 on Build GRCh38
rs536825259 Chr.17: 7382823-7382823 on Build GRCh38
rs544127209 Chr.17: 7382921-7382921 on Build GRCh38
rs560740452 Chr.17: 7383108-7383108 on Build GRCh38
rs770949791 Chr.17: 7383103-7383103 on Build GRCh38
rs778841967 Chr.17: 7383210-7383210 on Build GRCh38
rs1554947 Chr.17: 7382947-7382947 on Build GRCh38
rs761929004 Chr.17: 7382909-7382909 on Build GRCh38
rs747920038 Chr.17: 7382971-7382971 on Build GRCh38
rs374692022 Chr.17: 7382966-7382966 on Build GRCh38
rs369202866 Chr.17: 7383113-7383113 on Build GRCh38
rs554875958 Chr.17: 7382838-7382838 on Build GRCh38
rs898866321 Chr.17: 7382995-7382995 on Build GRCh38
rs767680748 Chr.17: 7383002-7383002 on Build GRCh38
rs757581070 Chr.17: 7382958-7382958 on Build GRCh38
rs765379680 Chr.17: 7382955-7382956 on Build GRCh38
rs752375512 Chr.17: 7383187-7383187 on Build GRCh38
rs549640660 Chr.17: 7383175-7383175 on Build GRCh38
rs749038186 Chr.17: 7383202-7383202 on Build GRCh38
rs566055005 Chr.17: 7382801-7382801 on Build GRCh38
rs138569412 Chr.17: 7383182-7383182 on Build GRCh38
rs747826892 Chr.17: 7383075-7383075 on Build GRCh38
rs763044398 Chr.17: 7382984-7382984 on Build GRCh38
rs765173531 Chr.17: 7382907-7382907 on Build GRCh38
rs974746184 Chr.17: 7382864-7382864 on Build GRCh38
rs958300907 Chr.17: 7383101-7383101 on Build GRCh38
rs200735706 Chr.17: 7383041-7383041 on Build GRCh38
rs1018868152 Chr.17: 7382775-7382775 on Build GRCh38
rs373258961 Chr.17: 7383067-7383067 on Build GRCh38
rs1037364088 Chr.17: 7382972-7382972 on Build GRCh38
rs144115801 Chr.17: 7383197-7383197 on Build GRCh38
rs565717325 Chr.17: 7382866-7382866 on Build GRCh38
rs966953790 Chr.17: 7382771-7382771 on Build GRCh38
rs367599004 Chr.17: 7383217-7383217 on Build GRCh38
rs746479432 Chr.17: 7383062-7383062 on Build GRCh38
rs916397521 Chr.17: 7382847-7382847 on Build GRCh38
rs17732702 Chr.17: 7383040-7383040 on Build GRCh38
rs768315951 Chr.17: 7382891-7382891 on Build GRCh38
rs766030944 Chr.17: 7383008-7383010 on Build GRCh38
rs745859668 Chr.17: 7383213-7383213 on Build GRCh38
rs757575289 Chr.17: 7383026-7383026 on Build GRCh38
rs368513840 Chr.17: 7383038-7383038 on Build GRCh38
rs200263301 Chr.17: 7383035-7383035 on Build GRCh38
rs780572745 Chr.17: 7383052-7383052 on Build GRCh38
rs777763861 Chr.17: 7383090-7383090 on Build GRCh38
rs531448636 Chr.17: 7383136-7383136 on Build GRCh38
rs769485134 Chr.17: 7382981-7382981 on Build GRCh38
rs752957084 Chr.17: 7382927-7382927 on Build GRCh38
rs754019389 Chr.17: 7383024-7383024 on Build GRCh38
rs374118434 Chr.17: 7382819-7382819 on Build GRCh38
rs779333797 Chr.17: 7382959-7382959 on Build GRCh38
rs958139606 Chr.17: 7382802-7382802 on Build GRCh38
rs776531171 Chr.17: 7382903-7382903 on Build GRCh38
rs752538066 Chr.17: 7382882-7382882 on Build GRCh38
rs528505771 Chr.17: 7383096-7383096 on Build GRCh38
rs368551026 Chr.17: 7382905-7382905 on Build GRCh38
rs750747615 Chr.17: 7383125-7383125 on Build GRCh38
rs1554948 Chr.17: 7383007-7383007 on Build GRCh38
rs775749356 Chr.17: 7383006-7383006 on Build GRCh38
rs774173462 Chr.17: 7383104-7383104 on Build GRCh38
rs377419297 Chr.17: 7383196-7383196 on Build GRCh38
rs1554949 Chr.17: 7383073-7383073 on Build GRCh38
rs773299841 Chr.17: 7382913-7382913 on Build GRCh38
rs373034596 Chr.17: 7383121-7383121 on Build GRCh38
rs35371814 Chr.17: 7383093-7383094 on Build GRCh38
rs769701142 Chr.17: 7383083-7383083 on Build GRCh38
rs770859777 Chr.17: 7383205-7383205 on Build GRCh38
rs766813936 Chr.17: 7383133-7383133 on Build GRCh38
rs199784773 Chr.17: 7382970-7382970 on Build GRCh38
rs756404365 Chr.17: 7382939-7382939 on Build GRCh38

View Full Product Details