Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7496126-7496621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
UniGene
Hs.270017
Gene Chromosome Location
Chr.17: 7484379-7514618 on Build GRCh38
Transcripts
NM_000937.4

SNP Detail

SNP ID Chromosome Location
rs955334378 Chr.17: 7496395-7496395 on Build GRCh38
rs766405787 Chr.17: 7496318-7496318 on Build GRCh38
rs776216864 Chr.17: 7496256-7496256 on Build GRCh38
rs578057578 Chr.17: 7496362-7496362 on Build GRCh38
rs565768507 Chr.17: 7496167-7496167 on Build GRCh38
rs772860031 Chr.17: 7496530-7496530 on Build GRCh38
rs977539951 Chr.17: 7496553-7496553 on Build GRCh38
rs747520179 Chr.17: 7496508-7496508 on Build GRCh38
rs767633932 Chr.17: 7496577-7496577 on Build GRCh38
rs757637430 Chr.17: 7496538-7496538 on Build GRCh38
rs766098379 Chr.17: 7496428-7496428 on Build GRCh38
rs774861060 Chr.17: 7496215-7496215 on Build GRCh38
rs779412281 Chr.17: 7496396-7496396 on Build GRCh38
rs774900335 Chr.17: 7496201-7496201 on Build GRCh38
rs765431216 Chr.17: 7496596-7496596 on Build GRCh38
rs758743434 Chr.17: 7496478-7496478 on Build GRCh38
rs771370687 Chr.17: 7496191-7496191 on Build GRCh38
rs772696430 Chr.17: 7496211-7496211 on Build GRCh38
rs763527500 Chr.17: 7496247-7496247 on Build GRCh38
rs747722190 Chr.17: 7496512-7496512 on Build GRCh38
rs754262279 Chr.17: 7496535-7496535 on Build GRCh38
rs746313882 Chr.17: 7496397-7496397 on Build GRCh38
rs778018246 Chr.17: 7496374-7496374 on Build GRCh38
rs772986014 Chr.17: 7496418-7496418 on Build GRCh38
rs764984502 Chr.17: 7496298-7496298 on Build GRCh38
rs377195955 Chr.17: 7496205-7496205 on Build GRCh38
rs368513162 Chr.17: 7496410-7496410 on Build GRCh38
rs764717051 Chr.17: 7496435-7496435 on Build GRCh38
rs776064758 Chr.17: 7496223-7496223 on Build GRCh38
rs7219725 Chr.17: 7496511-7496511 on Build GRCh38
rs1034485225 Chr.17: 7496148-7496148 on Build GRCh38
rs761930497 Chr.17: 7496594-7496594 on Build GRCh38
rs913804292 Chr.17: 7496414-7496414 on Build GRCh38
rs780963366 Chr.17: 7496399-7496399 on Build GRCh38
rs750267917 Chr.17: 7496302-7496302 on Build GRCh38
rs769492340 Chr.17: 7496412-7496412 on Build GRCh38
rs751453915 Chr.17: 7496346-7496346 on Build GRCh38
rs981648610 Chr.17: 7496333-7496333 on Build GRCh38
rs375186066 Chr.17: 7496571-7496571 on Build GRCh38
rs755252517 Chr.17: 7496171-7496172 on Build GRCh38
rs1017012603 Chr.17: 7496245-7496245 on Build GRCh38
rs371439650 Chr.17: 7496466-7496466 on Build GRCh38
rs780314379 Chr.17: 7496502-7496502 on Build GRCh38
rs756158822 Chr.17: 7496361-7496361 on Build GRCh38
rs776158336 Chr.17: 7496407-7496407 on Build GRCh38
rs868359721 Chr.17: 7496272-7496272 on Build GRCh38
rs762784138 Chr.17: 7496306-7496306 on Build GRCh38
rs372957049 Chr.17: 7496181-7496181 on Build GRCh38
rs375583094 Chr.17: 7496483-7496483 on Build GRCh38
rs373099792 Chr.17: 7496159-7496159 on Build GRCh38
rs991499527 Chr.17: 7496429-7496429 on Build GRCh38
rs764250199 Chr.17: 7496441-7496441 on Build GRCh38
rs199962958 Chr.17: 7496454-7496454 on Build GRCh38
rs769200105 Chr.17: 7496510-7496510 on Build GRCh38
rs754069168 Chr.17: 7496377-7496377 on Build GRCh38
rs370068867 Chr.17: 7496244-7496244 on Build GRCh38
rs772330323 Chr.17: 7496402-7496402 on Build GRCh38
rs2301609 Chr.17: 7496547-7496547 on Build GRCh38
rs764407012 Chr.17: 7496464-7496464 on Build GRCh38
rs924679256 Chr.17: 7496560-7496560 on Build GRCh38
rs754159394 Chr.17: 7496514-7496514 on Build GRCh38
rs746360283 Chr.17: 7496202-7496202 on Build GRCh38
rs878921473 Chr.17: 7496548-7496548 on Build GRCh38
rs201627353 Chr.17: 7496177-7496177 on Build GRCh38
rs922388452 Chr.17: 7496382-7496382 on Build GRCh38
rs780554203 Chr.17: 7496403-7496403 on Build GRCh38
rs368365915 Chr.17: 7496568-7496568 on Build GRCh38
rs201165105 Chr.17: 7496378-7496378 on Build GRCh38
rs889389857 Chr.17: 7496592-7496592 on Build GRCh38
rs758500020 Chr.17: 7496579-7496579 on Build GRCh38
rs761485518 Chr.17: 7496292-7496292 on Build GRCh38
rs762738793 Chr.17: 7496423-7496423 on Build GRCh38
rs751461593 Chr.17: 7496430-7496430 on Build GRCh38
rs759418786 Chr.17: 7496437-7496437 on Build GRCh38
rs896074219 Chr.17: 7496154-7496154 on Build GRCh38
rs750625038 Chr.17: 7496467-7496467 on Build GRCh38
rs775575528 Chr.17: 7496585-7496585 on Build GRCh38
rs768172654 Chr.17: 7496404-7496404 on Build GRCh38
rs375461597 Chr.17: 7496153-7496153 on Build GRCh38
rs757551715 Chr.17: 7496463-7496463 on Build GRCh38

View Full Product Details