Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10392990-10393534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH8
Entrez Gene ID
4626
Gene Name
myosin heavy chain 8
Gene Aliases
DA7, MyHC-peri, MyHC-pn, gtMHC-F
UniGene
Hs.700484
Gene Chromosome Location
Chr.17: 10390325-10421950 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Gene Symbol
MYHAS
Entrez Gene ID
100128560
Gene Name
myosin heavy chain gene cluster antisense RNA
Gene Aliases
-
UniGene
Hs.661174
Gene Chromosome Location
Chr.17: 10383132-10625540 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7378379 c.5236G>T p.E1746* Chr.17: 10393141-10393141 on Build GRCh38
5021933 c.5208C>T p.D1736D Chr.17: 10393169-10393169 on Build GRCh38
6346531 c.5256C>A p.R1752R Chr.17: 10393121-10393121 on Build GRCh38
3513627 c.5282C>T p.A1761V Chr.17: 10393095-10393095 on Build GRCh38
4498540 c.5220C>T p.L1740L Chr.17: 10393157-10393157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34433607 Chr.17: 10393122-10393122 on Build GRCh38
rs759676157 Chr.17: 10393083-10393083 on Build GRCh38
rs776562346 Chr.17: 10393357-10393357 on Build GRCh38
rs774984611 Chr.17: 10393038-10393038 on Build GRCh38
rs764923155 Chr.17: 10393065-10393065 on Build GRCh38
rs775814737 Chr.17: 10393168-10393168 on Build GRCh38
rs748026075 Chr.17: 10393154-10393154 on Build GRCh38
rs747630397 Chr.17: 10393229-10393229 on Build GRCh38
rs559658341 Chr.17: 10393362-10393362 on Build GRCh38
rs773155072 Chr.17: 10393250-10393250 on Build GRCh38
rs892572649 Chr.17: 10393040-10393040 on Build GRCh38
rs746549585 Chr.17: 10393147-10393147 on Build GRCh38
rs770996617 Chr.17: 10393272-10393272 on Build GRCh38
rs767877486 Chr.17: 10393162-10393162 on Build GRCh38
rs756213751 Chr.17: 10393089-10393089 on Build GRCh38
rs771633535 Chr.17: 10393139-10393139 on Build GRCh38
rs764256012 Chr.17: 10393172-10393172 on Build GRCh38
rs776508765 Chr.17: 10393057-10393057 on Build GRCh38
rs769827979 Chr.17: 10393073-10393074 on Build GRCh38
rs547388985 Chr.17: 10393291-10393291 on Build GRCh38
rs33969260 Chr.17: 10393169-10393169 on Build GRCh38
rs750772829 Chr.17: 10393189-10393189 on Build GRCh38
rs770375928 Chr.17: 10393253-10393253 on Build GRCh38
rs769699528 Chr.17: 10393159-10393159 on Build GRCh38
rs752840291 Chr.17: 10393088-10393088 on Build GRCh38
rs888090987 Chr.17: 10393243-10393243 on Build GRCh38
rs746645725 Chr.17: 10393223-10393223 on Build GRCh38
rs1053856177 Chr.17: 10393017-10393017 on Build GRCh38
rs745493875 Chr.17: 10393018-10393018 on Build GRCh38
rs183160629 Chr.17: 10393486-10393486 on Build GRCh38
rs779629920 Chr.17: 10393141-10393141 on Build GRCh38
rs768120382 Chr.17: 10393151-10393151 on Build GRCh38
rs1005248097 Chr.17: 10393290-10393290 on Build GRCh38
rs368960642 Chr.17: 10393225-10393225 on Build GRCh38
rs1025529306 Chr.17: 10393387-10393387 on Build GRCh38
rs760117922 Chr.17: 10393043-10393043 on Build GRCh38
rs574711257 Chr.17: 10393051-10393051 on Build GRCh38
rs546220244 Chr.17: 10393213-10393213 on Build GRCh38
rs767783112 Chr.17: 10393085-10393085 on Build GRCh38
rs927196603 Chr.17: 10393288-10393288 on Build GRCh38
rs1049687093 Chr.17: 10393236-10393236 on Build GRCh38
rs768599210 Chr.17: 10393056-10393056 on Build GRCh38
rs946071917 Chr.17: 10393402-10393402 on Build GRCh38
rs563469677 Chr.17: 10393096-10393096 on Build GRCh38
rs150763013 Chr.17: 10393403-10393403 on Build GRCh38
rs541994027 Chr.17: 10393066-10393066 on Build GRCh38
rs779138246 Chr.17: 10393123-10393123 on Build GRCh38
rs202102936 Chr.17: 10393257-10393257 on Build GRCh38
rs746175156 Chr.17: 10393129-10393129 on Build GRCh38
rs1029245494 Chr.17: 10393133-10393133 on Build GRCh38
rs1010708924 Chr.17: 10393037-10393037 on Build GRCh38
rs374898030 Chr.17: 10393152-10393152 on Build GRCh38
rs762752971 Chr.17: 10393160-10393160 on Build GRCh38
rs773323805 Chr.17: 10393103-10393104 on Build GRCh38
rs895781250 Chr.17: 10393380-10393380 on Build GRCh38
rs564320858 Chr.17: 10393226-10393226 on Build GRCh38
rs779718071 Chr.17: 10393214-10393214 on Build GRCh38
rs777179030 Chr.17: 10393059-10393059 on Build GRCh38
rs757707741 Chr.17: 10393180-10393180 on Build GRCh38
rs754435722 Chr.17: 10393173-10393173 on Build GRCh38
rs765715898 Chr.17: 10393183-10393183 on Build GRCh38
rs769633113 Chr.17: 10393233-10393233 on Build GRCh38
rs771677133 Chr.17: 10393027-10393027 on Build GRCh38

View Full Product Details