Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32305260-32305706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDL3
Entrez Gene ID
162494
Gene Name
rhomboid like 3
Gene Aliases
RHBDL4, VRHO
UniGene
Hs.655027
Gene Chromosome Location
Chr.17: 32266167-32324661 on Build GRCh38
Transcripts
NM_138328.2 XM_006721733.3 XM_006721734.3 XM_011524443.2 XM_017024272.1 XM_017024273.1 XM_017024274.1 XM_017024275.1 XM_017024276.1 XM_017024277.1 XM_017024278.1 XM_017024279.1 XM_017024280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180349 c.791C>T p.A264V Chr.17: 32305350-32305350 on Build GRCh38
705607 c.814G>T p.A272S Chr.17: 32305373-32305373 on Build GRCh38
1302631 c.845A>G p.Y282C Chr.17: 32305404-32305404 on Build GRCh38
72424 c.810G>A p.M270I Chr.17: 32305369-32305369 on Build GRCh38
2151296 c.853G>A p.V285I Chr.17: 32305412-32305412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745783049 Chr.17: 32305492-32305512 on Build GRCh38
rs916393899 Chr.17: 32305390-32305390 on Build GRCh38
rs776345799 Chr.17: 32305428-32305428 on Build GRCh38
rs568521840 Chr.17: 32305429-32305429 on Build GRCh38
rs775009726 Chr.17: 32305323-32305323 on Build GRCh38
rs370833380 Chr.17: 32305328-32305328 on Build GRCh38
rs1056128218 Chr.17: 32305303-32305303 on Build GRCh38
rs376169535 Chr.17: 32305326-32305326 on Build GRCh38
rs367894973 Chr.17: 32305477-32305477 on Build GRCh38
rs765166550 Chr.17: 32305322-32305322 on Build GRCh38
rs780275276 Chr.17: 32305290-32305290 on Build GRCh38
rs374414149 Chr.17: 32305546-32305546 on Build GRCh38
rs762984410 Chr.17: 32305438-32305438 on Build GRCh38
rs773967232 Chr.17: 32305452-32305452 on Build GRCh38
rs998819072 Chr.17: 32305626-32305626 on Build GRCh38
rs759466340 Chr.17: 32305304-32305304 on Build GRCh38
rs534402793 Chr.17: 32305550-32305550 on Build GRCh38
rs202194628 Chr.17: 32305411-32305411 on Build GRCh38
rs111435152 Chr.17: 32305459-32305459 on Build GRCh38
rs371133539 Chr.17: 32305455-32305455 on Build GRCh38
rs769673185 Chr.17: 32305433-32305433 on Build GRCh38
rs756933865 Chr.17: 32305293-32305297 on Build GRCh38
rs1022205798 Chr.17: 32305564-32305564 on Build GRCh38
rs373519235 Chr.17: 32305298-32305298 on Build GRCh38
rs767282195 Chr.17: 32305454-32305454 on Build GRCh38
rs763486448 Chr.17: 32305451-32305451 on Build GRCh38
rs770996568 Chr.17: 32305325-32305325 on Build GRCh38
rs939074115 Chr.17: 32305629-32305629 on Build GRCh38
rs867201553 Chr.17: 32305443-32305443 on Build GRCh38
rs745679411 Chr.17: 32305430-32305430 on Build GRCh38
rs998288899 Chr.17: 32305589-32305589 on Build GRCh38
rs113792841 Chr.17: 32305310-32305310 on Build GRCh38
rs1032973796 Chr.17: 32305640-32305640 on Build GRCh38
rs370027112 Chr.17: 32305291-32305291 on Build GRCh38
rs900037611 Chr.17: 32305556-32305556 on Build GRCh38
rs562310637 Chr.17: 32305297-32305297 on Build GRCh38
rs140788529 Chr.17: 32305552-32305552 on Build GRCh38
rs767977861 Chr.17: 32305371-32305371 on Build GRCh38
rs778679339 Chr.17: 32305501-32305501 on Build GRCh38
rs762396309 Chr.17: 32305324-32305324 on Build GRCh38
rs755042629 Chr.17: 32305389-32305389 on Build GRCh38
rs201455379 Chr.17: 32305431-32305431 on Build GRCh38
rs766951422 Chr.17: 32305463-32305463 on Build GRCh38
rs754278894 Chr.17: 32305465-32305465 on Build GRCh38
rs181963320 Chr.17: 32305400-32305400 on Build GRCh38
rs749339761 Chr.17: 32305381-32305381 on Build GRCh38
rs764758836 Chr.17: 32305605-32305605 on Build GRCh38
rs774394584 Chr.17: 32305461-32305461 on Build GRCh38
rs373170971 Chr.17: 32305376-32305376 on Build GRCh38
rs908172279 Chr.17: 32305677-32305677 on Build GRCh38
rs898644592 Chr.17: 32305387-32305387 on Build GRCh38
rs886775529 Chr.17: 32305306-32305306 on Build GRCh38
rs926480974 Chr.17: 32305609-32305609 on Build GRCh38
rs562779986 Chr.17: 32305372-32305372 on Build GRCh38
rs779099300 Chr.17: 32305396-32305396 on Build GRCh38
rs753792419 Chr.17: 32305354-32305354 on Build GRCh38
rs776470971 Chr.17: 32305299-32305299 on Build GRCh38
rs9905406 Chr.17: 32305598-32305598 on Build GRCh38
rs756620606 Chr.17: 32305373-32305373 on Build GRCh38
rs779642788 Chr.17: 32305636-32305636 on Build GRCh38
rs761550396 Chr.17: 32305453-32305453 on Build GRCh38
rs775325414 Chr.17: 32305435-32305435 on Build GRCh38
rs374315132 Chr.17: 32305462-32305462 on Build GRCh38
rs957704679 Chr.17: 32305678-32305678 on Build GRCh38
rs62064223 Chr.17: 32305421-32305421 on Build GRCh38
rs71354615 Chr.17: 32305598-32305600 on Build GRCh38
rs756979452 Chr.17: 32305367-32305367 on Build GRCh38
rs9905407 Chr.17: 32305600-32305600 on Build GRCh38
rs78277760 Chr.17: 32305551-32305551 on Build GRCh38

View Full Product Details