Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35858345-35858852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
UniGene
Hs.514820
Gene Chromosome Location
Chr.17: 35854946-35868891 on Build GRCh38
Transcripts
NM_001321395.1 NM_152781.3 NR_135630.1 XM_005278233.2 XM_005278235.1 XM_006721802.3 XM_011524586.2 XM_017024424.1

SNP Detail

SNP ID Chromosome Location
rs12946529 Chr.17: 35858631-35858631 on Build GRCh38
rs12937552 Chr.17: 35858629-35858629 on Build GRCh38
rs769106396 Chr.17: 35858551-35858551 on Build GRCh38
rs377679199 Chr.17: 35858511-35858511 on Build GRCh38
rs374136110 Chr.17: 35858496-35858496 on Build GRCh38
rs749777362 Chr.17: 35858540-35858540 on Build GRCh38
rs373568857 Chr.17: 35858392-35858392 on Build GRCh38
rs760702659 Chr.17: 35858442-35858442 on Build GRCh38
rs780579350 Chr.17: 35858536-35858536 on Build GRCh38
rs12944206 Chr.17: 35858606-35858606 on Build GRCh38
rs902457505 Chr.17: 35858623-35858623 on Build GRCh38
rs967203394 Chr.17: 35858818-35858818 on Build GRCh38
rs144377349 Chr.17: 35858472-35858472 on Build GRCh38
rs770568908 Chr.17: 35858383-35858383 on Build GRCh38
rs774447864 Chr.17: 35858552-35858552 on Build GRCh38
rs547809745 Chr.17: 35858713-35858713 on Build GRCh38
rs543384579 Chr.17: 35858470-35858470 on Build GRCh38
rs570205615 Chr.17: 35858770-35858770 on Build GRCh38
rs762049546 Chr.17: 35858427-35858427 on Build GRCh38
rs200346168 Chr.17: 35858637-35858637 on Build GRCh38
rs200249075 Chr.17: 35858399-35858399 on Build GRCh38
rs1042567034 Chr.17: 35858595-35858595 on Build GRCh38
rs984758322 Chr.17: 35858479-35858479 on Build GRCh38
rs142132238 Chr.17: 35858500-35858500 on Build GRCh38
rs745661978 Chr.17: 35858572-35858573 on Build GRCh38
rs963400489 Chr.17: 35858799-35858799 on Build GRCh38
rs777604593 Chr.17: 35858368-35858368 on Build GRCh38
rs772238491 Chr.17: 35858566-35858566 on Build GRCh38
rs774608619 Chr.17: 35858412-35858412 on Build GRCh38
rs893159917 Chr.17: 35858711-35858711 on Build GRCh38
rs778952878 Chr.17: 35858728-35858728 on Build GRCh38
rs377305682 Chr.17: 35858640-35858647 on Build GRCh38
rs190272527 Chr.17: 35858757-35858757 on Build GRCh38
rs535561369 Chr.17: 35858648-35858648 on Build GRCh38
rs767374895 Chr.17: 35858682-35858682 on Build GRCh38
rs775964469 Chr.17: 35858398-35858398 on Build GRCh38
rs1017588497 Chr.17: 35858777-35858777 on Build GRCh38
rs925504311 Chr.17: 35858617-35858617 on Build GRCh38
rs976514479 Chr.17: 35858811-35858811 on Build GRCh38
rs866992203 Chr.17: 35858773-35858773 on Build GRCh38
rs371347293 Chr.17: 35858716-35858716 on Build GRCh38
rs746278293 Chr.17: 35858532-35858532 on Build GRCh38
rs767740920 Chr.17: 35858437-35858437 on Build GRCh38
rs116758864 Chr.17: 35858492-35858492 on Build GRCh38
rs770191012 Chr.17: 35858534-35858534 on Build GRCh38
rs398078973 Chr.17: 35858535-35858535 on Build GRCh38
rs754026541 Chr.17: 35858691-35858691 on Build GRCh38
rs1054695555 Chr.17: 35858706-35858706 on Build GRCh38
rs950332105 Chr.17: 35858487-35858487 on Build GRCh38
rs529097232 Chr.17: 35858501-35858501 on Build GRCh38
rs777062047 Chr.17: 35858597-35858597 on Build GRCh38
rs770217214 Chr.17: 35858397-35858397 on Build GRCh38
rs993995835 Chr.17: 35858741-35858741 on Build GRCh38
rs757431805 Chr.17: 35858481-35858482 on Build GRCh38
rs12944169 Chr.17: 35858565-35858565 on Build GRCh38
rs369839459 Chr.17: 35858370-35858370 on Build GRCh38
rs1035971736 Chr.17: 35858400-35858400 on Build GRCh38
rs761888886 Chr.17: 35858560-35858560 on Build GRCh38
rs974508265 Chr.17: 35858800-35858800 on Build GRCh38
rs566796784 Chr.17: 35858744-35858744 on Build GRCh38
rs201500225 Chr.17: 35858393-35858393 on Build GRCh38
rs4796118 Chr.17: 35858804-35858804 on Build GRCh38
rs756863952 Chr.17: 35858504-35858504 on Build GRCh38
rs906016996 Chr.17: 35858723-35858723 on Build GRCh38
rs952642837 Chr.17: 35858429-35858429 on Build GRCh38
rs1035917888 Chr.17: 35858740-35858740 on Build GRCh38
rs765152384 Chr.17: 35858491-35858491 on Build GRCh38
rs117659319 Chr.17: 35858714-35858714 on Build GRCh38
rs35283303 Chr.17: 35858532-35858532 on Build GRCh38
rs1053134827 Chr.17: 35858665-35858665 on Build GRCh38
rs527827833 Chr.17: 35858692-35858692 on Build GRCh38
rs1023079440 Chr.17: 35858721-35858721 on Build GRCh38
rs370864478 Chr.17: 35858567-35858567 on Build GRCh38
rs923371253 Chr.17: 35858814-35858814 on Build GRCh38
rs533034638 Chr.17: 35858436-35858436 on Build GRCh38
rs934618409 Chr.17: 35858647-35858647 on Build GRCh38
rs758027984 Chr.17: 35858495-35858495 on Build GRCh38
rs115778225 Chr.17: 35858727-35858727 on Build GRCh38
rs753583305 Chr.17: 35858468-35858468 on Build GRCh38

View Full Product Details