Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36486548-36487037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNHIT3
Entrez Gene ID
9326
Gene Name
zinc finger HIT-type containing 3
Gene Aliases
TRIP3
UniGene
Hs.2210
Gene Chromosome Location
Chr.17: 36486627-36499310 on Build GRCh38
Transcripts
NM_001281432.1 NM_001281433.1 NM_001281434.1 NM_004773.3 NR_104009.1 NR_104010.1 NR_104011.1

SNP Detail

SNP ID Chromosome Location
rs769665365 Chr.17: 36486998-36486998 on Build GRCh38
rs200810980 Chr.17: 36486689-36486689 on Build GRCh38
rs199868804 Chr.17: 36486831-36486831 on Build GRCh38
rs749204894 Chr.17: 36486655-36486673 on Build GRCh38
rs780481605 Chr.17: 36486920-36486920 on Build GRCh38
rs773857034 Chr.17: 36486791-36486791 on Build GRCh38
rs760072890 Chr.17: 36486713-36486713 on Build GRCh38
rs746932496 Chr.17: 36486817-36486817 on Build GRCh38
rs561478406 Chr.17: 36486624-36486624 on Build GRCh38
rs4796224 Chr.17: 36486677-36486677 on Build GRCh38
rs569362060 Chr.17: 36486700-36486700 on Build GRCh38
rs980266339 Chr.17: 36486628-36486628 on Build GRCh38
rs558390471 Chr.17: 36486636-36486636 on Build GRCh38
rs759172655 Chr.17: 36486832-36486839 on Build GRCh38
rs749896766 Chr.17: 36486687-36486687 on Build GRCh38
rs755283829 Chr.17: 36486691-36486691 on Build GRCh38
rs538474249 Chr.17: 36486704-36486704 on Build GRCh38
rs748006012 Chr.17: 36486813-36486813 on Build GRCh38
rs774224518 Chr.17: 36486707-36486707 on Build GRCh38
rs748657014 Chr.17: 36486697-36486697 on Build GRCh38
rs963658889 Chr.17: 36486938-36486938 on Build GRCh38
rs914654157 Chr.17: 36486608-36486608 on Build GRCh38
rs530544811 Chr.17: 36486975-36486975 on Build GRCh38
rs760298652 Chr.17: 36486758-36486758 on Build GRCh38
rs768029722 Chr.17: 36486889-36486889 on Build GRCh38
rs767143290 Chr.17: 36486686-36486686 on Build GRCh38
rs781548748 Chr.17: 36486736-36486736 on Build GRCh38
rs1043648176 Chr.17: 36487011-36487011 on Build GRCh38
rs531491854 Chr.17: 36486606-36486606 on Build GRCh38
rs779540622 Chr.17: 36486692-36486692 on Build GRCh38
rs772176410 Chr.17: 36486752-36486752 on Build GRCh38
rs774035017 Chr.17: 36487006-36487006 on Build GRCh38
rs886127462 Chr.17: 36486641-36486641 on Build GRCh38
rs770906091 Chr.17: 36486735-36486735 on Build GRCh38
rs145622035 Chr.17: 36486762-36486762 on Build GRCh38
rs193277074 Chr.17: 36486814-36486814 on Build GRCh38
rs370819821 Chr.17: 36487016-36487016 on Build GRCh38
rs745814158 Chr.17: 36486996-36486996 on Build GRCh38
rs186664380 Chr.17: 36486751-36486751 on Build GRCh38
rs148890852 Chr.17: 36486940-36486940 on Build GRCh38
rs775382491 Chr.17: 36486671-36486671 on Build GRCh38
rs752009346 Chr.17: 36486786-36486786 on Build GRCh38
rs377271926 Chr.17: 36486783-36486783 on Build GRCh38
rs775105534 Chr.17: 36486828-36486828 on Build GRCh38
rs539591362 Chr.17: 36486988-36486988 on Build GRCh38
rs768334714 Chr.17: 36486706-36486706 on Build GRCh38
rs542033659 Chr.17: 36486869-36486869 on Build GRCh38
rs759051840 Chr.17: 36486827-36486827 on Build GRCh38
rs774311793 Chr.17: 36486750-36486750 on Build GRCh38
rs200363039 Chr.17: 36486942-36486942 on Build GRCh38
rs773261460 Chr.17: 36486757-36486757 on Build GRCh38
rs781054474 Chr.17: 36486994-36486994 on Build GRCh38
rs756451703 Chr.17: 36486599-36486599 on Build GRCh38
rs759131853 Chr.17: 36486766-36486766 on Build GRCh38
rs948138831 Chr.17: 36486842-36486849 on Build GRCh38
rs778682737 Chr.17: 36486680-36486680 on Build GRCh38
rs374337340 Chr.17: 36486775-36486775 on Build GRCh38
rs149756417 Chr.17: 36486726-36486726 on Build GRCh38
rs576252506 Chr.17: 36486834-36486834 on Build GRCh38
rs957322240 Chr.17: 36486999-36486999 on Build GRCh38
rs769285853 Chr.17: 36486953-36486953 on Build GRCh38
rs771007870 Chr.17: 36486665-36486665 on Build GRCh38
rs746085924 Chr.17: 36486739-36486739 on Build GRCh38
rs561810642 Chr.17: 36487010-36487010 on Build GRCh38
rs372257914 Chr.17: 36486759-36486759 on Build GRCh38
rs770851325 Chr.17: 36486819-36486819 on Build GRCh38
rs757730054 Chr.17: 36486888-36486888 on Build GRCh38
rs940357872 Chr.17: 36486654-36486654 on Build GRCh38
rs908112023 Chr.17: 36486741-36486741 on Build GRCh38
rs751358002 Chr.17: 36486989-36486989 on Build GRCh38
rs79177936 Chr.17: 36486609-36486609 on Build GRCh38
rs768251201 Chr.17: 36486958-36486958 on Build GRCh38
rs11542618 Chr.17: 36486761-36486761 on Build GRCh38
rs773207122 Chr.17: 36486661-36486661 on Build GRCh38
rs757720388 Chr.17: 36486788-36486788 on Build GRCh38
rs546623186 Chr.17: 36486638-36486638 on Build GRCh38
rs764827911 Chr.17: 36486770-36486770 on Build GRCh38
rs374569954 Chr.17: 36486672-36486672 on Build GRCh38
rs371328466 Chr.17: 36486688-36486688 on Build GRCh38
rs774613526 Chr.17: 36486959-36486959 on Build GRCh38
rs935338872 Chr.17: 36486647-36486647 on Build GRCh38
rs146112314 Chr.17: 36487001-36487001 on Build GRCh38
rs781163791 Chr.17: 36486948-36486948 on Build GRCh38
rs770356090 Chr.17: 36486685-36486686 on Build GRCh38
rs760898790 Chr.17: 36486664-36486664 on Build GRCh38
rs766578415 Chr.17: 36486983-36486983 on Build GRCh38
rs201411835 Chr.17: 36486684-36486684 on Build GRCh38
rs749525653 Chr.17: 36486921-36486921 on Build GRCh38
rs368024761 Chr.17: 36486716-36486716 on Build GRCh38
rs560117316 Chr.17: 36486955-36486955 on Build GRCh38
rs898922614 Chr.17: 36486669-36486669 on Build GRCh38
rs745436762 Chr.17: 36486950-36486950 on Build GRCh38
rs762207379 Chr.17: 36486963-36486963 on Build GRCh38
rs201228623 Chr.17: 36486954-36486954 on Build GRCh38
rs1003704440 Chr.17: 36486880-36486881 on Build GRCh38
rs773074626 Chr.17: 36486816-36486816 on Build GRCh38
rs753007806 Chr.17: 36486722-36486722 on Build GRCh38
rs770002937 Chr.17: 36486740-36486740 on Build GRCh38
rs749986394 Chr.17: 36486711-36486711 on Build GRCh38
rs1034437372 Chr.17: 36486842-36486842 on Build GRCh38
rs140081743 Chr.17: 36486703-36486703 on Build GRCh38
rs763379295 Chr.17: 36486829-36486829 on Build GRCh38
rs372589679 Chr.17: 36486790-36486790 on Build GRCh38
rs139241535 Chr.17: 36486723-36486723 on Build GRCh38
rs755192268 Chr.17: 36486923-36486923 on Build GRCh38
rs1057367440 Chr.17: 36486640-36486640 on Build GRCh38
rs772133790 Chr.17: 36486659-36486659 on Build GRCh38
rs200852859 Chr.17: 36486964-36486964 on Build GRCh38
rs756240392 Chr.17: 36486913-36486913 on Build GRCh38
rs1050632293 Chr.17: 36486979-36486979 on Build GRCh38
rs569582446 Chr.17: 36486986-36486986 on Build GRCh38
rs1047800473 Chr.17: 36486912-36486912 on Build GRCh38
rs762867908 Chr.17: 36486705-36486705 on Build GRCh38
rs747135956 Chr.17: 36486728-36486728 on Build GRCh38
rs767155108 Chr.17: 36486831-36486832 on Build GRCh38
rs768234500 Chr.17: 36487005-36487005 on Build GRCh38
rs933922050 Chr.17: 36486629-36486630 on Build GRCh38
rs745915202 Chr.17: 36486793-36486793 on Build GRCh38
rs777022871 Chr.17: 36486904-36486904 on Build GRCh38
rs772481728 Chr.17: 36487017-36487017 on Build GRCh38
rs776032979 Chr.17: 36486825-36486825 on Build GRCh38
rs750510364 Chr.17: 36486906-36486906 on Build GRCh38
rs757057781 Chr.17: 36486991-36486991 on Build GRCh38
rs372878191 Chr.17: 36486932-36486932 on Build GRCh38
rs780393657 Chr.17: 36486812-36486812 on Build GRCh38
rs938943430 Chr.17: 36486639-36486639 on Build GRCh38
rs762144332 Chr.17: 36486924-36486926 on Build GRCh38
rs112194209 Chr.17: 36486720-36486720 on Build GRCh38
rs561715809 Chr.17: 36486879-36486879 on Build GRCh38
rs906678475 Chr.17: 36486690-36486690 on Build GRCh38
rs755027653 Chr.17: 36486876-36486876 on Build GRCh38
rs143086637 Chr.17: 36486794-36486794 on Build GRCh38
rs980207386 Chr.17: 36486841-36486841 on Build GRCh38
rs760904333 Chr.17: 36486978-36486978 on Build GRCh38
rs143659130 Chr.17: 36486710-36486710 on Build GRCh38
rs1010807827 Chr.17: 36486826-36486826 on Build GRCh38
rs751532591 Chr.17: 36486682-36486682 on Build GRCh38
rs766104988 Chr.17: 36486715-36486715 on Build GRCh38
rs532017720 Chr.17: 36486841-36486842 on Build GRCh38
rs765537052 Chr.17: 36486972-36486972 on Build GRCh38
rs931759890 Chr.17: 36486760-36486760 on Build GRCh38
rs748759366 Chr.17: 36487003-36487003 on Build GRCh38
rs751895914 Chr.17: 36486884-36486884 on Build GRCh38
rs552279678 Chr.17: 36486985-36486985 on Build GRCh38
rs571069922 Chr.17: 36486974-36486974 on Build GRCh38
rs781148422 Chr.17: 36486792-36486792 on Build GRCh38

View Full Product Details