Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36591654-36592160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS11
Entrez Gene ID
79154
Gene Name
dehydrogenase/reductase 11
Gene Aliases
ARPG836, SDR24C1, spDHRS11
UniGene
Hs.462859
Gene Chromosome Location
Chr.17: 36591791-36600804 on Build GRCh38
Transcripts
NM_024308.3 XM_005257658.2 XM_011525233.2

SNP Detail

SNP ID Chromosome Location
rs532197802 Chr.17: 36591815-36591815 on Build GRCh38
rs1011756630 Chr.17: 36591920-36591921 on Build GRCh38
rs879260339 Chr.17: 36592086-36592086 on Build GRCh38
rs892743423 Chr.17: 36592014-36592014 on Build GRCh38
rs969834307 Chr.17: 36591994-36591994 on Build GRCh38
rs767085919 Chr.17: 36591701-36591701 on Build GRCh38
rs867558042 Chr.17: 36591908-36591908 on Build GRCh38
rs941216842 Chr.17: 36591728-36591728 on Build GRCh38
rs932653828 Chr.17: 36591868-36591868 on Build GRCh38
rs974353258 Chr.17: 36591921-36591921 on Build GRCh38
rs563733426 Chr.17: 36591865-36591865 on Build GRCh38
rs1011305361 Chr.17: 36591772-36591772 on Build GRCh38
rs931999322 Chr.17: 36591735-36591735 on Build GRCh38
rs550268452 Chr.17: 36592111-36592111 on Build GRCh38
rs747568347 Chr.17: 36592108-36592108 on Build GRCh38
rs754606247 Chr.17: 36592051-36592051 on Build GRCh38
rs747664624 Chr.17: 36592094-36592094 on Build GRCh38
rs571831892 Chr.17: 36592040-36592040 on Build GRCh38
rs1022035109 Chr.17: 36591842-36591842 on Build GRCh38
rs1047764246 Chr.17: 36592060-36592069 on Build GRCh38
rs549395888 Chr.17: 36591879-36591879 on Build GRCh38
rs891359696 Chr.17: 36591878-36591878 on Build GRCh38
rs890674238 Chr.17: 36591739-36591739 on Build GRCh38
rs893278055 Chr.17: 36591911-36591911 on Build GRCh38
rs920651194 Chr.17: 36591928-36591928 on Build GRCh38
rs960591719 Chr.17: 36591890-36591890 on Build GRCh38
rs796668065 Chr.17: 36592071-36592071 on Build GRCh38
rs909024108 Chr.17: 36591859-36591859 on Build GRCh38
rs529310393 Chr.17: 36591867-36591867 on Build GRCh38
rs560013035 Chr.17: 36591801-36591801 on Build GRCh38
rs1011519107 Chr.17: 36591809-36591809 on Build GRCh38
rs932075491 Chr.17: 36591932-36591932 on Build GRCh38
rs1051138125 Chr.17: 36591873-36591873 on Build GRCh38
rs766784719 Chr.17: 36591901-36591901 on Build GRCh38
rs778505157 Chr.17: 36592073-36592073 on Build GRCh38
rs1035389052 Chr.17: 36592023-36592023 on Build GRCh38
rs983673494 Chr.17: 36591845-36591845 on Build GRCh38
rs771773444 Chr.17: 36592097-36592097 on Build GRCh38
rs552992797 Chr.17: 36591677-36591677 on Build GRCh38
rs1011070389 Chr.17: 36592021-36592021 on Build GRCh38
rs890474162 Chr.17: 36591934-36591934 on Build GRCh38
rs565071162 Chr.17: 36591724-36591724 on Build GRCh38
rs1047595480 Chr.17: 36591733-36591733 on Build GRCh38
rs960348595 Chr.17: 36592104-36592104 on Build GRCh38
rs1022682055 Chr.17: 36591931-36591931 on Build GRCh38
rs999423728 Chr.17: 36591765-36591765 on Build GRCh38
rs1034699395 Chr.17: 36591869-36591869 on Build GRCh38
rs930506110 Chr.17: 36591807-36591807 on Build GRCh38
rs982679731 Chr.17: 36591899-36591899 on Build GRCh38
rs559836655 Chr.17: 36591881-36591881 on Build GRCh38
rs1001974175 Chr.17: 36592080-36592080 on Build GRCh38
rs920706257 Chr.17: 36591768-36591789 on Build GRCh38
rs930696763 Chr.17: 36591797-36591810 on Build GRCh38
rs1015508707 Chr.17: 36592114-36592114 on Build GRCh38
rs528766701 Chr.17: 36591907-36591907 on Build GRCh38
rs1037778700 Chr.17: 36591731-36591731 on Build GRCh38
rs551715766 Chr.17: 36591933-36591933 on Build GRCh38
rs962950560 Chr.17: 36592128-36592128 on Build GRCh38
rs111780615 Chr.17: 36591726-36591726 on Build GRCh38
rs1050388875 Chr.17: 36591736-36591736 on Build GRCh38
rs908485144 Chr.17: 36591905-36591905 on Build GRCh38
rs62069094 Chr.17: 36591732-36591732 on Build GRCh38
rs999139847 Chr.17: 36591885-36591886 on Build GRCh38
rs973899759 Chr.17: 36592137-36592137 on Build GRCh38
rs778411015 Chr.17: 36592100-36592100 on Build GRCh38
rs1009116028 Chr.17: 36591742-36591742 on Build GRCh38
rs543950975 Chr.17: 36591770-36591770 on Build GRCh38
rs963143057 Chr.17: 36592060-36592060 on Build GRCh38
rs573087335 Chr.17: 36591678-36591678 on Build GRCh38
rs544911585 Chr.17: 36591723-36591723 on Build GRCh38
rs939259650 Chr.17: 36591675-36591675 on Build GRCh38
rs575484103 Chr.17: 36591754-36591754 on Build GRCh38
rs1052687760 Chr.17: 36591986-36591986 on Build GRCh38
rs944731293 Chr.17: 36591942-36591942 on Build GRCh38
rs753485461 Chr.17: 36592043-36592043 on Build GRCh38
rs753818046 Chr.17: 36591773-36591780 on Build GRCh38
rs537212851 Chr.17: 36592064-36592064 on Build GRCh38

View Full Product Details