Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37614110-37614612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX52
Entrez Gene ID
11056
Gene Name
DEAD-box helicase 52
Gene Aliases
HUSSY19, ROK1
UniGene
Hs.590937
Gene Chromosome Location
Chr.17: 37609739-37643464 on Build GRCh38
Transcripts
NM_001291476.1 NM_007010.4 XM_011524232.2 XM_011524233.2

SNP Detail

SNP ID Chromosome Location
rs779391992 Chr.17: 37614320-37614320 on Build GRCh38
rs986974717 Chr.17: 37614257-37614257 on Build GRCh38
rs772343070 Chr.17: 37614352-37614352 on Build GRCh38
rs755592137 Chr.17: 37614305-37614305 on Build GRCh38
rs371644738 Chr.17: 37614369-37614369 on Build GRCh38
rs992732867 Chr.17: 37614204-37614204 on Build GRCh38
rs992805617 Chr.17: 37614590-37614590 on Build GRCh38
rs528469568 Chr.17: 37614406-37614406 on Build GRCh38
rs374401705 Chr.17: 37614276-37614276 on Build GRCh38
rs776746377 Chr.17: 37614364-37614364 on Build GRCh38
rs751402086 Chr.17: 37614251-37614251 on Build GRCh38
rs181867387 Chr.17: 37614580-37614580 on Build GRCh38
rs931759999 Chr.17: 37614280-37614283 on Build GRCh38
rs551713589 Chr.17: 37614474-37614474 on Build GRCh38
rs777993218 Chr.17: 37614355-37614355 on Build GRCh38
rs200428925 Chr.17: 37614396-37614396 on Build GRCh38
rs367580227 Chr.17: 37614361-37614361 on Build GRCh38
rs980176900 Chr.17: 37614479-37614479 on Build GRCh38
rs1050153937 Chr.17: 37614284-37614284 on Build GRCh38
rs754383924 Chr.17: 37614303-37614303 on Build GRCh38
rs765965929 Chr.17: 37614397-37614397 on Build GRCh38
rs1004741826 Chr.17: 37614554-37614554 on Build GRCh38
rs968853556 Chr.17: 37614475-37614475 on Build GRCh38
rs1057041945 Chr.17: 37614486-37614486 on Build GRCh38
rs368002824 Chr.17: 37614395-37614395 on Build GRCh38
rs1002807209 Chr.17: 37614440-37614440 on Build GRCh38
rs779780170 Chr.17: 37614300-37614300 on Build GRCh38
rs940578319 Chr.17: 37614247-37614247 on Build GRCh38
rs747557125 Chr.17: 37614434-37614434 on Build GRCh38
rs755588309 Chr.17: 37614297-37614297 on Build GRCh38
rs771039068 Chr.17: 37614363-37614363 on Build GRCh38
rs753439113 Chr.17: 37614398-37614398 on Build GRCh38
rs756825321 Chr.17: 37614258-37614258 on Build GRCh38
rs748486159 Chr.17: 37614326-37614326 on Build GRCh38
rs563838711 Chr.17: 37614221-37614221 on Build GRCh38
rs565255152 Chr.17: 37614560-37614560 on Build GRCh38
rs75350315 Chr.17: 37614210-37614210 on Build GRCh38
rs574421986 Chr.17: 37614269-37614269 on Build GRCh38
rs886341906 Chr.17: 37614504-37614504 on Build GRCh38
rs750089291 Chr.17: 37614392-37614392 on Build GRCh38
rs905822391 Chr.17: 37614415-37614415 on Build GRCh38
rs974419077 Chr.17: 37614149-37614149 on Build GRCh38
rs202139121 Chr.17: 37614346-37614346 on Build GRCh38
rs59823735 Chr.17: 37614378-37614378 on Build GRCh38
rs774300572 Chr.17: 37614368-37614368 on Build GRCh38
rs768617516 Chr.17: 37614367-37614367 on Build GRCh38
rs780517825 Chr.17: 37614401-37614401 on Build GRCh38

View Full Product Details