Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39665927-39666432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAP
Entrez Gene ID
8557
Gene Name
titin-cap
Gene Aliases
CMD1N, CMH25, LGMD2G, T-cap, TELE, telethonin
UniGene
Hs.77628
Gene Chromosome Location
Chr.17: 39665346-39666554 on Build GRCh38
Transcripts
NM_003673.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333085 c.436G>T p.V146F Chr.17: 39666041-39666041 on Build GRCh38
1640674 c.411A>G p.T137T Chr.17: 39666016-39666016 on Build GRCh38
6315906 c.429G>T p.V143V Chr.17: 39666034-39666034 on Build GRCh38
6252132 c.354G>T p.A118A Chr.17: 39665959-39665959 on Build GRCh38
5456610 c.389G>A p.R130H Chr.17: 39665994-39665994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941213158 Chr.17: 39666054-39666054 on Build GRCh38
rs779217085 Chr.17: 39666102-39666102 on Build GRCh38
rs748358368 Chr.17: 39665999-39665999 on Build GRCh38
rs397516863 Chr.17: 39666077-39666077 on Build GRCh38
rs149585781 Chr.17: 39666063-39666063 on Build GRCh38
rs876657586 Chr.17: 39665951-39665951 on Build GRCh38
rs755539784 Chr.17: 39666065-39666065 on Build GRCh38
rs985223813 Chr.17: 39666226-39666226 on Build GRCh38
rs769800632 Chr.17: 39666041-39666041 on Build GRCh38
rs45509691 Chr.17: 39666026-39666026 on Build GRCh38
rs950261878 Chr.17: 39666074-39666074 on Build GRCh38
rs759564316 Chr.17: 39666138-39666138 on Build GRCh38
rs780544264 Chr.17: 39666066-39666066 on Build GRCh38
rs964326898 Chr.17: 39666029-39666029 on Build GRCh38
rs908004856 Chr.17: 39666067-39666067 on Build GRCh38
rs762976588 Chr.17: 39666160-39666160 on Build GRCh38
rs786205287 Chr.17: 39666006-39666006 on Build GRCh38
rs777817968 Chr.17: 39666139-39666139 on Build GRCh38
rs761498487 Chr.17: 39666062-39666062 on Build GRCh38
rs886038534 Chr.17: 39665954-39665954 on Build GRCh38
rs1053651 Chr.17: 39666058-39666058 on Build GRCh38
rs774447573 Chr.17: 39666128-39666143 on Build GRCh38
rs755395354 Chr.17: 39666085-39666085 on Build GRCh38
rs964465412 Chr.17: 39666249-39666249 on Build GRCh38
rs375843419 Chr.17: 39666141-39666141 on Build GRCh38
rs45495400 Chr.17: 39666125-39666125 on Build GRCh38
rs773317399 Chr.17: 39666015-39666015 on Build GRCh38
rs902320555 Chr.17: 39666232-39666232 on Build GRCh38
rs397516864 Chr.17: 39666078-39666078 on Build GRCh38
rs1027238303 Chr.17: 39666300-39666300 on Build GRCh38
rs143233087 Chr.17: 39665958-39665958 on Build GRCh38
rs771072422 Chr.17: 39666142-39666142 on Build GRCh38
rs558480997 Chr.17: 39665982-39665982 on Build GRCh38
rs749685550 Chr.17: 39665983-39665983 on Build GRCh38
rs1057106389 Chr.17: 39666083-39666083 on Build GRCh38
rs779158527 Chr.17: 39666320-39666320 on Build GRCh38
rs763965899 Chr.17: 39666057-39666057 on Build GRCh38
rs1020883300 Chr.17: 39666203-39666203 on Build GRCh38
rs977426720 Chr.17: 39666172-39666172 on Build GRCh38
rs45506294 Chr.17: 39666185-39666185 on Build GRCh38
rs374886575 Chr.17: 39665993-39665993 on Build GRCh38
rs200333339 Chr.17: 39666131-39666131 on Build GRCh38
rs1031873194 Chr.17: 39666166-39666166 on Build GRCh38
rs185210946 Chr.17: 39666401-39666401 on Build GRCh38
rs147503632 Chr.17: 39665994-39665994 on Build GRCh38
rs1056790670 Chr.17: 39666279-39666279 on Build GRCh38
rs397516865 Chr.17: 39666098-39666098 on Build GRCh38
rs1012966829 Chr.17: 39666137-39666137 on Build GRCh38
rs956340161 Chr.17: 39666168-39666168 on Build GRCh38
rs748365337 Chr.17: 39666112-39666112 on Build GRCh38
rs45503594 Chr.17: 39666308-39666308 on Build GRCh38
rs909664509 Chr.17: 39666227-39666227 on Build GRCh38
rs794729176 Chr.17: 39666080-39666080 on Build GRCh38
rs974378301 Chr.17: 39666257-39666257 on Build GRCh38
rs45614332 Chr.17: 39666052-39666052 on Build GRCh38
rs771363240 Chr.17: 39666065-39666076 on Build GRCh38
rs773653957 Chr.17: 39665979-39665993 on Build GRCh38
rs762850913 Chr.17: 39666053-39666053 on Build GRCh38
rs794729177 Chr.17: 39666104-39666104 on Build GRCh38
rs1019098438 Chr.17: 39666237-39666237 on Build GRCh38
rs932397209 Chr.17: 39666050-39666050 on Build GRCh38
rs768942598 Chr.17: 39665992-39665992 on Build GRCh38
rs895778819 Chr.17: 39666306-39666306 on Build GRCh38
rs1008753988 Chr.17: 39666234-39666234 on Build GRCh38
rs45592941 Chr.17: 39666163-39666163 on Build GRCh38
rs775624321 Chr.17: 39666143-39666143 on Build GRCh38
rs749817987 Chr.17: 39666057-39666074 on Build GRCh38
rs772083153 Chr.17: 39666007-39666007 on Build GRCh38
rs772638575 Chr.17: 39666406-39666406 on Build GRCh38
rs1010794675 Chr.17: 39666194-39666194 on Build GRCh38

View Full Product Details