Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64496714-64497203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLG2
Entrez Gene ID
11232
Gene Name
DNA polymerase gamma 2, accessory subunit
Gene Aliases
HP55, MTPOLB, PEOA4, POLB, POLG-BETA, POLGB
UniGene
Hs.437009
Gene Chromosome Location
Chr.17: 64477785-64497066 on Build GRCh38
Transcripts
NM_007215.3 XM_006721651.2 XM_017024072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982898 c.165G>A p.E55E Chr.17: 64496804-64496804 on Build GRCh38
6752093 c.59G>T p.G20V Chr.17: 64496910-64496910 on Build GRCh38
6023237 c.56C>T p.S19F Chr.17: 64496913-64496913 on Build GRCh38
3672532 c.102G>T p.T34T Chr.17: 64496867-64496867 on Build GRCh38
6243052 c.175G>A p.A59T Chr.17: 64496794-64496794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782625172 Chr.17: 64496901-64496901 on Build GRCh38
rs781977946 Chr.17: 64497081-64497081 on Build GRCh38
rs782727922 Chr.17: 64496977-64496977 on Build GRCh38
rs782416621 Chr.17: 64496748-64496748 on Build GRCh38
rs782196764 Chr.17: 64497011-64497011 on Build GRCh38
rs200431365 Chr.17: 64496807-64496807 on Build GRCh38
rs892296046 Chr.17: 64497053-64497053 on Build GRCh38
rs560778693 Chr.17: 64497061-64497061 on Build GRCh38
rs782520895 Chr.17: 64496976-64496976 on Build GRCh38
rs782621029 Chr.17: 64496794-64496794 on Build GRCh38
rs782676579 Chr.17: 64496818-64496818 on Build GRCh38
rs569460394 Chr.17: 64497035-64497035 on Build GRCh38
rs782621486 Chr.17: 64496908-64496908 on Build GRCh38
rs782366432 Chr.17: 64497009-64497009 on Build GRCh38
rs904953893 Chr.17: 64497150-64497150 on Build GRCh38
rs112240735 Chr.17: 64496968-64496968 on Build GRCh38
rs552693721 Chr.17: 64497029-64497029 on Build GRCh38
rs1040051640 Chr.17: 64497043-64497043 on Build GRCh38
rs782445961 Chr.17: 64496986-64496986 on Build GRCh38
rs372002094 Chr.17: 64497082-64497082 on Build GRCh38
rs901799025 Chr.17: 64497114-64497114 on Build GRCh38
rs965620329 Chr.17: 64496736-64496736 on Build GRCh38
rs901109833 Chr.17: 64497044-64497044 on Build GRCh38
rs782291957 Chr.17: 64496993-64496993 on Build GRCh38
rs1026556126 Chr.17: 64497174-64497174 on Build GRCh38
rs782103896 Chr.17: 64496951-64496951 on Build GRCh38
rs892328648 Chr.17: 64497138-64497138 on Build GRCh38
rs367696388 Chr.17: 64497003-64497003 on Build GRCh38
rs117792639 Chr.17: 64496889-64496889 on Build GRCh38
rs939575561 Chr.17: 64496967-64496967 on Build GRCh38
rs372637401 Chr.17: 64497008-64497008 on Build GRCh38
rs781936160 Chr.17: 64496958-64496958 on Build GRCh38
rs781847382 Chr.17: 64496974-64496974 on Build GRCh38
rs1049172633 Chr.17: 64497032-64497032 on Build GRCh38
rs782466597 Chr.17: 64496792-64496792 on Build GRCh38
rs781840547 Chr.17: 64496862-64496862 on Build GRCh38
rs782118666 Chr.17: 64496942-64496942 on Build GRCh38
rs41563537 Chr.17: 64497151-64497151 on Build GRCh38
rs782076699 Chr.17: 64496835-64496835 on Build GRCh38
rs782553388 Chr.17: 64496896-64496896 on Build GRCh38
rs782787611 Chr.17: 64496742-64496742 on Build GRCh38
rs144148008 Chr.17: 64496788-64496788 on Build GRCh38
rs782583101 Chr.17: 64496880-64496880 on Build GRCh38
rs782038358 Chr.17: 64496935-64496935 on Build GRCh38
rs952215489 Chr.17: 64497180-64497180 on Build GRCh38
rs781955943 Chr.17: 64496943-64496943 on Build GRCh38
rs140885161 Chr.17: 64496965-64496965 on Build GRCh38
rs548473967 Chr.17: 64496846-64496847 on Build GRCh38
rs782180971 Chr.17: 64496806-64496806 on Build GRCh38
rs930718069 Chr.17: 64497024-64497024 on Build GRCh38
rs556280294 Chr.17: 64497090-64497090 on Build GRCh38
rs889151949 Chr.17: 64497033-64497033 on Build GRCh38
rs547806545 Chr.17: 64497040-64497040 on Build GRCh38
rs782090879 Chr.17: 64497083-64497083 on Build GRCh38
rs781993155 Chr.17: 64496824-64496824 on Build GRCh38
rs41558714 Chr.17: 64497158-64497158 on Build GRCh38
rs886053253 Chr.17: 64497055-64497055 on Build GRCh38
rs960651032 Chr.17: 64497172-64497172 on Build GRCh38
rs998014247 Chr.17: 64496741-64496741 on Build GRCh38
rs540779392 Chr.17: 64497149-64497149 on Build GRCh38
rs139333917 Chr.17: 64496746-64496746 on Build GRCh38
rs553575891 Chr.17: 64497050-64497050 on Build GRCh38
rs555028462 Chr.17: 64497106-64497106 on Build GRCh38
rs886053252 Chr.17: 64497053-64497053 on Build GRCh38
rs1010643325 Chr.17: 64497056-64497056 on Build GRCh38
rs782128051 Chr.17: 64496853-64496853 on Build GRCh38
rs782445874 Chr.17: 64496868-64496868 on Build GRCh38
rs574879318 Chr.17: 64497111-64497111 on Build GRCh38
rs376183896 Chr.17: 64497016-64497016 on Build GRCh38
rs375375131 Chr.17: 64497121-64497121 on Build GRCh38
rs373387408 Chr.17: 64496953-64496953 on Build GRCh38
rs532597297 Chr.17: 64496959-64496959 on Build GRCh38
rs146504115 Chr.17: 64496847-64496847 on Build GRCh38
rs1010675817 Chr.17: 64497145-64497145 on Build GRCh38
rs782746699 Chr.17: 64496859-64496859 on Build GRCh38
rs972271544 Chr.17: 64497089-64497089 on Build GRCh38
rs782418665 Chr.17: 64496977-64496978 on Build GRCh38
rs782252477 Chr.17: 64496904-64496904 on Build GRCh38
rs886053251 Chr.17: 64497034-64497034 on Build GRCh38
rs782350159 Chr.17: 64496839-64496839 on Build GRCh38
rs138418765 Chr.17: 64497038-64497038 on Build GRCh38
rs34508296 Chr.17: 64497108-64497109 on Build GRCh38
rs368241192 Chr.17: 64497018-64497018 on Build GRCh38
rs532600526 Chr.17: 64497173-64497173 on Build GRCh38
rs782806271 Chr.17: 64496973-64496973 on Build GRCh38
rs781834593 Chr.17: 64496980-64496980 on Build GRCh38
rs910621672 Chr.17: 64497101-64497101 on Build GRCh38
rs782505866 Chr.17: 64497002-64497002 on Build GRCh38
rs561230251 Chr.17: 64496799-64496799 on Build GRCh38
rs782720561 Chr.17: 64496765-64496765 on Build GRCh38
rs969072131 Chr.17: 64496930-64496930 on Build GRCh38
rs969482784 Chr.17: 64497075-64497075 on Build GRCh38
rs782515312 Chr.17: 64497073-64497073 on Build GRCh38
rs1034695512 Chr.17: 64497165-64497165 on Build GRCh38
rs782626115 Chr.17: 64496786-64496786 on Build GRCh38
rs782372036 Chr.17: 64496823-64496823 on Build GRCh38
rs781866535 Chr.17: 64496850-64496850 on Build GRCh38
rs782570089 Chr.17: 64496781-64496781 on Build GRCh38
rs113949663 Chr.17: 64497094-64497094 on Build GRCh38
rs1052294892 Chr.17: 64497129-64497129 on Build GRCh38
rs567697094 Chr.17: 64497045-64497045 on Build GRCh38
rs764343802 Chr.17: 64496950-64496950 on Build GRCh38
rs933946682 Chr.17: 64497117-64497117 on Build GRCh38
rs537261236 Chr.17: 64497088-64497088 on Build GRCh38
rs201278573 Chr.17: 64496844-64496844 on Build GRCh38
rs782194725 Chr.17: 64496905-64496905 on Build GRCh38
rs781820330 Chr.17: 64496745-64496745 on Build GRCh38
rs542115612 Chr.17: 64497100-64497100 on Build GRCh38
rs782056821 Chr.17: 64496753-64496753 on Build GRCh38
rs782664223 Chr.17: 64496813-64496813 on Build GRCh38
rs9897606 Chr.17: 64497049-64497049 on Build GRCh38
rs781939715 Chr.17: 64496840-64496840 on Build GRCh38
rs781897145 Chr.17: 64496766-64496766 on Build GRCh38

View Full Product Details