Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67071997-67072538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELZ
Entrez Gene ID
9931
Gene Name
helicase with zinc finger
Gene Aliases
DHRC, DRHC, HUMORF5
UniGene
Hs.370140
Gene Chromosome Location
Chr.17: 67070438-67245989 on Build GRCh38
Transcripts
NM_014877.3 XM_005257888.4 XM_005257889.4 XM_006722214.3 XM_006722215.3 XM_006722216.3 XM_011525544.2 XM_017025477.1 XM_017025478.1

SNP Detail

SNP ID Chromosome Location
rs957741457 Chr.17: 67072440-67072440 on Build GRCh38
rs944709232 Chr.17: 67072319-67072319 on Build GRCh38
rs904605044 Chr.17: 67072102-67072102 on Build GRCh38
rs1053129686 Chr.17: 67072312-67072312 on Build GRCh38
rs752231338 Chr.17: 67072268-67072268 on Build GRCh38
rs1049185462 Chr.17: 67072347-67072347 on Build GRCh38
rs367843508 Chr.17: 67072261-67072261 on Build GRCh38
rs980343290 Chr.17: 67072430-67072430 on Build GRCh38
rs996782271 Chr.17: 67072336-67072336 on Build GRCh38
rs1053076420 Chr.17: 67072107-67072107 on Build GRCh38
rs774448750 Chr.17: 67072197-67072197 on Build GRCh38
rs900694983 Chr.17: 67072141-67072141 on Build GRCh38
rs112474087 Chr.17: 67072285-67072285 on Build GRCh38
rs202155947 Chr.17: 67072016-67072016 on Build GRCh38
rs552474214 Chr.17: 67072370-67072370 on Build GRCh38
rs979936784 Chr.17: 67072423-67072423 on Build GRCh38
rs1014574140 Chr.17: 67072122-67072122 on Build GRCh38
rs989000633 Chr.17: 67072446-67072446 on Build GRCh38
rs952456028 Chr.17: 67072196-67072196 on Build GRCh38
rs201824911 Chr.17: 67072349-67072349 on Build GRCh38
rs571046635 Chr.17: 67072403-67072403 on Build GRCh38
rs187340392 Chr.17: 67072259-67072259 on Build GRCh38
rs1015285615 Chr.17: 67072367-67072367 on Build GRCh38
rs150736345 Chr.17: 67072355-67072355 on Build GRCh38
rs531068708 Chr.17: 67072356-67072356 on Build GRCh38
rs535015912 Chr.17: 67072055-67072055 on Build GRCh38
rs917156418 Chr.17: 67072260-67072260 on Build GRCh38
rs554247671 Chr.17: 67072178-67072178 on Build GRCh38
rs780232901 Chr.17: 67072500-67072500 on Build GRCh38
rs530118730 Chr.17: 67072311-67072311 on Build GRCh38
rs200377725 Chr.17: 67072339-67072340 on Build GRCh38
rs970268937 Chr.17: 67072429-67072429 on Build GRCh38
rs917042972 Chr.17: 67072062-67072062 on Build GRCh38
rs939906247 Chr.17: 67072029-67072029 on Build GRCh38
rs970288762 Chr.17: 67072152-67072152 on Build GRCh38
rs182192526 Chr.17: 67072024-67072024 on Build GRCh38
rs373603399 Chr.17: 67072103-67072103 on Build GRCh38
rs980229242 Chr.17: 67072162-67072162 on Build GRCh38
rs755780068 Chr.17: 67072280-67072280 on Build GRCh38
rs926242307 Chr.17: 67072438-67072438 on Build GRCh38
rs758804788 Chr.17: 67072420-67072420 on Build GRCh38
rs1013919957 Chr.17: 67072416-67072416 on Build GRCh38
rs1041048429 Chr.17: 67072322-67072322 on Build GRCh38
rs1009541320 Chr.17: 67072129-67072129 on Build GRCh38
rs983758507 Chr.17: 67072202-67072202 on Build GRCh38
rs759503588 Chr.17: 67072209-67072209 on Build GRCh38
rs1024677773 Chr.17: 67072417-67072417 on Build GRCh38
rs992677009 Chr.17: 67072040-67072040 on Build GRCh38
rs960498746 Chr.17: 67072380-67072380 on Build GRCh38
rs7501436 Chr.17: 67072130-67072130 on Build GRCh38
rs1028066474 Chr.17: 67072164-67072164 on Build GRCh38
rs139893483 Chr.17: 67072101-67072101 on Build GRCh38
rs1013169402 Chr.17: 67072398-67072398 on Build GRCh38
rs948569418 Chr.17: 67072063-67072063 on Build GRCh38
rs892057904 Chr.17: 67072108-67072108 on Build GRCh38
rs996339420 Chr.17: 67072154-67072154 on Build GRCh38
rs1053587776 Chr.17: 67072213-67072213 on Build GRCh38
rs576127164 Chr.17: 67072231-67072231 on Build GRCh38
rs988921226 Chr.17: 67072506-67072506 on Build GRCh38
rs921956783 Chr.17: 67072513-67072513 on Build GRCh38
rs925754770 Chr.17: 67072437-67072437 on Build GRCh38
rs142176925 Chr.17: 67072244-67072244 on Build GRCh38
rs900748615 Chr.17: 67072323-67072323 on Build GRCh38
rs183072439 Chr.17: 67072349-67072349 on Build GRCh38
rs961335449 Chr.17: 67072233-67072233 on Build GRCh38
rs559740825 Chr.17: 67072303-67072303 on Build GRCh38
rs891774865 Chr.17: 67072314-67072314 on Build GRCh38

View Full Product Details