Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67552068-67552602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITPNC1
Entrez Gene ID
26207
Gene Name
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1
UniGene
Hs.591185
Gene Chromosome Location
Chr.17: 67377281-67697263 on Build GRCh38
Transcripts
NM_012417.3 NM_181671.2 XM_005257216.2 XM_017024442.1 XM_017024443.1 XM_017024444.1 XM_017024445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4068889 c.274T>C p.Y92H Chr.17: 67552333-67552333 on Build GRCh38
2797522 c.228C>G p.V76V Chr.17: 67552287-67552287 on Build GRCh38
7341588 c.260delG p.W87fs*1 Chr.17: 67552319-67552319 on Build GRCh38
6271755 c.244G>T p.V82L Chr.17: 67552303-67552303 on Build GRCh38
1194203 c.206C>T p.P69L Chr.17: 67552265-67552265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762191575 Chr.17: 67552271-67552271 on Build GRCh38
rs766277799 Chr.17: 67552360-67552360 on Build GRCh38
rs146525851 Chr.17: 67552402-67552402 on Build GRCh38
rs1011156965 Chr.17: 67552455-67552455 on Build GRCh38
rs933584183 Chr.17: 67552101-67552101 on Build GRCh38
rs749882106 Chr.17: 67552230-67552230 on Build GRCh38
rs919305945 Chr.17: 67552149-67552149 on Build GRCh38
rs1047310922 Chr.17: 67552463-67552463 on Build GRCh38
rs924909203 Chr.17: 67552198-67552198 on Build GRCh38
rs200978982 Chr.17: 67552480-67552480 on Build GRCh38
rs941984108 Chr.17: 67552131-67552131 on Build GRCh38
rs774906366 Chr.17: 67552241-67552241 on Build GRCh38
rs764011538 Chr.17: 67552268-67552268 on Build GRCh38
rs910544366 Chr.17: 67552118-67552118 on Build GRCh38
rs887036455 Chr.17: 67552468-67552468 on Build GRCh38
rs1013320184 Chr.17: 67552545-67552549 on Build GRCh38
rs923550673 Chr.17: 67552096-67552096 on Build GRCh38
rs958729805 Chr.17: 67552537-67552538 on Build GRCh38
rs2570044 Chr.17: 67552567-67552567 on Build GRCh38
rs751391832 Chr.17: 67552363-67552363 on Build GRCh38
rs556072325 Chr.17: 67552502-67552502 on Build GRCh38
rs754725967 Chr.17: 67552375-67552375 on Build GRCh38
rs537606672 Chr.17: 67552483-67552483 on Build GRCh38
rs538955420 Chr.17: 67552240-67552240 on Build GRCh38
rs894163160 Chr.17: 67552452-67552452 on Build GRCh38
rs769131622 Chr.17: 67552212-67552212 on Build GRCh38
rs780917661 Chr.17: 67552391-67552391 on Build GRCh38
rs773777003 Chr.17: 67552229-67552229 on Build GRCh38
rs986560249 Chr.17: 67552107-67552107 on Build GRCh38
rs1039488139 Chr.17: 67552439-67552439 on Build GRCh38
rs551404991 Chr.17: 67552250-67552250 on Build GRCh38
rs2916133 Chr.17: 67552424-67552424 on Build GRCh38
rs767885662 Chr.17: 67552247-67552247 on Build GRCh38
rs1002752783 Chr.17: 67552479-67552479 on Build GRCh38
rs371764256 Chr.17: 67552395-67552395 on Build GRCh38
rs967090449 Chr.17: 67552478-67552478 on Build GRCh38
rs977977473 Chr.17: 67552197-67552197 on Build GRCh38
rs111501038 Chr.17: 67552254-67552254 on Build GRCh38
rs375389796 Chr.17: 67552143-67552143 on Build GRCh38
rs759949321 Chr.17: 67552245-67552245 on Build GRCh38
rs753947061 Chr.17: 67552269-67552269 on Build GRCh38
rs369774854 Chr.17: 67552260-67552260 on Build GRCh38
rs1030479138 Chr.17: 67552539-67552539 on Build GRCh38
rs572329156 Chr.17: 67552467-67552468 on Build GRCh38
rs1021337029 Chr.17: 67552456-67552456 on Build GRCh38
rs968942893 Chr.17: 67552122-67552122 on Build GRCh38
rs999466452 Chr.17: 67552527-67552527 on Build GRCh38
rs769258603 Chr.17: 67552112-67552112 on Build GRCh38
rs533656475 Chr.17: 67552325-67552325 on Build GRCh38

View Full Product Details