Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69520859-69521364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K6
Entrez Gene ID
5608
Gene Name
mitogen-activated protein kinase kinase 6
Gene Aliases
MAPKK6, MEK6, MKK6, PRKMK6, SAPKK-3, SAPKK3
UniGene
Hs.463978
Gene Chromosome Location
Chr.17: 69414697-69553854 on Build GRCh38
Transcripts
NM_002758.3 XM_005257515.2 XM_005257516.2 XM_006721975.3 XM_011525025.2 XM_011525026.2 XM_011525027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4580029 c.516G>A p.K172K Chr.17: 69521081-69521081 on Build GRCh38
6399911 c.531C>A p.H177Q Chr.17: 69521096-69521096 on Build GRCh38
1710709 c.521C>T p.S174F Chr.17: 69521086-69521086 on Build GRCh38
71554 c.488T>G p.V163G Chr.17: 69521053-69521053 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916579453 Chr.17: 69521244-69521244 on Build GRCh38
rs140864293 Chr.17: 69521256-69521256 on Build GRCh38
rs1043924879 Chr.17: 69520989-69520989 on Build GRCh38
rs765320378 Chr.17: 69521074-69521074 on Build GRCh38
rs550608688 Chr.17: 69521188-69521188 on Build GRCh38
rs942463643 Chr.17: 69521328-69521328 on Build GRCh38
rs768157576 Chr.17: 69521127-69521127 on Build GRCh38
rs369122561 Chr.17: 69521300-69521300 on Build GRCh38
rs541990700 Chr.17: 69521213-69521214 on Build GRCh38
rs762012848 Chr.17: 69521153-69521153 on Build GRCh38
rs555528866 Chr.17: 69520903-69520903 on Build GRCh38
rs777491735 Chr.17: 69521012-69521012 on Build GRCh38
rs763154937 Chr.17: 69521096-69521096 on Build GRCh38
rs182968958 Chr.17: 69520984-69520984 on Build GRCh38
rs954158692 Chr.17: 69521242-69521242 on Build GRCh38
rs375928354 Chr.17: 69521111-69521111 on Build GRCh38
rs17690945 Chr.17: 69521284-69521284 on Build GRCh38
rs562767129 Chr.17: 69521192-69521192 on Build GRCh38
rs761904827 Chr.17: 69521049-69521049 on Build GRCh38
rs762448110 Chr.17: 69521203-69521203 on Build GRCh38
rs146455212 Chr.17: 69520968-69520968 on Build GRCh38
rs533636025 Chr.17: 69521285-69521285 on Build GRCh38
rs565511915 Chr.17: 69520901-69520901 on Build GRCh38
rs763784943 Chr.17: 69521150-69521151 on Build GRCh38
rs540472624 Chr.17: 69521165-69521165 on Build GRCh38
rs41301823 Chr.17: 69521181-69521181 on Build GRCh38
rs1046151305 Chr.17: 69521115-69521115 on Build GRCh38
rs1034458537 Chr.17: 69521177-69521177 on Build GRCh38
rs572550194 Chr.17: 69521027-69521027 on Build GRCh38
rs772037812 Chr.17: 69521032-69521032 on Build GRCh38
rs760076232 Chr.17: 69521112-69521112 on Build GRCh38
rs1051729623 Chr.17: 69521291-69521291 on Build GRCh38
rs770753196 Chr.17: 69521023-69521023 on Build GRCh38
rs577968620 Chr.17: 69520982-69520982 on Build GRCh38
rs1039427806 Chr.17: 69521329-69521329 on Build GRCh38
rs529036757 Chr.17: 69521185-69521185 on Build GRCh38
rs1030428346 Chr.17: 69521241-69521241 on Build GRCh38
rs924234833 Chr.17: 69521275-69521275 on Build GRCh38
rs752194353 Chr.17: 69521164-69521164 on Build GRCh38
rs773490930 Chr.17: 69521081-69521081 on Build GRCh38
rs538155497 Chr.17: 69520926-69520926 on Build GRCh38
rs906241550 Chr.17: 69521118-69521118 on Build GRCh38
rs766755714 Chr.17: 69521106-69521106 on Build GRCh38
rs61758052 Chr.17: 69521339-69521339 on Build GRCh38
rs910375372 Chr.17: 69521262-69521262 on Build GRCh38
rs869301249 Chr.17: 69521180-69521180 on Build GRCh38
rs188720020 Chr.17: 69521227-69521227 on Build GRCh38
rs548042421 Chr.17: 69521010-69521010 on Build GRCh38
rs776064610 Chr.17: 69521099-69521099 on Build GRCh38
rs748966336 Chr.17: 69521022-69521022 on Build GRCh38
rs941909014 Chr.17: 69521054-69521054 on Build GRCh38
rs776150672 Chr.17: 69521299-69521299 on Build GRCh38
rs919130752 Chr.17: 69521325-69521325 on Build GRCh38
rs1051269639 Chr.17: 69521154-69521154 on Build GRCh38
rs776652694 Chr.17: 69521040-69521040 on Build GRCh38
rs574038760 Chr.17: 69520907-69520907 on Build GRCh38
rs910490285 Chr.17: 69521014-69521014 on Build GRCh38

View Full Product Details