Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70133855-70134355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ16
Entrez Gene ID
3773
Gene Name
potassium voltage-gated channel subfamily J member 16
Gene Aliases
BIR9, KIR5.1
UniGene
Hs.463985
Gene Chromosome Location
Chr.17: 70075225-70135608 on Build GRCh38
Transcripts
NM_001270422.1 NM_001291622.1 NM_001291623.1 NM_001291624.1 NM_001291625.1 NM_018658.2 NM_170741.2 NM_170742.2 XM_005257337.4 XM_006721885.3 XM_006721886.3 XM_006721887.3 XM_011524781.2 XM_017024609.1 XM_017024610.1 XM_017024611.1 XM_017024612.1 XM_017024613.1

SNP Detail

SNP ID Chromosome Location
rs1025504774 Chr.17: 70134237-70134237 on Build GRCh38
rs143099409 Chr.17: 70134176-70134176 on Build GRCh38
rs1032595254 Chr.17: 70134258-70134258 on Build GRCh38
rs538718053 Chr.17: 70134179-70134179 on Build GRCh38
rs981660808 Chr.17: 70133892-70133892 on Build GRCh38
rs935645399 Chr.17: 70134069-70134069 on Build GRCh38
rs780650063 Chr.17: 70134230-70134230 on Build GRCh38
rs751599481 Chr.17: 70134093-70134093 on Build GRCh38
rs758571879 Chr.17: 70133951-70133951 on Build GRCh38
rs1013624009 Chr.17: 70134166-70134166 on Build GRCh38
rs145531616 Chr.17: 70133944-70133944 on Build GRCh38
rs991679554 Chr.17: 70133902-70133902 on Build GRCh38
rs1036316463 Chr.17: 70134149-70134149 on Build GRCh38
rs887733037 Chr.17: 70134205-70134205 on Build GRCh38
rs879589147 Chr.17: 70134302-70134302 on Build GRCh38
rs1052077941 Chr.17: 70134046-70134046 on Build GRCh38
rs370231007 Chr.17: 70134130-70134130 on Build GRCh38
rs866514545 Chr.17: 70134306-70134306 on Build GRCh38
rs1040169406 Chr.17: 70133878-70133878 on Build GRCh38
rs996083093 Chr.17: 70133891-70133891 on Build GRCh38
rs977453964 Chr.17: 70134187-70134187 on Build GRCh38
rs1029347741 Chr.17: 70133893-70133893 on Build GRCh38
rs77037824 Chr.17: 70134094-70134094 on Build GRCh38
rs368828946 Chr.17: 70133913-70133913 on Build GRCh38
rs533854701 Chr.17: 70134257-70134257 on Build GRCh38
rs969979640 Chr.17: 70133995-70133995 on Build GRCh38
rs749060546 Chr.17: 70134312-70134312 on Build GRCh38
rs765317058 Chr.17: 70133879-70133879 on Build GRCh38
rs945746602 Chr.17: 70134128-70134128 on Build GRCh38
rs116228233 Chr.17: 70134304-70134304 on Build GRCh38
rs149075675 Chr.17: 70134142-70134142 on Build GRCh38
rs958977610 Chr.17: 70134162-70134162 on Build GRCh38
rs959085482 Chr.17: 70133901-70133901 on Build GRCh38
rs766250741 Chr.17: 70134311-70134311 on Build GRCh38
rs1003737801 Chr.17: 70134295-70134295 on Build GRCh38
rs933342316 Chr.17: 70134019-70134019 on Build GRCh38
rs912874995 Chr.17: 70134282-70134282 on Build GRCh38
rs920395437 Chr.17: 70134260-70134260 on Build GRCh38
rs924701332 Chr.17: 70134206-70134206 on Build GRCh38
rs142712667 Chr.17: 70134273-70134273 on Build GRCh38
rs992210211 Chr.17: 70133952-70133952 on Build GRCh38
rs910478233 Chr.17: 70134047-70134047 on Build GRCh38
rs1006210649 Chr.17: 70133931-70133931 on Build GRCh38
rs531968867 Chr.17: 70134102-70134102 on Build GRCh38
rs184101075 Chr.17: 70134293-70134293 on Build GRCh38
rs1025111871 Chr.17: 70133978-70133978 on Build GRCh38
rs9916760 Chr.17: 70134218-70134218 on Build GRCh38
rs573579581 Chr.17: 70134286-70134286 on Build GRCh38
rs1006096304 Chr.17: 70134212-70134212 on Build GRCh38
rs749751870 Chr.17: 70134303-70134303 on Build GRCh38
rs967057271 Chr.17: 70134300-70134300 on Build GRCh38
rs757475428 Chr.17: 70134135-70134135 on Build GRCh38
rs925555418 Chr.17: 70134035-70134035 on Build GRCh38
rs1047471634 Chr.17: 70134285-70134285 on Build GRCh38
rs148218624 Chr.17: 70134213-70134213 on Build GRCh38
rs993235923 Chr.17: 70134126-70134126 on Build GRCh38
rs1050355380 Chr.17: 70134200-70134200 on Build GRCh38
rs988402200 Chr.17: 70134281-70134281 on Build GRCh38
rs764068936 Chr.17: 70134003-70134003 on Build GRCh38
rs931836001 Chr.17: 70134181-70134181 on Build GRCh38
rs573775023 Chr.17: 70134177-70134177 on Build GRCh38
rs969571115 Chr.17: 70134244-70134244 on Build GRCh38
rs912907044 Chr.17: 70134106-70134106 on Build GRCh38
rs906286565 Chr.17: 70134291-70134291 on Build GRCh38
rs1035962160 Chr.17: 70133900-70133900 on Build GRCh38

View Full Product Details