Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 72849550-72850060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A11
Entrez Gene ID
201266
Gene Name
solute carrier family 39 member 11
Gene Aliases
C17orf26, ZIP11
UniGene
Hs.221127
Gene Chromosome Location
Chr.17: 72645946-73092714 on Build GRCh38
Transcripts
NM_001159770.1 NM_139177.3 XM_005257134.3 XM_006721754.3 XM_006721755.2 XM_006721756.2 XM_006721757.2 XM_006721758.3 XM_006721759.1 XM_011524493.1 XM_011524494.2 XM_011524495.2 XM_011524496.2 XM_011524498.2 XM_017024330.1 XM_017024331.1 XM_017024332.1 XM_017024333.1 XM_017024334.1 XM_017024335.1 XM_017024336.1 XM_017024337.1 XM_017024338.1 XM_017024339.1 XM_017024340.1 XM_017024341.1 XM_017024342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5055733 c.578C>T p.A193V Chr.17: 72849657-72849657 on Build GRCh38
5766875 c.441G>A p.E147E Chr.17: 72849794-72849794 on Build GRCh38
6428369 c.599C>A p.P200Q Chr.17: 72849636-72849636 on Build GRCh38
3521549 c.517C>T p.R173* Chr.17: 72849718-72849718 on Build GRCh38
2157714 c.474_475insC p.A159fs*63 Chr.17: 72849760-72849761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773801459 Chr.17: 72849810-72849811 on Build GRCh38
rs757727469 Chr.17: 72849811-72849814 on Build GRCh38
rs1056378693 Chr.17: 72849586-72849586 on Build GRCh38
rs753167027 Chr.17: 72849780-72849780 on Build GRCh38
rs866679630 Chr.17: 72849695-72849695 on Build GRCh38
rs779903144 Chr.17: 72849845-72849845 on Build GRCh38
rs758085667 Chr.17: 72849833-72849833 on Build GRCh38
rs771897276 Chr.17: 72849641-72849641 on Build GRCh38
rs1017073908 Chr.17: 72849863-72849863 on Build GRCh38
rs780316851 Chr.17: 72849821-72849826 on Build GRCh38
rs141628970 Chr.17: 72849697-72849697 on Build GRCh38
rs745553592 Chr.17: 72849817-72849817 on Build GRCh38
rs962304074 Chr.17: 72849673-72849673 on Build GRCh38
rs755451091 Chr.17: 72849611-72849611 on Build GRCh38
rs922567855 Chr.17: 72850012-72850012 on Build GRCh38
rs202177814 Chr.17: 72849824-72849824 on Build GRCh38
rs766704695 Chr.17: 72849665-72849665 on Build GRCh38
rs377648414 Chr.17: 72849854-72849854 on Build GRCh38
rs369301863 Chr.17: 72849791-72849791 on Build GRCh38
rs752034112 Chr.17: 72849606-72849606 on Build GRCh38
rs756830187 Chr.17: 72849794-72849794 on Build GRCh38
rs915334342 Chr.17: 72849984-72849984 on Build GRCh38
rs893758686 Chr.17: 72849582-72849582 on Build GRCh38
rs774632972 Chr.17: 72849731-72849731 on Build GRCh38
rs545661942 Chr.17: 72849694-72849694 on Build GRCh38
rs141064511 Chr.17: 72849613-72849613 on Build GRCh38
rs766503635 Chr.17: 72849763-72849763 on Build GRCh38
rs34998299 Chr.17: 72849811-72849812 on Build GRCh38
rs113128349 Chr.17: 72849604-72849604 on Build GRCh38
rs758786197 Chr.17: 72849598-72849598 on Build GRCh38
rs759105797 Chr.17: 72849809-72849810 on Build GRCh38
rs753335316 Chr.17: 72849793-72849793 on Build GRCh38
rs74444525 Chr.17: 72849680-72849680 on Build GRCh38
rs771950666 Chr.17: 72849714-72849714 on Build GRCh38
rs765429147 Chr.17: 72849649-72849649 on Build GRCh38
rs945345892 Chr.17: 72849701-72849701 on Build GRCh38
rs745762301 Chr.17: 72849640-72849640 on Build GRCh38
rs374851095 Chr.17: 72849764-72849764 on Build GRCh38
rs540411380 Chr.17: 72849717-72849717 on Build GRCh38
rs373163649 Chr.17: 72849693-72849693 on Build GRCh38
rs761961296 Chr.17: 72849646-72849646 on Build GRCh38
rs879183033 Chr.17: 72849810-72849810 on Build GRCh38
rs959498350 Chr.17: 72849947-72849947 on Build GRCh38
rs747143797 Chr.17: 72849824-72849825 on Build GRCh38
rs561779094 Chr.17: 72849724-72849724 on Build GRCh38
rs773435044 Chr.17: 72849752-72849752 on Build GRCh38
rs145922024 Chr.17: 72849698-72849698 on Build GRCh38
rs970953677 Chr.17: 72849678-72849678 on Build GRCh38
rs751853412 Chr.17: 72849668-72849668 on Build GRCh38
rs763540337 Chr.17: 72849788-72849788 on Build GRCh38
rs780593694 Chr.17: 72849603-72849603 on Build GRCh38
rs765918599 Chr.17: 72849796-72849797 on Build GRCh38
rs4436821 Chr.17: 72849977-72849977 on Build GRCh38
rs1009042465 Chr.17: 72849650-72849650 on Build GRCh38
rs945473265 Chr.17: 72849986-72849986 on Build GRCh38
rs796160881 Chr.17: 72849824-72849825 on Build GRCh38
rs528825217 Chr.17: 72850014-72850014 on Build GRCh38
rs769924632 Chr.17: 72849743-72849743 on Build GRCh38
rs952062875 Chr.17: 72849936-72849936 on Build GRCh38
rs768700741 Chr.17: 72849856-72849856 on Build GRCh38
rs745351473 Chr.17: 72849590-72849590 on Build GRCh38
rs753299534 Chr.17: 72849687-72849687 on Build GRCh38
rs1042822085 Chr.17: 72850007-72850007 on Build GRCh38
rs755439919 Chr.17: 72849674-72849674 on Build GRCh38
rs750722448 Chr.17: 72849597-72849597 on Build GRCh38
rs562433242 Chr.17: 72849869-72849869 on Build GRCh38
rs763180972 Chr.17: 72849760-72849760 on Build GRCh38
rs763268192 Chr.17: 72849652-72849652 on Build GRCh38
rs377363641 Chr.17: 72849619-72849619 on Build GRCh38
rs369510171 Chr.17: 72849623-72849623 on Build GRCh38
rs369931536 Chr.17: 72849588-72849588 on Build GRCh38
rs1042373569 Chr.17: 72849727-72849727 on Build GRCh38
rs926441615 Chr.17: 72849574-72849574 on Build GRCh38
rs748033585 Chr.17: 72849825-72849828 on Build GRCh38
rs139816411 Chr.17: 72849707-72849707 on Build GRCh38
rs764746579 Chr.17: 72849811-72849813 on Build GRCh38
rs146605355 Chr.17: 72849579-72849579 on Build GRCh38
rs796898025 Chr.17: 72849810-72849811 on Build GRCh38
rs1027614641 Chr.17: 72849683-72849683 on Build GRCh38
rs867450002 Chr.17: 72849583-72849583 on Build GRCh38
rs1009012726 Chr.17: 72849871-72849871 on Build GRCh38
rs144832523 Chr.17: 72849651-72849651 on Build GRCh38
rs778686610 Chr.17: 72849705-72849705 on Build GRCh38
rs762455692 Chr.17: 72849695-72849699 on Build GRCh38
rs774022089 Chr.17: 72849637-72849637 on Build GRCh38
rs992247056 Chr.17: 72850005-72850005 on Build GRCh38
rs765524019 Chr.17: 72849593-72849593 on Build GRCh38
rs146219801 Chr.17: 72849782-72849782 on Build GRCh38
rs4998297 Chr.17: 72849888-72849888 on Build GRCh38
rs1023963709 Chr.17: 72849889-72849889 on Build GRCh38
rs1033758535 Chr.17: 72849974-72849974 on Build GRCh38
rs777386726 Chr.17: 72849620-72849620 on Build GRCh38
rs143231715 Chr.17: 72849765-72849765 on Build GRCh38
rs747610696 Chr.17: 72849614-72849614 on Build GRCh38
rs778405587 Chr.17: 72849800-72849800 on Build GRCh38
rs961137544 Chr.17: 72849867-72849867 on Build GRCh38
rs527641599 Chr.17: 72849919-72849919 on Build GRCh38
rs770674217 Chr.17: 72849628-72849628 on Build GRCh38
rs575276322 Chr.17: 72849939-72849939 on Build GRCh38
rs765467213 Chr.17: 72849811-72849811 on Build GRCh38
rs796269291 Chr.17: 72849804-72849804 on Build GRCh38
rs926725307 Chr.17: 72849730-72849730 on Build GRCh38
rs552858374 Chr.17: 72849893-72849893 on Build GRCh38
rs760638934 Chr.17: 72850025-72850025 on Build GRCh38
rs753879340 Chr.17: 72849584-72849584 on Build GRCh38

View Full Product Details