Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74250187-74250692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
UniGene
Hs.27935
Gene Chromosome Location
Chr.17: 74213557-74262020 on Build GRCh38
Transcripts
NM_032646.5 NM_052869.1 XM_005257824.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385731 c.1051C>T p.L351L Chr.17: 74250292-74250292 on Build GRCh38
6340713 c.1087G>T p.A363S Chr.17: 74250328-74250328 on Build GRCh38
80707 c.1077C>T p.H359H Chr.17: 74250318-74250318 on Build GRCh38
7095279 c.1053G>A p.L351L Chr.17: 74250294-74250294 on Build GRCh38
3521693 c.1047C>T p.L349L Chr.17: 74250288-74250288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190757424 Chr.17: 74250449-74250449 on Build GRCh38
rs145583965 Chr.17: 74250408-74250408 on Build GRCh38
rs376529142 Chr.17: 74250237-74250237 on Build GRCh38
rs374479940 Chr.17: 74250332-74250332 on Build GRCh38
rs145865780 Chr.17: 74250609-74250609 on Build GRCh38
rs187140941 Chr.17: 74250429-74250429 on Build GRCh38
rs541457097 Chr.17: 74250464-74250464 on Build GRCh38
rs770811667 Chr.17: 74250243-74250244 on Build GRCh38
rs1012376367 Chr.17: 74250545-74250545 on Build GRCh38
rs61066438 Chr.17: 74250613-74250613 on Build GRCh38
rs547977014 Chr.17: 74250212-74250212 on Build GRCh38
rs577666880 Chr.17: 74250366-74250366 on Build GRCh38
rs756388556 Chr.17: 74250539-74250539 on Build GRCh38
rs751274095 Chr.17: 74250245-74250248 on Build GRCh38
rs766624555 Chr.17: 74250544-74250544 on Build GRCh38
rs779111791 Chr.17: 74250591-74250591 on Build GRCh38
rs756796469 Chr.17: 74250257-74250257 on Build GRCh38
rs751464110 Chr.17: 74250240-74250241 on Build GRCh38
rs552739368 Chr.17: 74250531-74250531 on Build GRCh38
rs139685298 Chr.17: 74250320-74250320 on Build GRCh38
rs7225106 Chr.17: 74250589-74250589 on Build GRCh38
rs774690992 Chr.17: 74250330-74250330 on Build GRCh38
rs7225236 Chr.17: 74250668-74250668 on Build GRCh38
rs746998838 Chr.17: 74250310-74250310 on Build GRCh38
rs777539141 Chr.17: 74250364-74250374 on Build GRCh38
rs761211360 Chr.17: 74250360-74250360 on Build GRCh38
rs557116023 Chr.17: 74250470-74250470 on Build GRCh38
rs146342198 Chr.17: 74250313-74250313 on Build GRCh38
rs958172858 Chr.17: 74250430-74250430 on Build GRCh38
rs1047557621 Chr.17: 74250288-74250288 on Build GRCh38
rs781108211 Chr.17: 74250240-74250247 on Build GRCh38
rs746839705 Chr.17: 74250398-74250398 on Build GRCh38
rs10606052 Chr.17: 74250247-74250250 on Build GRCh38
rs1056124340 Chr.17: 74250462-74250462 on Build GRCh38
rs942769831 Chr.17: 74250446-74250446 on Build GRCh38
rs753231519 Chr.17: 74250380-74250380 on Build GRCh38
rs139483749 Chr.17: 74250244-74250247 on Build GRCh38
rs369690684 Chr.17: 74250232-74250232 on Build GRCh38
rs71286189 Chr.17: 74250659-74250668 on Build GRCh38
rs533602384 Chr.17: 74250469-74250469 on Build GRCh38
rs750120873 Chr.17: 74250388-74250388 on Build GRCh38
rs754734664 Chr.17: 74250239-74250240 on Build GRCh38
rs146608677 Chr.17: 74250344-74250344 on Build GRCh38
rs779777186 Chr.17: 74250397-74250397 on Build GRCh38
rs770520453 Chr.17: 74250398-74250399 on Build GRCh38
rs1039901291 Chr.17: 74250472-74250472 on Build GRCh38
rs770999919 Chr.17: 74250328-74250328 on Build GRCh38
rs536203519 Chr.17: 74250439-74250439 on Build GRCh38
rs377277767 Chr.17: 74250611-74250611 on Build GRCh38
rs748841859 Chr.17: 74250396-74250398 on Build GRCh38
rs766334773 Chr.17: 74250240-74250243 on Build GRCh38
rs990775222 Chr.17: 74250413-74250413 on Build GRCh38
rs764690950 Chr.17: 74250373-74250373 on Build GRCh38
rs923662500 Chr.17: 74250641-74250641 on Build GRCh38
rs959092697 Chr.17: 74250370-74250370 on Build GRCh38
rs763038400 Chr.17: 74250234-74250234 on Build GRCh38
rs911332636 Chr.17: 74250440-74250440 on Build GRCh38
rs144932863 Chr.17: 74250509-74250509 on Build GRCh38
rs978711563 Chr.17: 74250421-74250421 on Build GRCh38
rs539834937 Chr.17: 74250381-74250381 on Build GRCh38
rs758168455 Chr.17: 74250390-74250390 on Build GRCh38
rs7225791 Chr.17: 74250659-74250659 on Build GRCh38
rs769901818 Chr.17: 74250223-74250223 on Build GRCh38
rs906169288 Chr.17: 74250572-74250572 on Build GRCh38
rs758260713 Chr.17: 74250280-74250280 on Build GRCh38
rs745652917 Chr.17: 74250260-74250260 on Build GRCh38
rs922639130 Chr.17: 74250667-74250667 on Build GRCh38
rs924079446 Chr.17: 74250213-74250213 on Build GRCh38
rs779936673 Chr.17: 74250289-74250289 on Build GRCh38
rs7224615 Chr.17: 74250327-74250327 on Build GRCh38
rs964161476 Chr.17: 74250654-74250654 on Build GRCh38
rs762287571 Chr.17: 74250468-74250468 on Build GRCh38
rs370613156 Chr.17: 74250258-74250258 on Build GRCh38
rs112542987 Chr.17: 74250497-74250497 on Build GRCh38
rs997806045 Chr.17: 74250276-74250276 on Build GRCh38
rs768029128 Chr.17: 74250340-74250340 on Build GRCh38
rs373524645 Chr.17: 74250236-74250236 on Build GRCh38
rs971384616 Chr.17: 74250211-74250211 on Build GRCh38
rs764796381 Chr.17: 74250383-74250383 on Build GRCh38
rs73996939 Chr.17: 74250247-74250247 on Build GRCh38
rs147304485 Chr.17: 74250448-74250448 on Build GRCh38
rs759974151 Chr.17: 74250335-74250335 on Build GRCh38
rs769768447 Chr.17: 74250543-74250543 on Build GRCh38
rs1023533920 Chr.17: 74250590-74250590 on Build GRCh38
rs1037536882 Chr.17: 74250512-74250512 on Build GRCh38
rs922553715 Chr.17: 74250457-74250457 on Build GRCh38
rs965307676 Chr.17: 74250343-74250343 on Build GRCh38
rs111326582 Chr.17: 74250246-74250247 on Build GRCh38
rs951515995 Chr.17: 74250368-74250368 on Build GRCh38
rs7223962 Chr.17: 74250570-74250570 on Build GRCh38
rs755651777 Chr.17: 74250246-74250246 on Build GRCh38
rs1054354023 Chr.17: 74250463-74250463 on Build GRCh38

View Full Product Details