Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75318406-75318912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRB2
Entrez Gene ID
2885
Gene Name
growth factor receptor bound protein 2
Gene Aliases
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2
UniGene
Hs.444356
Gene Chromosome Location
Chr.17: 75318076-75405709 on Build GRCh38
Transcripts
NM_002086.4 NM_203506.2

SNP Detail

SNP ID Chromosome Location
rs944914638 Chr.17: 75318621-75318621 on Build GRCh38
rs960656505 Chr.17: 75318831-75318831 on Build GRCh38
rs750418078 Chr.17: 75318665-75318665 on Build GRCh38
rs941040148 Chr.17: 75318537-75318537 on Build GRCh38
rs779814410 Chr.17: 75318586-75318587 on Build GRCh38
rs534312310 Chr.17: 75318484-75318484 on Build GRCh38
rs936214820 Chr.17: 75318598-75318598 on Build GRCh38
rs962124073 Chr.17: 75318462-75318462 on Build GRCh38
rs933687460 Chr.17: 75318503-75318506 on Build GRCh38
rs185635804 Chr.17: 75318767-75318767 on Build GRCh38
rs189362846 Chr.17: 75318832-75318832 on Build GRCh38
rs1003555810 Chr.17: 75318766-75318766 on Build GRCh38
rs966376212 Chr.17: 75318866-75318866 on Build GRCh38
rs751214712 Chr.17: 75318513-75318513 on Build GRCh38
rs1020589041 Chr.17: 75318847-75318847 on Build GRCh38
rs993484842 Chr.17: 75318664-75318664 on Build GRCh38
rs774807830 Chr.17: 75318471-75318471 on Build GRCh38
rs889385010 Chr.17: 75318608-75318608 on Build GRCh38
rs887858390 Chr.17: 75318736-75318736 on Build GRCh38
rs1033641160 Chr.17: 75318505-75318505 on Build GRCh38
rs979121315 Chr.17: 75318575-75318575 on Build GRCh38
rs767390659 Chr.17: 75318599-75318599 on Build GRCh38
rs35280208 Chr.17: 75318881-75318881 on Build GRCh38
rs922344960 Chr.17: 75318490-75318490 on Build GRCh38
rs897801632 Chr.17: 75318657-75318657 on Build GRCh38
rs141324646 Chr.17: 75318573-75318573 on Build GRCh38
rs182246061 Chr.17: 75318472-75318472 on Build GRCh38
rs1045502967 Chr.17: 75318752-75318752 on Build GRCh38
rs546117270 Chr.17: 75318473-75318473 on Build GRCh38
rs530036763 Chr.17: 75318833-75318833 on Build GRCh38
rs945145379 Chr.17: 75318547-75318547 on Build GRCh38
rs992459731 Chr.17: 75318839-75318839 on Build GRCh38
rs1033896691 Chr.17: 75318795-75318795 on Build GRCh38
rs553836699 Chr.17: 75318596-75318596 on Build GRCh38
rs557179700 Chr.17: 75318441-75318441 on Build GRCh38
rs550044557 Chr.17: 75318853-75318853 on Build GRCh38
rs1040394872 Chr.17: 75318656-75318656 on Build GRCh38
rs3211129 Chr.17: 75318525-75318525 on Build GRCh38
rs180961849 Chr.17: 75318882-75318882 on Build GRCh38
rs913528599 Chr.17: 75318522-75318522 on Build GRCh38
rs553259050 Chr.17: 75318483-75318483 on Build GRCh38
rs907054132 Chr.17: 75318759-75318759 on Build GRCh38
rs367852151 Chr.17: 75318516-75318516 on Build GRCh38
rs764755902 Chr.17: 75318443-75318443 on Build GRCh38

View Full Product Details