Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75829891-75830379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC13D
Entrez Gene ID
201294
Gene Name
unc-13 homolog D
Gene Aliases
FHL3, HLH3, HPLH3, Munc13-4
UniGene
Hs.41045
Gene Chromosome Location
Chr.17: 75827225-75844717 on Build GRCh38
Transcripts
NM_199242.2 XM_011524504.1 XM_011524506.1 XM_011524507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243118 c.2830+6T>G p.? Chr.17: 75830356-75830356 on Build GRCh38
6213344 c.2891C>T p.A964V Chr.17: 75830091-75830091 on Build GRCh38
6352156 c.2943G>A p.E981E Chr.17: 75830039-75830039 on Build GRCh38
1610782 c.2863G>T p.E955* Chr.17: 75830119-75830119 on Build GRCh38
298536 c.2905_2906insC p.Q969fs*12 Chr.17: 75830076-75830077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563286724 Chr.17: 75830178-75830178 on Build GRCh38
rs750433063 Chr.17: 75830348-75830348 on Build GRCh38
rs373259998 Chr.17: 75830323-75830323 on Build GRCh38
rs1005334519 Chr.17: 75830164-75830164 on Build GRCh38
rs569980766 Chr.17: 75830090-75830090 on Build GRCh38
rs751314219 Chr.17: 75830115-75830115 on Build GRCh38
rs756015508 Chr.17: 75830012-75830012 on Build GRCh38
rs964810880 Chr.17: 75829982-75829982 on Build GRCh38
rs758575692 Chr.17: 75830350-75830350 on Build GRCh38
rs950614253 Chr.17: 75830277-75830277 on Build GRCh38
rs910703782 Chr.17: 75830301-75830301 on Build GRCh38
rs766306470 Chr.17: 75830092-75830092 on Build GRCh38
rs924698827 Chr.17: 75830025-75830025 on Build GRCh38
rs752611933 Chr.17: 75829992-75829993 on Build GRCh38
rs538544253 Chr.17: 75830011-75830011 on Build GRCh38
rs775943310 Chr.17: 75830029-75830029 on Build GRCh38
rs957679067 Chr.17: 75830287-75830287 on Build GRCh38
rs770967156 Chr.17: 75829981-75829981 on Build GRCh38
rs779204571 Chr.17: 75830335-75830335 on Build GRCh38
rs761351250 Chr.17: 75830085-75830085 on Build GRCh38
rs958549241 Chr.17: 75830175-75830175 on Build GRCh38
rs1030829119 Chr.17: 75830252-75830252 on Build GRCh38
rs373385387 Chr.17: 75830174-75830174 on Build GRCh38
rs917235131 Chr.17: 75829947-75829947 on Build GRCh38
rs114246714 Chr.17: 75830319-75830319 on Build GRCh38
rs548771105 Chr.17: 75830253-75830253 on Build GRCh38
rs776470044 Chr.17: 75830066-75830066 on Build GRCh38
rs772473891 Chr.17: 75830015-75830015 on Build GRCh38
rs777727127 Chr.17: 75830063-75830065 on Build GRCh38
rs377281943 Chr.17: 75830088-75830088 on Build GRCh38
rs1011813695 Chr.17: 75830215-75830215 on Build GRCh38
rs774891090 Chr.17: 75829984-75829984 on Build GRCh38
rs185682655 Chr.17: 75830144-75830144 on Build GRCh38
rs772529830 Chr.17: 75830202-75830202 on Build GRCh38
rs923676310 Chr.17: 75830357-75830357 on Build GRCh38
rs567552311 Chr.17: 75830165-75830165 on Build GRCh38
rs141576434 Chr.17: 75830324-75830324 on Build GRCh38
rs990279155 Chr.17: 75830298-75830298 on Build GRCh38
rs144744401 Chr.17: 75830089-75830089 on Build GRCh38
rs760819177 Chr.17: 75829954-75829954 on Build GRCh38
rs576805665 Chr.17: 75830204-75830204 on Build GRCh38
rs574589859 Chr.17: 75830123-75830123 on Build GRCh38
rs1031907718 Chr.17: 75829930-75829930 on Build GRCh38
rs149809768 Chr.17: 75830171-75830171 on Build GRCh38
rs1011594442 Chr.17: 75829971-75829971 on Build GRCh38
rs193012444 Chr.17: 75829974-75829974 on Build GRCh38
rs757354369 Chr.17: 75830140-75830140 on Build GRCh38
rs757367802 Chr.17: 75830333-75830333 on Build GRCh38
rs537006659 Chr.17: 75829929-75829929 on Build GRCh38
rs777951171 Chr.17: 75830120-75830120 on Build GRCh38
rs528665939 Chr.17: 75830229-75830229 on Build GRCh38
rs373478612 Chr.17: 75830065-75830065 on Build GRCh38
rs754791533 Chr.17: 75830116-75830116 on Build GRCh38
rs983427172 Chr.17: 75830017-75830017 on Build GRCh38
rs997636090 Chr.17: 75830241-75830241 on Build GRCh38
rs949174966 Chr.17: 75830110-75830110 on Build GRCh38
rs377644361 Chr.17: 75830176-75830176 on Build GRCh38
rs118049905 Chr.17: 75830086-75830086 on Build GRCh38
rs113380644 Chr.17: 75830228-75830228 on Build GRCh38
rs201023196 Chr.17: 75830355-75830355 on Build GRCh38
rs867535106 Chr.17: 75830091-75830091 on Build GRCh38
rs970780021 Chr.17: 75830351-75830351 on Build GRCh38
rs546158627 Chr.17: 75830212-75830212 on Build GRCh38
rs1016012320 Chr.17: 75830302-75830302 on Build GRCh38
rs999144862 Chr.17: 75829916-75829916 on Build GRCh38
rs971355406 Chr.17: 75830214-75830214 on Build GRCh38
rs958601301 Chr.17: 75829936-75829936 on Build GRCh38
rs749341201 Chr.17: 75829979-75829979 on Build GRCh38
rs35628234 Chr.17: 75830039-75830039 on Build GRCh38
rs889317033 Chr.17: 75830059-75830059 on Build GRCh38
rs1030194494 Chr.17: 75830013-75830013 on Build GRCh38
rs762554409 Chr.17: 75830312-75830312 on Build GRCh38
rs778921171 Chr.17: 75830143-75830143 on Build GRCh38
rs368800716 Chr.17: 75829922-75829922 on Build GRCh38
rs369860531 Chr.17: 75830170-75830170 on Build GRCh38
rs772571668 Chr.17: 75829937-75829937 on Build GRCh38
rs2290769 Chr.17: 75830325-75830325 on Build GRCh38
rs1055031933 Chr.17: 75830050-75830050 on Build GRCh38
rs569582051 Chr.17: 75829991-75829991 on Build GRCh38
rs561798314 Chr.17: 75830276-75830276 on Build GRCh38

View Full Product Details