Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75890056-75890559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM65
Entrez Gene ID
201292
Gene Name
tripartite motif containing 65
Gene Aliases
4732463G12Rik
UniGene
Hs.189823
Gene Chromosome Location
Chr.17: 75880379-75897008 on Build GRCh38
Transcripts
NM_001256124.1 NM_173547.3 XM_006721760.3 XM_006721761.3 XM_011524499.2 XM_011524500.2 XM_011524501.2 XM_011524502.2 XM_017024344.1

SNP Detail

SNP ID Chromosome Location
rs779213015 Chr.17: 75890307-75890307 on Build GRCh38
rs76603306 Chr.17: 75890476-75890476 on Build GRCh38
rs370073665 Chr.17: 75890287-75890287 on Build GRCh38
rs981222940 Chr.17: 75890147-75890147 on Build GRCh38
rs1030855946 Chr.17: 75890378-75890378 on Build GRCh38
rs956547686 Chr.17: 75890430-75890430 on Build GRCh38
rs928489106 Chr.17: 75890163-75890163 on Build GRCh38
rs934089173 Chr.17: 75890493-75890493 on Build GRCh38
rs79120131 Chr.17: 75890451-75890451 on Build GRCh38
rs769016458 Chr.17: 75890215-75890215 on Build GRCh38
rs922749498 Chr.17: 75890161-75890161 on Build GRCh38
rs898818871 Chr.17: 75890311-75890311 on Build GRCh38
rs747350533 Chr.17: 75890418-75890418 on Build GRCh38
rs974788254 Chr.17: 75890508-75890508 on Build GRCh38
rs561624528 Chr.17: 75890457-75890457 on Build GRCh38
rs934121667 Chr.17: 75890168-75890168 on Build GRCh38
rs11554205 Chr.17: 75890385-75890385 on Build GRCh38
rs780434159 Chr.17: 75890341-75890341 on Build GRCh38
rs191589801 Chr.17: 75890516-75890516 on Build GRCh38
rs947040812 Chr.17: 75890244-75890244 on Build GRCh38
rs148341513 Chr.17: 75890270-75890270 on Build GRCh38
rs932856093 Chr.17: 75890523-75890523 on Build GRCh38
rs1023768786 Chr.17: 75890352-75890352 on Build GRCh38
rs988976472 Chr.17: 75890186-75890186 on Build GRCh38
rs891307551 Chr.17: 75890340-75890340 on Build GRCh38
rs540482343 Chr.17: 75890196-75890196 on Build GRCh38
rs982853746 Chr.17: 75890504-75890504 on Build GRCh38
rs969977566 Chr.17: 75890137-75890137 on Build GRCh38
rs934384808 Chr.17: 75890177-75890177 on Build GRCh38
rs572935973 Chr.17: 75890310-75890310 on Build GRCh38
rs1003351718 Chr.17: 75890377-75890377 on Build GRCh38
rs553095123 Chr.17: 75890458-75890458 on Build GRCh38
rs577282035 Chr.17: 75890368-75890368 on Build GRCh38
rs969925184 Chr.17: 75890399-75890399 on Build GRCh38
rs750985980 Chr.17: 75890212-75890212 on Build GRCh38
rs887330165 Chr.17: 75890306-75890306 on Build GRCh38
rs1016090611 Chr.17: 75890351-75890351 on Build GRCh38
rs141534491 Chr.17: 75890475-75890475 on Build GRCh38
rs914205619 Chr.17: 75890243-75890243 on Build GRCh38
rs1035456559 Chr.17: 75890450-75890450 on Build GRCh38
rs995901519 Chr.17: 75890362-75890362 on Build GRCh38
rs758901940 Chr.17: 75890229-75890229 on Build GRCh38

View Full Product Details