Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54188732-54189197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBD2
Entrez Gene ID
8932
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
DMTase, NY-CO-41
UniGene
Hs.25674
Gene Chromosome Location
Chr.18: 54151601-54224788 on Build GRCh38
Transcripts
NM_003927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341473 c.818G>A p.R273Q Chr.18: 54188896-54188896 on Build GRCh38
4700964 c.798_799insA p.V267fs*10 Chr.18: 54188915-54188916 on Build GRCh38
1680370 c.830A>C p.Q277P Chr.18: 54188884-54188884 on Build GRCh38
5388251 c.766G>C p.V256L Chr.18: 54188948-54188948 on Build GRCh38
6149397 c.703G>C p.G235R Chr.18: 54189011-54189011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758479176 Chr.18: 54189026-54189026 on Build GRCh38
rs903185321 Chr.18: 54189137-54189137 on Build GRCh38
rs777679228 Chr.18: 54189048-54189048 on Build GRCh38
rs200382180 Chr.18: 54188925-54188925 on Build GRCh38
rs778326530 Chr.18: 54188867-54188867 on Build GRCh38
rs376348912 Chr.18: 54189016-54189016 on Build GRCh38
rs761498673 Chr.18: 54189129-54189129 on Build GRCh38
rs200843117 Chr.18: 54188904-54188904 on Build GRCh38
rs747514913 Chr.18: 54188879-54188879 on Build GRCh38
rs969750614 Chr.18: 54188816-54188816 on Build GRCh38
rs1019595219 Chr.18: 54189109-54189109 on Build GRCh38
rs1023538431 Chr.18: 54188791-54188791 on Build GRCh38
rs77251753 Chr.18: 54188992-54188992 on Build GRCh38
rs111469458 Chr.18: 54188832-54188832 on Build GRCh38
rs761615321 Chr.18: 54188988-54188988 on Build GRCh38
rs898988315 Chr.18: 54188818-54188818 on Build GRCh38
rs755904678 Chr.18: 54189004-54189004 on Build GRCh38
rs550954262 Chr.18: 54188997-54188997 on Build GRCh38
rs768463202 Chr.18: 54189033-54189034 on Build GRCh38
rs755142224 Chr.18: 54188827-54188827 on Build GRCh38
rs772107709 Chr.18: 54188859-54188859 on Build GRCh38
rs771505040 Chr.18: 54188901-54188901 on Build GRCh38
rs771662925 Chr.18: 54188787-54188787 on Build GRCh38
rs769506412 Chr.18: 54188928-54188928 on Build GRCh38
rs941527339 Chr.18: 54189027-54189027 on Build GRCh38
rs530458078 Chr.18: 54189075-54189075 on Build GRCh38
rs752916688 Chr.18: 54189025-54189025 on Build GRCh38
rs562865980 Chr.18: 54189010-54189010 on Build GRCh38
rs996936118 Chr.18: 54189162-54189162 on Build GRCh38
rs201061908 Chr.18: 54188844-54188844 on Build GRCh38
rs199780930 Chr.18: 54188943-54188943 on Build GRCh38
rs762513492 Chr.18: 54188937-54188937 on Build GRCh38
rs867194627 Chr.18: 54188949-54188949 on Build GRCh38
rs976012147 Chr.18: 54189068-54189068 on Build GRCh38
rs775350753 Chr.18: 54188929-54188929 on Build GRCh38
rs747138343 Chr.18: 54189051-54189051 on Build GRCh38
rs538949604 Chr.18: 54189153-54189153 on Build GRCh38
rs774850786 Chr.18: 54189042-54189044 on Build GRCh38
rs373847404 Chr.18: 54189002-54189002 on Build GRCh38
rs750247920 Chr.18: 54188826-54188826 on Build GRCh38
rs540749358 Chr.18: 54188784-54188784 on Build GRCh38
rs921521303 Chr.18: 54189074-54189074 on Build GRCh38
rs751482492 Chr.18: 54188963-54188963 on Build GRCh38

View Full Product Details